Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157993195-157993701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIRREL
Entrez Gene ID
55243
Gene Name
kin of IRRE like (Drosophila)
Gene Aliases
NEPH1
UniGene
Hs.272234
Gene Chromosome Location
Chr.1: 157993273-158100262 on Build GRCh38
Transcripts
NM_001286349.1 NM_018240.6 XM_005245305.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1212522 c.16G>A p.V6I Chr.1: 157993692-157993692 on Build GRCh38
6215673 c.1A>G p.M1V Chr.1: 157993677-157993677 on Build GRCh38
6215672 c.1A>G p.M1V Chr.1: 157993677-157993677 on Build GRCh38
1212521 c.16G>A p.V6I Chr.1: 157993692-157993692 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs967734343 Chr.1: 157993524-157993524 on Build GRCh38
rs1045440544 Chr.1: 157993630-157993630 on Build GRCh38
rs375424709 Chr.1: 157993628-157993628 on Build GRCh38
rs956311997 Chr.1: 157993459-157993459 on Build GRCh38
rs564241954 Chr.1: 157993333-157993333 on Build GRCh38
rs531568299 Chr.1: 157993356-157993356 on Build GRCh38
rs768411475 Chr.1: 157993631-157993631 on Build GRCh38
rs535225713 Chr.1: 157993242-157993243 on Build GRCh38
rs953616937 Chr.1: 157993517-157993517 on Build GRCh38
rs1017679555 Chr.1: 157993411-157993411 on Build GRCh38
rs980967446 Chr.1: 157993534-157993534 on Build GRCh38
rs965826318 Chr.1: 157993462-157993462 on Build GRCh38
rs148023130 Chr.1: 157993573-157993580 on Build GRCh38
rs548115457 Chr.1: 157993634-157993634 on Build GRCh38
rs1008062842 Chr.1: 157993408-157993408 on Build GRCh38
rs561778153 Chr.1: 157993456-157993456 on Build GRCh38
rs985032251 Chr.1: 157993526-157993526 on Build GRCh38
rs1049545948 Chr.1: 157993385-157993385 on Build GRCh38
rs775233343 Chr.1: 157993407-157993407 on Build GRCh38
rs148283601 Chr.1: 157993243-157993244 on Build GRCh38
rs1032829067 Chr.1: 157993410-157993410 on Build GRCh38
rs559263033 Chr.1: 157993276-157993276 on Build GRCh38
rs926341090 Chr.1: 157993600-157993600 on Build GRCh38
rs566252539 Chr.1: 157993590-157993597 on Build GRCh38
rs961030404 Chr.1: 157993566-157993566 on Build GRCh38
rs566131297 Chr.1: 157993664-157993664 on Build GRCh38
rs1044787892 Chr.1: 157993219-157993219 on Build GRCh38
rs957099134 Chr.1: 157993243-157993243 on Build GRCh38
rs946461667 Chr.1: 157993217-157993217 on Build GRCh38
rs533580668 Chr.1: 157993673-157993673 on Build GRCh38
rs1022387130 Chr.1: 157993490-157993490 on Build GRCh38
rs919069195 Chr.1: 157993646-157993646 on Build GRCh38
rs545974281 Chr.1: 157993297-157993297 on Build GRCh38
rs997628756 Chr.1: 157993454-157993454 on Build GRCh38
rs1028629298 Chr.1: 157993480-157993481 on Build GRCh38
rs902044761 Chr.1: 157993444-157993444 on Build GRCh38
rs926478031 Chr.1: 157993332-157993332 on Build GRCh38
rs1045073699 Chr.1: 157993398-157993398 on Build GRCh38
rs905220074 Chr.1: 157993651-157993651 on Build GRCh38
rs932253953 Chr.1: 157993289-157993289 on Build GRCh38
rs949378144 Chr.1: 157993387-157993387 on Build GRCh38
rs939163163 Chr.1: 157993352-157993352 on Build GRCh38
rs917953646 Chr.1: 157993622-157993622 on Build GRCh38
rs896477409 Chr.1: 157993386-157993386 on Build GRCh38
rs969969087 Chr.1: 157993217-157993217 on Build GRCh38
rs984895864 Chr.1: 157993309-157993309 on Build GRCh38
rs990378731 Chr.1: 157993480-157993480 on Build GRCh38
rs73014860 Chr.1: 157993245-157993245 on Build GRCh38
rs929210403 Chr.1: 157993379-157993379 on Build GRCh38
rs909381914 Chr.1: 157993537-157993537 on Build GRCh38
rs181486539 Chr.1: 157993278-157993278 on Build GRCh38
rs867405613 Chr.1: 157993607-157993607 on Build GRCh38
rs1009982206 Chr.1: 157993451-157993451 on Build GRCh38
rs976131387 Chr.1: 157993479-157993479 on Build GRCh38
rs534277525 Chr.1: 157993620-157993620 on Build GRCh38
rs575329407 Chr.1: 157993377-157993377 on Build GRCh38

View Full Product Details