Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160547493-160548040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD84
Entrez Gene ID
8832
Gene Name
CD84 molecule
Gene Aliases
LY9B, SLAMF5, hCD84, mCD84
UniGene
Hs.398093
Gene Chromosome Location
Chr.1: 160541094-160579516 on Build GRCh38
Transcripts
NM_001184879.1 NM_001184881.1 NM_001184882.1 NM_003874.3 XM_011510094.2 XM_011510095.2

SNP Detail

SNP ID Chromosome Location
rs560435860 Chr.1: 160547695-160547695 on Build GRCh38
rs55672637 Chr.1: 160547792-160547792 on Build GRCh38
rs926290441 Chr.1: 160547999-160547999 on Build GRCh38
rs909913287 Chr.1: 160547823-160547823 on Build GRCh38
rs899687997 Chr.1: 160547570-160547570 on Build GRCh38
rs1034915621 Chr.1: 160547749-160547749 on Build GRCh38
rs562379061 Chr.1: 160547926-160547926 on Build GRCh38
rs759048125 Chr.1: 160547637-160547637 on Build GRCh38
rs963242251 Chr.1: 160547947-160547947 on Build GRCh38
rs61802607 Chr.1: 160547602-160547602 on Build GRCh38
rs573386272 Chr.1: 160547654-160547654 on Build GRCh38
rs146505028 Chr.1: 160547592-160547592 on Build GRCh38
rs748880809 Chr.1: 160547690-160547690 on Build GRCh38
rs941256136 Chr.1: 160547825-160547825 on Build GRCh38
rs921192517 Chr.1: 160547878-160547878 on Build GRCh38
rs915399202 Chr.1: 160547743-160547743 on Build GRCh38
rs998445388 Chr.1: 160547582-160547582 on Build GRCh38
rs879247568 Chr.1: 160547824-160547824 on Build GRCh38
rs547579841 Chr.1: 160547932-160547932 on Build GRCh38
rs527807693 Chr.1: 160547913-160547913 on Build GRCh38
rs892215330 Chr.1: 160547938-160547938 on Build GRCh38
rs374014864 Chr.1: 160547689-160547689 on Build GRCh38
rs970636439 Chr.1: 160547995-160547995 on Build GRCh38
rs190341339 Chr.1: 160547727-160547727 on Build GRCh38
rs1042615205 Chr.1: 160547768-160547768 on Build GRCh38
rs963574887 Chr.1: 160547612-160547612 on Build GRCh38
rs982744198 Chr.1: 160547814-160547814 on Build GRCh38
rs3766388 Chr.1: 160547689-160547689 on Build GRCh38
rs576458805 Chr.1: 160547665-160547665 on Build GRCh38
rs1050916158 Chr.1: 160547905-160547905 on Build GRCh38
rs769298589 Chr.1: 160547682-160547682 on Build GRCh38
rs757882751 Chr.1: 160547773-160547773 on Build GRCh38
rs970374819 Chr.1: 160547692-160547692 on Build GRCh38
rs969369818 Chr.1: 160547708-160547708 on Build GRCh38
rs986326600 Chr.1: 160547579-160547579 on Build GRCh38
rs1054334857 Chr.1: 160547742-160547742 on Build GRCh38
rs941321931 Chr.1: 160547552-160547552 on Build GRCh38
rs917297954 Chr.1: 160547787-160547787 on Build GRCh38
rs141119084 Chr.1: 160547860-160547860 on Build GRCh38
rs374692238 Chr.1: 160547738-160547738 on Build GRCh38
rs570750512 Chr.1: 160547933-160547933 on Build GRCh38
rs1049718213 Chr.1: 160547828-160547828 on Build GRCh38
rs993774229 Chr.1: 160547953-160547953 on Build GRCh38
rs1009355565 Chr.1: 160547980-160547980 on Build GRCh38
rs1039928474 Chr.1: 160547561-160547561 on Build GRCh38
rs992970621 Chr.1: 160547763-160547763 on Build GRCh38
rs11265434 Chr.1: 160547726-160547726 on Build GRCh38
rs540409818 Chr.1: 160547691-160547691 on Build GRCh38
rs1043401431 Chr.1: 160547997-160547997 on Build GRCh38
rs1055880 Chr.1: 160547902-160547902 on Build GRCh38
rs562644168 Chr.1: 160547750-160547750 on Build GRCh38

View Full Product Details