Product Details
- Primer Sequences
- Log in to view
- Primer Chromosome Location
-
Chr.7:
102640325-102640820
on Build GRCh38
View in UCSC browser
- Format
- Each primer will be delivered in a single tube, dried-down
- Quick Reference Guide
- Primer Designer™ Tool Quick Reference
- Ampliseq ID's
Gene Details
- Gene Symbol
- POLR2J2
- Entrez Gene ID
-
246721
- Gene Name
- RNA polymerase II subunit J2
- Gene Aliases
-
HRPB11B, RPB11b1
- UniGene
- Hs.610255
- Gene Chromosome Location
-
Chr.7:
102636748-102671729
on Build GRCh38
- Transcripts
-
NM_032959.5
NM_001114403.2
- Gene Symbol
- UPK3BL
- Entrez Gene ID
-
100134938
- Gene Name
- uroplakin 3B-like
- Gene Aliases
-
UPLP
- UniGene
- Hs.610255
- Gene Chromosome Location
-
Chr.7:
102637025-102642791
on Build GRCh38
- Transcripts
-
NM_032959.5
NM_001114403.2
Cosmic Detail
Cosmic ID |
Mutation |
AA |
Transcript Location |
3949667 |
c.344G>T |
p.G115V |
Chr.7:
102640345-102640345
on Build GRCh38
|
5426455 |
c.295C>A |
p.P99T |
Chr.7:
102640394-102640394
on Build GRCh38
|
SNP Detail
SNP ID |
Chromosome Location |
rs796333081
|
Chr.7:
102640800-102640800
on Build GRCh38
|
rs748395918
|
Chr.7:
102640754-102640754
on Build GRCh38
|
View Full Product Details