Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 112480919-112481429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSMEM1
Entrez Gene ID
286006
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
C7orf53
UniGene
Hs.396189
Gene Chromosome Location
Chr.7: 112480219-112491062 on Build GRCh38
Transcripts
NM_001134468.1 NM_182597.2 XM_011516075.2 XM_011516076.2

SNP Detail

SNP ID Chromosome Location
rs1016356482 Chr.7: 112481132-112481132 on Build GRCh38
rs955133863 Chr.7: 112481219-112481219 on Build GRCh38
rs775018962 Chr.7: 112481024-112481024 on Build GRCh38
rs975235661 Chr.7: 112481178-112481178 on Build GRCh38
rs1000505556 Chr.7: 112481076-112481076 on Build GRCh38
rs941060327 Chr.7: 112481287-112481295 on Build GRCh38
rs190445740 Chr.7: 112481324-112481324 on Build GRCh38
rs771524040 Chr.7: 112480974-112480974 on Build GRCh38
rs747767579 Chr.7: 112481293-112481294 on Build GRCh38
rs1010056220 Chr.7: 112481396-112481396 on Build GRCh38
rs922317633 Chr.7: 112481204-112481204 on Build GRCh38
rs986427595 Chr.7: 112481387-112481387 on Build GRCh38
rs1006351661 Chr.7: 112481131-112481131 on Build GRCh38
rs574111315 Chr.7: 112480981-112480981 on Build GRCh38
rs537164104 Chr.7: 112480972-112480972 on Build GRCh38
rs113001091 Chr.7: 112481286-112481286 on Build GRCh38
rs759631963 Chr.7: 112481077-112481077 on Build GRCh38
rs766109870 Chr.7: 112481291-112481293 on Build GRCh38
rs185635769 Chr.7: 112481265-112481265 on Build GRCh38
rs75748102 Chr.7: 112481341-112481341 on Build GRCh38
rs199798004 Chr.7: 112481294-112481294 on Build GRCh38
rs1002993872 Chr.7: 112481004-112481004 on Build GRCh38
rs772196370 Chr.7: 112481205-112481205 on Build GRCh38
rs1039012548 Chr.7: 112481290-112481290 on Build GRCh38
rs141244847 Chr.7: 112481283-112481286 on Build GRCh38
rs369045031 Chr.7: 112481290-112481291 on Build GRCh38
rs770387888 Chr.7: 112481184-112481184 on Build GRCh38
rs1034394459 Chr.7: 112481052-112481052 on Build GRCh38
rs982587048 Chr.7: 112481261-112481261 on Build GRCh38
rs1032960587 Chr.7: 112481103-112481103 on Build GRCh38
rs1041745116 Chr.7: 112480973-112480973 on Build GRCh38
rs765195872 Chr.7: 112480959-112480960 on Build GRCh38
rs139343566 Chr.7: 112481100-112481100 on Build GRCh38
rs555716375 Chr.7: 112480978-112480978 on Build GRCh38
rs374926291 Chr.7: 112481395-112481395 on Build GRCh38
rs748672303 Chr.7: 112480963-112480963 on Build GRCh38
rs938135546 Chr.7: 112481327-112481327 on Build GRCh38
rs1010547071 Chr.7: 112480955-112480955 on Build GRCh38
rs972705455 Chr.7: 112481245-112481245 on Build GRCh38
rs151147805 Chr.7: 112481294-112481297 on Build GRCh38

View Full Product Details