Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 124822245-124822740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POT1
Entrez Gene ID
25913
Gene Name
protection of telomeres 1
Gene Aliases
CMM10, GLM9, HPOT1
UniGene
Hs.31968
Gene Chromosome Location
Chr.7: 124822386-124929983 on Build GRCh38
Transcripts
NM_001042594.1 NM_015450.2 NR_003102.1 NR_003103.1 NR_003104.1 XM_006715917.3 XM_011516006.1 XM_011516007.1 XM_017011942.1

SNP Detail

SNP ID Chromosome Location
rs17246404 Chr.7: 124822607-124822607 on Build GRCh38
rs529937671 Chr.7: 124822695-124822695 on Build GRCh38
rs751168958 Chr.7: 124822541-124822541 on Build GRCh38
rs886854290 Chr.7: 124822326-124822326 on Build GRCh38
rs1000978344 Chr.7: 124822715-124822715 on Build GRCh38
rs1051493054 Chr.7: 124822619-124822619 on Build GRCh38
rs993683402 Chr.7: 124822645-124822645 on Build GRCh38
rs567700636 Chr.7: 124822615-124822615 on Build GRCh38
rs572841207 Chr.7: 124822351-124822351 on Build GRCh38
rs992841156 Chr.7: 124822514-124822514 on Build GRCh38
rs148739261 Chr.7: 124822334-124822335 on Build GRCh38
rs144505427 Chr.7: 124822590-124822590 on Build GRCh38
rs528668733 Chr.7: 124822546-124822546 on Build GRCh38
rs958928070 Chr.7: 124822300-124822300 on Build GRCh38
rs544417496 Chr.7: 124822717-124822717 on Build GRCh38
rs1020628817 Chr.7: 124822276-124822276 on Build GRCh38
rs543350821 Chr.7: 124822443-124822443 on Build GRCh38
rs759868783 Chr.7: 124822497-124822497 on Build GRCh38
rs903605135 Chr.7: 124822289-124822289 on Build GRCh38
rs1033199711 Chr.7: 124822297-124822297 on Build GRCh38
rs76436625 Chr.7: 124822601-124822601 on Build GRCh38
rs1020775985 Chr.7: 124822668-124822668 on Build GRCh38
rs1009406124 Chr.7: 124822666-124822666 on Build GRCh38
rs574227671 Chr.7: 124822389-124822389 on Build GRCh38
rs544668410 Chr.7: 124822563-124822563 on Build GRCh38
rs945234822 Chr.7: 124822499-124822499 on Build GRCh38
rs919225924 Chr.7: 124822383-124822383 on Build GRCh38
rs761372337 Chr.7: 124822544-124822544 on Build GRCh38
rs772836291 Chr.7: 124822305-124822305 on Build GRCh38
rs187584000 Chr.7: 124822491-124822491 on Build GRCh38
rs926580905 Chr.7: 124822474-124822474 on Build GRCh38
rs759675051 Chr.7: 124822620-124822620 on Build GRCh38
rs575148932 Chr.7: 124822338-124822338 on Build GRCh38
rs370988165 Chr.7: 124822517-124822517 on Build GRCh38
rs979444353 Chr.7: 124822468-124822468 on Build GRCh38
rs929501853 Chr.7: 124822520-124822520 on Build GRCh38
rs918107521 Chr.7: 124822292-124822292 on Build GRCh38
rs75558173 Chr.7: 124822337-124822338 on Build GRCh38
rs192260864 Chr.7: 124822530-124822530 on Build GRCh38
rs554583130 Chr.7: 124822369-124822369 on Build GRCh38
rs1000581343 Chr.7: 124822295-124822295 on Build GRCh38
rs765610244 Chr.7: 124822528-124822528 on Build GRCh38
rs1054913577 Chr.7: 124822596-124822596 on Build GRCh38
rs971681879 Chr.7: 124822629-124822629 on Build GRCh38
rs545050832 Chr.7: 124822568-124822568 on Build GRCh38
rs182894575 Chr.7: 124822340-124822340 on Build GRCh38
rs1046834559 Chr.7: 124822307-124822307 on Build GRCh38
rs147972124 Chr.7: 124822338-124822339 on Build GRCh38
rs62478915 Chr.7: 124822703-124822703 on Build GRCh38
rs1026290363 Chr.7: 124822681-124822681 on Build GRCh38
rs984816723 Chr.7: 124822426-124822426 on Build GRCh38
rs930742899 Chr.7: 124822405-124822405 on Build GRCh38
rs537858943 Chr.7: 124822455-124822455 on Build GRCh38
rs76846113 Chr.7: 124822339-124822339 on Build GRCh38
rs943245204 Chr.7: 124822454-124822454 on Build GRCh38
rs749845454 Chr.7: 124822697-124822697 on Build GRCh38

View Full Product Details