Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127373256-127373756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF800
Entrez Gene ID
168850
Gene Name
zinc finger protein 800
Gene Aliases
-
UniGene
Hs.159006
Gene Chromosome Location
Chr.7: 127342871-127392803 on Build GRCh38
Transcripts
NM_176814.4 XM_005250181.3 XM_006715867.3 XM_006715868.3 XM_006715869.3 XM_011515859.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7120753 c.1822G>A p.E608K Chr.7: 127373514-127373514 on Build GRCh38
7322794 c.1870G>T p.G624* Chr.7: 127373466-127373466 on Build GRCh38
5871736 c.1942C>T p.P648S Chr.7: 127373394-127373394 on Build GRCh38
5997418 c.1622T>G p.V541G Chr.7: 127373714-127373714 on Build GRCh38
6833238 c.1882G>T p.A628S Chr.7: 127373454-127373454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776136075 Chr.7: 127373724-127373724 on Build GRCh38
rs774573647 Chr.7: 127373371-127373371 on Build GRCh38
rs140198904 Chr.7: 127373585-127373585 on Build GRCh38
rs144304458 Chr.7: 127373677-127373678 on Build GRCh38
rs754758617 Chr.7: 127373459-127373459 on Build GRCh38
rs749644328 Chr.7: 127373659-127373659 on Build GRCh38
rs778626181 Chr.7: 127373556-127373556 on Build GRCh38
rs540077251 Chr.7: 127373364-127373364 on Build GRCh38
rs754952182 Chr.7: 127373554-127373554 on Build GRCh38
rs772201913 Chr.7: 127373482-127373482 on Build GRCh38
rs745786731 Chr.7: 127373339-127373339 on Build GRCh38
rs771369916 Chr.7: 127373405-127373405 on Build GRCh38
rs761597629 Chr.7: 127373432-127373432 on Build GRCh38
rs767031632 Chr.7: 127373441-127373441 on Build GRCh38
rs544145908 Chr.7: 127373491-127373491 on Build GRCh38
rs760412847 Chr.7: 127373679-127373679 on Build GRCh38
rs756054944 Chr.7: 127373353-127373353 on Build GRCh38
rs746364506 Chr.7: 127373619-127373619 on Build GRCh38
rs758608925 Chr.7: 127373590-127373590 on Build GRCh38
rs754172798 Chr.7: 127373305-127373305 on Build GRCh38
rs765753904 Chr.7: 127373418-127373418 on Build GRCh38
rs772139812 Chr.7: 127373389-127373389 on Build GRCh38
rs948638473 Chr.7: 127373285-127373285 on Build GRCh38
rs761122716 Chr.7: 127373300-127373300 on Build GRCh38
rs774132118 Chr.7: 127373304-127373304 on Build GRCh38
rs775929057 Chr.7: 127373647-127373647 on Build GRCh38
rs758156176 Chr.7: 127373286-127373289 on Build GRCh38
rs751252192 Chr.7: 127373426-127373426 on Build GRCh38
rs554318069 Chr.7: 127373284-127373284 on Build GRCh38
rs1040458600 Chr.7: 127373643-127373643 on Build GRCh38
rs180822910 Chr.7: 127373618-127373618 on Build GRCh38
rs374406949 Chr.7: 127373367-127373367 on Build GRCh38
rs777256124 Chr.7: 127373303-127373304 on Build GRCh38
rs527854334 Chr.7: 127373602-127373602 on Build GRCh38
rs759406657 Chr.7: 127373728-127373728 on Build GRCh38
rs745538075 Chr.7: 127373303-127373303 on Build GRCh38
rs1049805866 Chr.7: 127373396-127373396 on Build GRCh38
rs760186657 Chr.7: 127373512-127373512 on Build GRCh38
rs370009019 Chr.7: 127373442-127373442 on Build GRCh38
rs917134675 Chr.7: 127373285-127373286 on Build GRCh38
rs566464280 Chr.7: 127373549-127373549 on Build GRCh38
rs749883086 Chr.7: 127373530-127373530 on Build GRCh38
rs751054156 Chr.7: 127373312-127373312 on Build GRCh38
rs770436899 Chr.7: 127373632-127373632 on Build GRCh38
rs938016581 Chr.7: 127373595-127373595 on Build GRCh38
rs749924711 Chr.7: 127373444-127373444 on Build GRCh38
rs200602569 Chr.7: 127373450-127373450 on Build GRCh38
rs770885574 Chr.7: 127373603-127373603 on Build GRCh38
rs758081373 Chr.7: 127373307-127373307 on Build GRCh38
rs779024867 Chr.7: 127373321-127373321 on Build GRCh38
rs561952635 Chr.7: 127373399-127373399 on Build GRCh38
rs746557903 Chr.7: 127373310-127373310 on Build GRCh38
rs770252626 Chr.7: 127373520-127373520 on Build GRCh38
rs749115583 Chr.7: 127373366-127373366 on Build GRCh38
rs961232177 Chr.7: 127373283-127373283 on Build GRCh38
rs201097436 Chr.7: 127373310-127373310 on Build GRCh38
rs756812736 Chr.7: 127373315-127373315 on Build GRCh38
rs768803409 Chr.7: 127373370-127373370 on Build GRCh38
rs902663412 Chr.7: 127373676-127373676 on Build GRCh38
rs913287247 Chr.7: 127373550-127373550 on Build GRCh38
rs765975561 Chr.7: 127373543-127373543 on Build GRCh38
rs752496744 Chr.7: 127373567-127373567 on Build GRCh38
rs748343975 Chr.7: 127373479-127373479 on Build GRCh38
rs867664985 Chr.7: 127373414-127373414 on Build GRCh38
rs761191819 Chr.7: 127373400-127373400 on Build GRCh38
rs776985561 Chr.7: 127373416-127373416 on Build GRCh38
rs771409270 Chr.7: 127373499-127373499 on Build GRCh38
rs373818543 Chr.7: 127373451-127373451 on Build GRCh38
rs767107811 Chr.7: 127373524-127373524 on Build GRCh38
rs888109796 Chr.7: 127373409-127373409 on Build GRCh38
rs772715607 Chr.7: 127373671-127373671 on Build GRCh38
rs983292287 Chr.7: 127373345-127373345 on Build GRCh38
rs551012942 Chr.7: 127373533-127373533 on Build GRCh38
rs766741002 Chr.7: 127373301-127373301 on Build GRCh38
rs775750914 Chr.7: 127373627-127373627 on Build GRCh38
rs200843686 Chr.7: 127373286-127373286 on Build GRCh38
rs1014075786 Chr.7: 127373465-127373465 on Build GRCh38
rs567879157 Chr.7: 127373716-127373716 on Build GRCh38
rs76411607 Chr.7: 127373521-127373521 on Build GRCh38
rs535286073 Chr.7: 127373383-127373383 on Build GRCh38
rs900739484 Chr.7: 127373322-127373323 on Build GRCh38
rs562133327 Chr.7: 127373511-127373511 on Build GRCh38
rs1009593504 Chr.7: 127373693-127373693 on Build GRCh38
rs753574901 Chr.7: 127373458-127373458 on Build GRCh38
rs770455761 Chr.7: 127373328-127373329 on Build GRCh38
rs780633277 Chr.7: 127373332-127373332 on Build GRCh38
rs761509560 Chr.7: 127373523-127373523 on Build GRCh38
rs778750764 Chr.7: 127373460-127373460 on Build GRCh38
rs771703793 Chr.7: 127373665-127373665 on Build GRCh38
rs748430315 Chr.7: 127373381-127373381 on Build GRCh38

View Full Product Details