Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151187749-151188256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB10
Entrez Gene ID
136371
Gene Name
ankyrin repeat and SOCS box containing 10
Gene Aliases
GLC1F
UniGene
Hs.647081
Gene Chromosome Location
Chr.7: 151175698-151187832 on Build GRCh38
Transcripts
NM_080871.3

SNP Detail

SNP ID Chromosome Location
rs903830612 Chr.7: 151188069-151188069 on Build GRCh38
rs532988684 Chr.7: 151188215-151188215 on Build GRCh38
rs976650374 Chr.7: 151188162-151188167 on Build GRCh38
rs952133738 Chr.7: 151187863-151187863 on Build GRCh38
rs965030801 Chr.7: 151188151-151188151 on Build GRCh38
rs1026795358 Chr.7: 151188059-151188059 on Build GRCh38
rs927043514 Chr.7: 151188187-151188187 on Build GRCh38
rs543467168 Chr.7: 151188231-151188231 on Build GRCh38
rs534229411 Chr.7: 151188016-151188016 on Build GRCh38
rs868471299 Chr.7: 151188065-151188065 on Build GRCh38
rs976060801 Chr.7: 151188124-151188124 on Build GRCh38
rs116644649 Chr.7: 151187857-151187857 on Build GRCh38
rs947552067 Chr.7: 151188141-151188141 on Build GRCh38
rs186881820 Chr.7: 151188058-151188058 on Build GRCh38
rs1012779948 Chr.7: 151187918-151187918 on Build GRCh38
rs568698047 Chr.7: 151187894-151187894 on Build GRCh38
rs897061202 Chr.7: 151188051-151188051 on Build GRCh38
rs935500835 Chr.7: 151187881-151187897 on Build GRCh38
rs754530981 Chr.7: 151187795-151187795 on Build GRCh38
rs976113280 Chr.7: 151187912-151187912 on Build GRCh38
rs752618849 Chr.7: 151187944-151187944 on Build GRCh38
rs564115272 Chr.7: 151187799-151187799 on Build GRCh38
rs914708845 Chr.7: 151188140-151188140 on Build GRCh38
rs907054200 Chr.7: 151188113-151188113 on Build GRCh38
rs777581712 Chr.7: 151188134-151188134 on Build GRCh38
rs943569016 Chr.7: 151187828-151187828 on Build GRCh38
rs761342620 Chr.7: 151187892-151187892 on Build GRCh38
rs951687263 Chr.7: 151188123-151188123 on Build GRCh38
rs918270118 Chr.7: 151188220-151188220 on Build GRCh38
rs964714866 Chr.7: 151187909-151187909 on Build GRCh38
rs1024175217 Chr.7: 151188112-151188112 on Build GRCh38
rs923349714 Chr.7: 151188049-151188049 on Build GRCh38
rs548513091 Chr.7: 151187893-151187893 on Build GRCh38
rs190175152 Chr.7: 151188162-151188162 on Build GRCh38
rs1013161738 Chr.7: 151188106-151188106 on Build GRCh38
rs374738713 Chr.7: 151187811-151187811 on Build GRCh38
rs549938943 Chr.7: 151187817-151187817 on Build GRCh38
rs1044542864 Chr.7: 151187997-151187997 on Build GRCh38
rs1006270667 Chr.7: 151188085-151188085 on Build GRCh38
rs919505839 Chr.7: 151187770-151187770 on Build GRCh38
rs1035581114 Chr.7: 151187803-151187803 on Build GRCh38
rs533052241 Chr.7: 151187813-151187813 on Build GRCh38
rs947135293 Chr.7: 151187995-151187995 on Build GRCh38
rs923512277 Chr.7: 151187914-151187914 on Build GRCh38
rs77953646 Chr.7: 151188119-151188119 on Build GRCh38
rs925386775 Chr.7: 151188226-151188226 on Build GRCh38
rs73478660 Chr.7: 151188211-151188211 on Build GRCh38
rs980332217 Chr.7: 151188143-151188143 on Build GRCh38
rs945544253 Chr.7: 151188208-151188208 on Build GRCh38
rs1030363979 Chr.7: 151188125-151188125 on Build GRCh38
rs535616251 Chr.7: 151187941-151187941 on Build GRCh38
rs989488009 Chr.7: 151188137-151188137 on Build GRCh38
rs73727105 Chr.7: 151188017-151188017 on Build GRCh38
rs910673208 Chr.7: 151187826-151187826 on Build GRCh38
rs541964128 Chr.7: 151188118-151188118 on Build GRCh38
rs569455610 Chr.7: 151187827-151187827 on Build GRCh38
rs894415488 Chr.7: 151188095-151188095 on Build GRCh38
rs988979347 Chr.7: 151187934-151187934 on Build GRCh38
rs914740360 Chr.7: 151187940-151187940 on Build GRCh38
rs556812661 Chr.7: 151188022-151188022 on Build GRCh38
rs918890419 Chr.7: 151187942-151187942 on Build GRCh38
rs934922784 Chr.7: 151187920-151187920 on Build GRCh38
rs1049309659 Chr.7: 151187804-151187804 on Build GRCh38
rs943522195 Chr.7: 151188024-151188024 on Build GRCh38
rs888279130 Chr.7: 151188082-151188082 on Build GRCh38
rs74440219 Chr.7: 151188205-151188205 on Build GRCh38
rs556211461 Chr.7: 151188025-151188025 on Build GRCh38
rs1040833260 Chr.7: 151187837-151187837 on Build GRCh38

View Full Product Details