Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 152100656-152101165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT11
Entrez Gene ID
63917
Gene Name
polypeptide N-acetylgalactosaminyltransferase 11
Gene Aliases
GALNAC-T11, GALNACT11
UniGene
Hs.647109
Gene Chromosome Location
Chr.7: 152025674-152122347 on Build GRCh38
Transcripts
NM_001304514.1 NM_022087.3 XM_006716082.2 XM_006716083.2 XM_006716084.2 XM_017012499.1 XM_017012500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6227905 c.314G>A p.R105H Chr.7: 152100816-152100816 on Build GRCh38
3636444 c.375C>T p.D125D Chr.7: 152100877-152100877 on Build GRCh38
1449372 c.310G>T p.E104* Chr.7: 152100812-152100812 on Build GRCh38
5014766 c.411G>T p.R137S Chr.7: 152100913-152100913 on Build GRCh38
7287767 c.306T>C p.F102F Chr.7: 152100808-152100808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998229728 Chr.7: 152100988-152100988 on Build GRCh38
rs376537764 Chr.7: 152100872-152100872 on Build GRCh38
rs761842320 Chr.7: 152100765-152100765 on Build GRCh38
rs994374201 Chr.7: 152101023-152101023 on Build GRCh38
rs139294517 Chr.7: 152100817-152100817 on Build GRCh38
rs752521339 Chr.7: 152101122-152101122 on Build GRCh38
rs978654864 Chr.7: 152101068-152101068 on Build GRCh38
rs1044763417 Chr.7: 152100906-152100906 on Build GRCh38
rs373357941 Chr.7: 152100859-152100859 on Build GRCh38
rs74427639 Chr.7: 152100816-152100816 on Build GRCh38
rs149600575 Chr.7: 152100818-152100818 on Build GRCh38
rs771556840 Chr.7: 152100790-152100790 on Build GRCh38
rs943998246 Chr.7: 152100723-152100723 on Build GRCh38
rs897372522 Chr.7: 152100858-152100858 on Build GRCh38
rs763723817 Chr.7: 152100962-152100962 on Build GRCh38
rs770002172 Chr.7: 152100936-152100936 on Build GRCh38
rs990865823 Chr.7: 152101124-152101124 on Build GRCh38
rs751066614 Chr.7: 152100838-152100838 on Build GRCh38
rs573200341 Chr.7: 152100928-152100928 on Build GRCh38
rs555262229 Chr.7: 152100750-152100752 on Build GRCh38
rs952797461 Chr.7: 152101075-152101075 on Build GRCh38
rs199513189 Chr.7: 152100892-152100892 on Build GRCh38
rs775895140 Chr.7: 152100954-152100954 on Build GRCh38
rs563431288 Chr.7: 152101088-152101088 on Build GRCh38
rs560398003 Chr.7: 152100781-152100784 on Build GRCh38
rs1018004507 Chr.7: 152100935-152100935 on Build GRCh38
rs967680792 Chr.7: 152101015-152101015 on Build GRCh38
rs911195247 Chr.7: 152100713-152100713 on Build GRCh38
rs755245810 Chr.7: 152100750-152100750 on Build GRCh38
rs367881769 Chr.7: 152100995-152100995 on Build GRCh38
rs1006638765 Chr.7: 152100934-152100934 on Build GRCh38
rs201149772 Chr.7: 152100773-152100773 on Build GRCh38
rs767384306 Chr.7: 152100863-152100863 on Build GRCh38
rs370112474 Chr.7: 152100881-152100881 on Build GRCh38
rs760903182 Chr.7: 152100921-152100921 on Build GRCh38
rs376988569 Chr.7: 152100966-152100966 on Build GRCh38
rs761250556 Chr.7: 152100963-152100963 on Build GRCh38
rs750240669 Chr.7: 152100866-152100866 on Build GRCh38
rs978986611 Chr.7: 152100769-152100769 on Build GRCh38
rs773367690 Chr.7: 152100849-152100849 on Build GRCh38
rs762662352 Chr.7: 152100960-152100960 on Build GRCh38
rs200711472 Chr.7: 152100888-152100888 on Build GRCh38
rs1027153834 Chr.7: 152101065-152101065 on Build GRCh38
rs748294582 Chr.7: 152100767-152100767 on Build GRCh38
rs111896957 Chr.7: 152100999-152100999 on Build GRCh38
rs779220999 Chr.7: 152100878-152100878 on Build GRCh38
rs748371084 Chr.7: 152100879-152100879 on Build GRCh38
rs200793325 Chr.7: 152100772-152100772 on Build GRCh38
rs748355673 Chr.7: 152100910-152100910 on Build GRCh38
rs911239694 Chr.7: 152101118-152101118 on Build GRCh38
rs759761432 Chr.7: 152100815-152100815 on Build GRCh38
rs369630394 Chr.7: 152100793-152100793 on Build GRCh38
rs140670959 Chr.7: 152100916-152100916 on Build GRCh38
rs763635935 Chr.7: 152100831-152100831 on Build GRCh38
rs747556230 Chr.7: 152100787-152100787 on Build GRCh38
rs1041336394 Chr.7: 152100737-152100737 on Build GRCh38
rs28473773 Chr.7: 152100738-152100738 on Build GRCh38
rs147242850 Chr.7: 152100828-152100828 on Build GRCh38
rs771577564 Chr.7: 152100912-152100912 on Build GRCh38

View Full Product Details