Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160997280-160997787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F11R
Entrez Gene ID
50848
Gene Name
F11 receptor
Gene Aliases
CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1
UniGene
Hs.517293
Gene Chromosome Location
Chr.1: 160995211-161021343 on Build GRCh38
Transcripts
NM_016946.4

SNP Detail

SNP ID Chromosome Location
rs1006417101 Chr.1: 160997569-160997569 on Build GRCh38
rs549341014 Chr.1: 160997547-160997547 on Build GRCh38
rs1022314118 Chr.1: 160997415-160997415 on Build GRCh38
rs144948407 Chr.1: 160997719-160997719 on Build GRCh38
rs962267730 Chr.1: 160997353-160997353 on Build GRCh38
rs925312815 Chr.1: 160997413-160997413 on Build GRCh38
rs969434200 Chr.1: 160997520-160997520 on Build GRCh38
rs532784318 Chr.1: 160997516-160997516 on Build GRCh38
rs1025563995 Chr.1: 160997614-160997614 on Build GRCh38
rs980899951 Chr.1: 160997551-160997551 on Build GRCh38
rs925260460 Chr.1: 160997639-160997639 on Build GRCh38
rs913120490 Chr.1: 160997747-160997748 on Build GRCh38
rs11542268 Chr.1: 160997325-160997325 on Build GRCh38
rs1048015203 Chr.1: 160997696-160997696 on Build GRCh38
rs992634993 Chr.1: 160997365-160997365 on Build GRCh38
rs942430051 Chr.1: 160997750-160997750 on Build GRCh38
rs12565932 Chr.1: 160997665-160997665 on Build GRCh38
rs149893770 Chr.1: 160997661-160997661 on Build GRCh38
rs901568366 Chr.1: 160997516-160997517 on Build GRCh38
rs1040625543 Chr.1: 160997513-160997513 on Build GRCh38
rs1013719624 Chr.1: 160997610-160997610 on Build GRCh38
rs956690041 Chr.1: 160997381-160997381 on Build GRCh38
rs917977429 Chr.1: 160997396-160997399 on Build GRCh38
rs545307903 Chr.1: 160997398-160997398 on Build GRCh38
rs565738788 Chr.1: 160997711-160997711 on Build GRCh38
rs972715360 Chr.1: 160997571-160997571 on Build GRCh38
rs773359086 Chr.1: 160997452-160997452 on Build GRCh38
rs930897901 Chr.1: 160997681-160997681 on Build GRCh38
rs190273556 Chr.1: 160997567-160997567 on Build GRCh38
rs998491494 Chr.1: 160997334-160997334 on Build GRCh38
rs962619708 Chr.1: 160997605-160997605 on Build GRCh38
rs565207228 Chr.1: 160997449-160997449 on Build GRCh38
rs948124165 Chr.1: 160997398-160997399 on Build GRCh38
rs980727083 Chr.1: 160997410-160997410 on Build GRCh38
rs928106135 Chr.1: 160997561-160997561 on Build GRCh38
rs34789796 Chr.1: 160997384-160997384 on Build GRCh38
rs545608497 Chr.1: 160997695-160997695 on Build GRCh38
rs763156582 Chr.1: 160997576-160997576 on Build GRCh38
rs945878750 Chr.1: 160997480-160997480 on Build GRCh38
rs185746191 Chr.1: 160997344-160997344 on Build GRCh38
rs770212502 Chr.1: 160997748-160997748 on Build GRCh38
rs969455715 Chr.1: 160997615-160997615 on Build GRCh38
rs1015201184 Chr.1: 160997575-160997575 on Build GRCh38
rs980886292 Chr.1: 160997625-160997625 on Build GRCh38

View Full Product Details