Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167126074-167126562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP27
Entrez Gene ID
92235
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
-
UniGene
Hs.632462
Gene Chromosome Location
Chr.1: 167094048-167129165 on Build GRCh38
Transcripts
NM_001080426.1 XM_005245603.3 XM_011510146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
676459 c.1020C>T p.T340T Chr.1: 167126151-167126151 on Build GRCh38
7211400 c.1042_1044delGAG p.E348delE Chr.1: 167126173-167126175 on Build GRCh38
4024783 c.1334G>A p.S445N Chr.1: 167126465-167126465 on Build GRCh38
6122373 c.1384C>A p.H462N Chr.1: 167126515-167126515 on Build GRCh38
1317633 c.1030_1032delGAG p.E348delE Chr.1: 167126161-167126163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751835407 Chr.1: 167126280-167126280 on Build GRCh38
rs139389530 Chr.1: 167126481-167126481 on Build GRCh38
rs759073443 Chr.1: 167126175-167126175 on Build GRCh38
rs142057898 Chr.1: 167126249-167126249 on Build GRCh38
rs899547238 Chr.1: 167126521-167126521 on Build GRCh38
rs201292191 Chr.1: 167126351-167126351 on Build GRCh38
rs760051518 Chr.1: 167126504-167126504 on Build GRCh38
rs145405415 Chr.1: 167126398-167126398 on Build GRCh38
rs1022400914 Chr.1: 167126106-167126106 on Build GRCh38
rs762804909 Chr.1: 167126163-167126163 on Build GRCh38
rs747123131 Chr.1: 167126105-167126105 on Build GRCh38
rs760150269 Chr.1: 167126385-167126385 on Build GRCh38
rs369055427 Chr.1: 167126156-167126156 on Build GRCh38
rs780471336 Chr.1: 167126204-167126204 on Build GRCh38
rs936556006 Chr.1: 167126289-167126289 on Build GRCh38
rs777847059 Chr.1: 167126420-167126420 on Build GRCh38
rs781362677 Chr.1: 167126219-167126219 on Build GRCh38
rs768833604 Chr.1: 167126113-167126113 on Build GRCh38
rs138628687 Chr.1: 167126337-167126337 on Build GRCh38
rs922481006 Chr.1: 167126510-167126510 on Build GRCh38
rs373216642 Chr.1: 167126160-167126160 on Build GRCh38
rs768421904 Chr.1: 167126488-167126488 on Build GRCh38
rs1041155513 Chr.1: 167126357-167126357 on Build GRCh38
rs759998805 Chr.1: 167126447-167126447 on Build GRCh38
rs756297966 Chr.1: 167126457-167126457 on Build GRCh38
rs376866922 Chr.1: 167126460-167126460 on Build GRCh38
rs140692288 Chr.1: 167126327-167126327 on Build GRCh38
rs199616293 Chr.1: 167126264-167126264 on Build GRCh38
rs778873469 Chr.1: 167126461-167126461 on Build GRCh38
rs6668826 Chr.1: 167126528-167126528 on Build GRCh38
rs772332757 Chr.1: 167126506-167126506 on Build GRCh38
rs778631763 Chr.1: 167126492-167126492 on Build GRCh38
rs775910849 Chr.1: 167126310-167126310 on Build GRCh38
rs181149914 Chr.1: 167126326-167126326 on Build GRCh38
rs761906549 Chr.1: 167126512-167126512 on Build GRCh38
rs151182288 Chr.1: 167126421-167126421 on Build GRCh38
rs920414811 Chr.1: 167126272-167126272 on Build GRCh38
rs545663733 Chr.1: 167126422-167126422 on Build GRCh38
rs755310428 Chr.1: 167126295-167126295 on Build GRCh38
rs775976978 Chr.1: 167126507-167126507 on Build GRCh38
rs769688201 Chr.1: 167126226-167126226 on Build GRCh38
rs752917323 Chr.1: 167126408-167126408 on Build GRCh38
rs770721361 Chr.1: 167126331-167126331 on Build GRCh38
rs781317499 Chr.1: 167126368-167126369 on Build GRCh38
rs766979136 Chr.1: 167126180-167126180 on Build GRCh38
rs879321156 Chr.1: 167126353-167126353 on Build GRCh38
rs964207778 Chr.1: 167126116-167126116 on Build GRCh38
rs754443154 Chr.1: 167126476-167126476 on Build GRCh38
rs760225047 Chr.1: 167126248-167126248 on Build GRCh38
rs758855328 Chr.1: 167126270-167126270 on Build GRCh38
rs754706168 Chr.1: 167126531-167126531 on Build GRCh38
rs544611272 Chr.1: 167126502-167126502 on Build GRCh38
rs746654567 Chr.1: 167126483-167126483 on Build GRCh38
rs900930229 Chr.1: 167126374-167126374 on Build GRCh38
rs199963325 Chr.1: 167126092-167126092 on Build GRCh38
rs774028355 Chr.1: 167126232-167126232 on Build GRCh38
rs778897145 Chr.1: 167126167-167126167 on Build GRCh38
rs748091814 Chr.1: 167126370-167126372 on Build GRCh38
rs745646399 Chr.1: 167126341-167126341 on Build GRCh38
rs373269072 Chr.1: 167126391-167126391 on Build GRCh38
rs781560890 Chr.1: 167126296-167126296 on Build GRCh38
rs376387120 Chr.1: 167126392-167126392 on Build GRCh38
rs373488300 Chr.1: 167126445-167126445 on Build GRCh38
rs962263097 Chr.1: 167126227-167126227 on Build GRCh38
rs758159742 Chr.1: 167126469-167126469 on Build GRCh38
rs1006466176 Chr.1: 167126206-167126206 on Build GRCh38
rs762708373 Chr.1: 167126228-167126228 on Build GRCh38
rs527890632 Chr.1: 167126539-167126539 on Build GRCh38
rs535432607 Chr.1: 167126373-167126373 on Build GRCh38
rs750313262 Chr.1: 167126464-167126464 on Build GRCh38
rs140256913 Chr.1: 167126101-167126101 on Build GRCh38
rs372437301 Chr.1: 167126161-167126163 on Build GRCh38
rs754343401 Chr.1: 167126184-167126184 on Build GRCh38
rs866594794 Chr.1: 167126340-167126340 on Build GRCh38
rs758757651 Chr.1: 167126201-167126201 on Build GRCh38
rs1038320391 Chr.1: 167126529-167126529 on Build GRCh38
rs200815707 Chr.1: 167126434-167126434 on Build GRCh38
rs757644272 Chr.1: 167126185-167126185 on Build GRCh38
rs1043488724 Chr.1: 167126383-167126383 on Build GRCh38
rs201991477 Chr.1: 167126183-167126183 on Build GRCh38
rs747760393 Chr.1: 167126533-167126533 on Build GRCh38
rs776148882 Chr.1: 167126437-167126437 on Build GRCh38
rs113517663 Chr.1: 167126255-167126255 on Build GRCh38
rs767024238 Chr.1: 167126235-167126235 on Build GRCh38
rs976062812 Chr.1: 167126151-167126151 on Build GRCh38
rs892153799 Chr.1: 167126327-167126328 on Build GRCh38
rs149064504 Chr.1: 167126541-167126541 on Build GRCh38
rs756182069 Chr.1: 167126416-167126416 on Build GRCh38
rs769377705 Chr.1: 167126499-167126499 on Build GRCh38
rs200522320 Chr.1: 167126234-167126234 on Build GRCh38
rs146957255 Chr.1: 167126244-167126244 on Build GRCh38
rs767683915 Chr.1: 167126405-167126405 on Build GRCh38
rs374350764 Chr.1: 167126120-167126120 on Build GRCh38
rs756467892 Chr.1: 167126425-167126425 on Build GRCh38
rs776731321 Chr.1: 167126119-167126119 on Build GRCh38
rs1045386845 Chr.1: 167126290-167126290 on Build GRCh38
rs147722091 Chr.1: 167126208-167126208 on Build GRCh38
rs926598511 Chr.1: 167126468-167126468 on Build GRCh38
rs772938358 Chr.1: 167126162-167126162 on Build GRCh38
rs892185013 Chr.1: 167126097-167126097 on Build GRCh38
rs781012884 Chr.1: 167126532-167126532 on Build GRCh38
rs562826508 Chr.1: 167126511-167126511 on Build GRCh38
rs774975802 Chr.1: 167126446-167126446 on Build GRCh38
rs761615404 Chr.1: 167126141-167126141 on Build GRCh38
rs769769327 Chr.1: 167126381-167126383 on Build GRCh38
rs780524685 Chr.1: 167126277-167126277 on Build GRCh38
rs375239703 Chr.1: 167126527-167126527 on Build GRCh38
rs780676357 Chr.1: 167126489-167126489 on Build GRCh38
rs533498775 Chr.1: 167126518-167126518 on Build GRCh38
rs573171187 Chr.1: 167126444-167126444 on Build GRCh38
rs117105149 Chr.1: 167126231-167126231 on Build GRCh38
rs755364076 Chr.1: 167126400-167126400 on Build GRCh38
rs144860364 Chr.1: 167126540-167126540 on Build GRCh38
rs373710936 Chr.1: 167126166-167126166 on Build GRCh38
rs768888738 Chr.1: 167126209-167126209 on Build GRCh38
rs745699854 Chr.1: 167126424-167126424 on Build GRCh38
rs746111627 Chr.1: 167126095-167126095 on Build GRCh38
rs878895226 Chr.1: 167126429-167126429 on Build GRCh38
rs750817864 Chr.1: 167126195-167126195 on Build GRCh38
rs370233672 Chr.1: 167126497-167126497 on Build GRCh38
rs754989702 Chr.1: 167126314-167126343 on Build GRCh38
rs748218088 Chr.1: 167126305-167126305 on Build GRCh38
rs766979901 Chr.1: 167126397-167126397 on Build GRCh38
rs375985480 Chr.1: 167126224-167126224 on Build GRCh38
rs777563120 Chr.1: 167126531-167126531 on Build GRCh38
rs140173478 Chr.1: 167126454-167126454 on Build GRCh38
rs142610963 Chr.1: 167126458-167126458 on Build GRCh38
rs945247492 Chr.1: 167126354-167126354 on Build GRCh38
rs779434911 Chr.1: 167126144-167126149 on Build GRCh38
rs746634720 Chr.1: 167126426-167126426 on Build GRCh38
rs780416294 Chr.1: 167126103-167126103 on Build GRCh38
rs753039685 Chr.1: 167126320-167126320 on Build GRCh38
rs750366251 Chr.1: 167126520-167126520 on Build GRCh38

View Full Product Details