Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167992970-167993471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF6
Entrez Gene ID
55827
Gene Name
DDB1 and CUL4 associated factor 6
Gene Aliases
1200006M05Rik, ARCAP, IQWD1, MSTP055, NRIP, PC326
UniGene
Hs.435741
Gene Chromosome Location
Chr.1: 167936247-168075843 on Build GRCh38
Transcripts
NM_001017977.2 NM_001198956.1 NM_001198957.1 NM_018442.3 XM_005245331.4 XM_005245332.4 XM_005245333.4 XM_017001779.1 XM_017001780.1 XM_017001781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024842 c.763A>C p.R255R Chr.1: 167993300-167993300 on Build GRCh38
3984442 c.808G>A p.V270I Chr.1: 167993345-167993345 on Build GRCh38
7129670 c.700G>A p.G234S Chr.1: 167993237-167993237 on Build GRCh38
4398700 c.758C>T p.S253F Chr.1: 167993295-167993295 on Build GRCh38
3721414 c.726C>T p.A242A Chr.1: 167993263-167993263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778329582 Chr.1: 167993280-167993280 on Build GRCh38
rs970293387 Chr.1: 167993063-167993063 on Build GRCh38
rs752417831 Chr.1: 167993256-167993256 on Build GRCh38
rs930797531 Chr.1: 167993233-167993233 on Build GRCh38
rs762153897 Chr.1: 167993221-167993221 on Build GRCh38
rs745699952 Chr.1: 167993133-167993133 on Build GRCh38
rs747460982 Chr.1: 167993202-167993202 on Build GRCh38
rs763894032 Chr.1: 167993393-167993393 on Build GRCh38
rs373660631 Chr.1: 167993088-167993088 on Build GRCh38
rs973718958 Chr.1: 167993175-167993175 on Build GRCh38
rs1015360445 Chr.1: 167993310-167993310 on Build GRCh38
rs781658607 Chr.1: 167993314-167993314 on Build GRCh38
rs776857844 Chr.1: 167993218-167993218 on Build GRCh38
rs200843977 Chr.1: 167993264-167993264 on Build GRCh38
rs114673055 Chr.1: 167993140-167993140 on Build GRCh38
rs995929968 Chr.1: 167993066-167993066 on Build GRCh38
rs978653317 Chr.1: 167993073-167993073 on Build GRCh38
rs773326971 Chr.1: 167993358-167993358 on Build GRCh38
rs1028764176 Chr.1: 167993429-167993429 on Build GRCh38
rs746329187 Chr.1: 167993174-167993174 on Build GRCh38
rs368193640 Chr.1: 167993278-167993278 on Build GRCh38
rs372291616 Chr.1: 167993366-167993366 on Build GRCh38
rs1049258261 Chr.1: 167993241-167993241 on Build GRCh38
rs769286167 Chr.1: 167993207-167993209 on Build GRCh38
rs749913884 Chr.1: 167993416-167993416 on Build GRCh38
rs926019074 Chr.1: 167993080-167993080 on Build GRCh38
rs764787359 Chr.1: 167993414-167993414 on Build GRCh38
rs767409390 Chr.1: 167993249-167993249 on Build GRCh38
rs780564591 Chr.1: 167993188-167993188 on Build GRCh38
rs1013962018 Chr.1: 167993345-167993345 on Build GRCh38
rs968372390 Chr.1: 167993054-167993054 on Build GRCh38
rs748640034 Chr.1: 167993428-167993428 on Build GRCh38
rs769014287 Chr.1: 167993216-167993216 on Build GRCh38
rs767332265 Chr.1: 167993381-167993381 on Build GRCh38
rs142328126 Chr.1: 167993421-167993421 on Build GRCh38
rs938751673 Chr.1: 167993292-167993292 on Build GRCh38
rs769886782 Chr.1: 167993224-167993224 on Build GRCh38
rs148012547 Chr.1: 167993344-167993344 on Build GRCh38
rs574051701 Chr.1: 167993296-167993296 on Build GRCh38
rs773380695 Chr.1: 167993226-167993226 on Build GRCh38
rs143999777 Chr.1: 167993261-167993261 on Build GRCh38
rs1021325154 Chr.1: 167993007-167993007 on Build GRCh38
rs774565385 Chr.1: 167993369-167993369 on Build GRCh38
rs190266994 Chr.1: 167993145-167993145 on Build GRCh38
rs763156521 Chr.1: 167993228-167993228 on Build GRCh38
rs866592053 Chr.1: 167993105-167993105 on Build GRCh38
rs561877453 Chr.1: 167993382-167993382 on Build GRCh38
rs865966002 Chr.1: 167993178-167993178 on Build GRCh38
rs759601372 Chr.1: 167993377-167993377 on Build GRCh38
rs766516732 Chr.1: 167993234-167993234 on Build GRCh38
rs777627850 Chr.1: 167993218-167993218 on Build GRCh38
rs149756774 Chr.1: 167993359-167993359 on Build GRCh38
rs556649114 Chr.1: 167993112-167993112 on Build GRCh38
rs911977621 Chr.1: 167993443-167993443 on Build GRCh38
rs188682339 Chr.1: 167993412-167993412 on Build GRCh38
rs755464947 Chr.1: 167993282-167993282 on Build GRCh38
rs770392371 Chr.1: 167993186-167993186 on Build GRCh38
rs781580588 Chr.1: 167993422-167993422 on Build GRCh38
rs778495040 Chr.1: 167993011-167993011 on Build GRCh38
rs535545584 Chr.1: 167993087-167993087 on Build GRCh38
rs748428757 Chr.1: 167993323-167993323 on Build GRCh38
rs756447466 Chr.1: 167993432-167993432 on Build GRCh38
rs1010892221 Chr.1: 167992997-167992997 on Build GRCh38
rs529336881 Chr.1: 167993390-167993390 on Build GRCh38
rs1022884271 Chr.1: 167993037-167993037 on Build GRCh38
rs759477011 Chr.1: 167993235-167993235 on Build GRCh38
rs148884987 Chr.1: 167993383-167993383 on Build GRCh38
rs978985051 Chr.1: 167993368-167993368 on Build GRCh38
rs988876329 Chr.1: 167993123-167993123 on Build GRCh38
rs1044475466 Chr.1: 167993244-167993244 on Build GRCh38
rs753483406 Chr.1: 167993265-167993265 on Build GRCh38
rs753496627 Chr.1: 167993400-167993400 on Build GRCh38

View Full Product Details