Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169793550-169794086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL18
Entrez Gene ID
92342
Gene Name
methyltransferase like 18
Gene Aliases
AsTP2, C1orf156, HPM1
UniGene
Hs.33922
Gene Chromosome Location
Chr.1: 169792524-169794976 on Build GRCh38
Transcripts
NM_001320199.1 NM_001320201.1 NM_033418.3 XM_006711627.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6711644 c.127G>C p.E43Q Chr.1: 169793569-169793569 on Build GRCh38
5047413 c.124G>A p.E42K Chr.1: 169793572-169793572 on Build GRCh38
5978153 c.113G>A p.S38N Chr.1: 169793583-169793583 on Build GRCh38
5844459 c.121G>A p.G41R Chr.1: 169793575-169793575 on Build GRCh38
4975608 c.17A>T p.N6I Chr.1: 169793679-169793679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773461599 Chr.1: 169793676-169793676 on Build GRCh38
rs12059182 Chr.1: 169793985-169793985 on Build GRCh38
rs535664569 Chr.1: 169793654-169793654 on Build GRCh38
rs769948548 Chr.1: 169793616-169793616 on Build GRCh38
rs551779239 Chr.1: 169794022-169794022 on Build GRCh38
rs765912527 Chr.1: 169793662-169793662 on Build GRCh38
rs539887999 Chr.1: 169793739-169793739 on Build GRCh38
rs71713237 Chr.1: 169793906-169793906 on Build GRCh38
rs544006707 Chr.1: 169793606-169793606 on Build GRCh38
rs755909503 Chr.1: 169793687-169793687 on Build GRCh38
rs1048161340 Chr.1: 169793764-169793764 on Build GRCh38
rs111419588 Chr.1: 169793907-169793907 on Build GRCh38
rs574744291 Chr.1: 169793948-169793948 on Build GRCh38
rs756255684 Chr.1: 169793905-169793905 on Build GRCh38
rs781299878 Chr.1: 169793701-169793701 on Build GRCh38
rs916321614 Chr.1: 169794015-169794015 on Build GRCh38
rs749673109 Chr.1: 169793638-169793638 on Build GRCh38
rs769018139 Chr.1: 169793719-169793719 on Build GRCh38
rs181253765 Chr.1: 169794037-169794037 on Build GRCh38
rs866429994 Chr.1: 169793842-169793842 on Build GRCh38
rs763099452 Chr.1: 169793743-169793743 on Build GRCh38
rs141615875 Chr.1: 169793849-169793849 on Build GRCh38
rs970270184 Chr.1: 169793972-169793972 on Build GRCh38
rs749585335 Chr.1: 169793718-169793718 on Build GRCh38
rs1018758162 Chr.1: 169793821-169793821 on Build GRCh38
rs866678613 Chr.1: 169793905-169793905 on Build GRCh38
rs751530290 Chr.1: 169793698-169793698 on Build GRCh38
rs61807814 Chr.1: 169793984-169793984 on Build GRCh38
rs944252600 Chr.1: 169793671-169793671 on Build GRCh38
rs199794642 Chr.1: 169793921-169793922 on Build GRCh38
rs1041219126 Chr.1: 169793674-169793674 on Build GRCh38
rs775899587 Chr.1: 169793628-169793628 on Build GRCh38
rs757207495 Chr.1: 169793700-169793700 on Build GRCh38
rs928794576 Chr.1: 169793964-169793964 on Build GRCh38
rs898606688 Chr.1: 169793926-169793926 on Build GRCh38
rs756309982 Chr.1: 169793714-169793714 on Build GRCh38
rs149017105 Chr.1: 169793604-169793604 on Build GRCh38
rs750516308 Chr.1: 169793710-169793710 on Build GRCh38
rs978088976 Chr.1: 169793932-169793932 on Build GRCh38
rs529365670 Chr.1: 169793854-169793854 on Build GRCh38
rs147719562 Chr.1: 169793651-169793651 on Build GRCh38
rs557955943 Chr.1: 169793771-169793771 on Build GRCh38
rs1014550629 Chr.1: 169793923-169793923 on Build GRCh38
rs371427681 Chr.1: 169793665-169793665 on Build GRCh38
rs1052431139 Chr.1: 169793632-169793632 on Build GRCh38
rs1038032006 Chr.1: 169793653-169793653 on Build GRCh38
rs752501395 Chr.1: 169793677-169793677 on Build GRCh38
rs898177103 Chr.1: 169794059-169794059 on Build GRCh38
rs1003874865 Chr.1: 169793846-169793846 on Build GRCh38
rs528132166 Chr.1: 169794028-169794028 on Build GRCh38
rs964154093 Chr.1: 169793847-169793847 on Build GRCh38
rs967588245 Chr.1: 169793918-169793918 on Build GRCh38
rs553272071 Chr.1: 169793901-169793901 on Build GRCh38
rs1021674382 Chr.1: 169793885-169793885 on Build GRCh38
rs770020853 Chr.1: 169793653-169793665 on Build GRCh38
rs562049305 Chr.1: 169794043-169794043 on Build GRCh38
rs1008289849 Chr.1: 169793806-169793806 on Build GRCh38
rs10489177 Chr.1: 169793666-169793666 on Build GRCh38
rs764822937 Chr.1: 169793669-169793669 on Build GRCh38
rs936678146 Chr.1: 169793591-169793591 on Build GRCh38
rs955399802 Chr.1: 169793762-169793762 on Build GRCh38
rs748269153 Chr.1: 169793617-169793617 on Build GRCh38
rs781185874 Chr.1: 169793598-169793598 on Build GRCh38
rs1038375215 Chr.1: 169793922-169793922 on Build GRCh38
rs143505821 Chr.1: 169793685-169793685 on Build GRCh38
rs774839631 Chr.1: 169793652-169793652 on Build GRCh38
rs529517771 Chr.1: 169794051-169794051 on Build GRCh38
rs994051036 Chr.1: 169793703-169793703 on Build GRCh38
rs564001385 Chr.1: 169794000-169794000 on Build GRCh38
rs368584829 Chr.1: 169793587-169793587 on Build GRCh38
rs542048210 Chr.1: 169793954-169793954 on Build GRCh38
rs10919245 Chr.1: 169793998-169793998 on Build GRCh38
rs573217771 Chr.1: 169793811-169793811 on Build GRCh38
rs764066076 Chr.1: 169793692-169793692 on Build GRCh38
rs375981531 Chr.1: 169793716-169793716 on Build GRCh38

View Full Product Details