Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96145272-96145768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF6
Entrez Gene ID
392255
Gene Name
growth differentiation factor 6
Gene Aliases
BMP-13, BMP13, CDMP2, KFM, KFS, KFS1, KFSL, LCA17, MCOP4, MCOPCB6, SCDO4, SGM1
UniGene
Hs.492277
Gene Chromosome Location
Chr.8: 96142330-96160792 on Build GRCh38
Transcripts
NM_001001557.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5723944 c.616G>A p.G206R Chr.8: 96145315-96145315 on Build GRCh38
3902269 c.525G>A p.A175A Chr.8: 96145406-96145406 on Build GRCh38
6670277 c.469G>T p.D157Y Chr.8: 96145462-96145462 on Build GRCh38
297994 c.489C>T p.G163G Chr.8: 96145442-96145442 on Build GRCh38
4997723 c.625C>A p.P209T Chr.8: 96145306-96145306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901058669 Chr.8: 96145315-96145315 on Build GRCh38
rs369127445 Chr.8: 96145496-96145496 on Build GRCh38
rs958381615 Chr.8: 96145450-96145450 on Build GRCh38
rs1005295163 Chr.8: 96145433-96145433 on Build GRCh38
rs758826687 Chr.8: 96145683-96145683 on Build GRCh38
rs993069013 Chr.8: 96145462-96145462 on Build GRCh38
rs936122584 Chr.8: 96145560-96145560 on Build GRCh38
rs866756897 Chr.8: 96145353-96145353 on Build GRCh38
rs528471783 Chr.8: 96145385-96145385 on Build GRCh38
rs770401948 Chr.8: 96145464-96145464 on Build GRCh38
rs745516566 Chr.8: 96145447-96145447 on Build GRCh38
rs374740957 Chr.8: 96145527-96145527 on Build GRCh38
rs759041718 Chr.8: 96145342-96145342 on Build GRCh38
rs1016475475 Chr.8: 96145667-96145667 on Build GRCh38
rs552234954 Chr.8: 96145300-96145300 on Build GRCh38
rs759752149 Chr.8: 96145494-96145494 on Build GRCh38
rs752118839 Chr.8: 96145704-96145704 on Build GRCh38
rs1006400954 Chr.8: 96145649-96145649 on Build GRCh38
rs977566933 Chr.8: 96145533-96145533 on Build GRCh38
rs777218109 Chr.8: 96145348-96145348 on Build GRCh38
rs1040377646 Chr.8: 96145583-96145583 on Build GRCh38
rs374477999 Chr.8: 96145610-96145610 on Build GRCh38
rs370351474 Chr.8: 96145513-96145513 on Build GRCh38
rs773901485 Chr.8: 96145338-96145338 on Build GRCh38
rs773952649 Chr.8: 96145562-96145563 on Build GRCh38
rs577607274 Chr.8: 96145394-96145394 on Build GRCh38
rs387906794 Chr.8: 96145336-96145336 on Build GRCh38
rs200585226 Chr.8: 96145448-96145448 on Build GRCh38
rs958419622 Chr.8: 96145738-96145738 on Build GRCh38
rs766573793 Chr.8: 96145380-96145380 on Build GRCh38
rs888284937 Chr.8: 96145699-96145699 on Build GRCh38
rs564008975 Chr.8: 96145310-96145310 on Build GRCh38
rs751734883 Chr.8: 96145382-96145382 on Build GRCh38
rs777541984 Chr.8: 96145319-96145319 on Build GRCh38
rs749067069 Chr.8: 96145415-96145415 on Build GRCh38
rs1034090525 Chr.8: 96145731-96145731 on Build GRCh38
rs572155748 Chr.8: 96145674-96145674 on Build GRCh38
rs763310577 Chr.8: 96145470-96145470 on Build GRCh38
rs757029731 Chr.8: 96145439-96145439 on Build GRCh38
rs886063209 Chr.8: 96145432-96145432 on Build GRCh38
rs961245472 Chr.8: 96145330-96145330 on Build GRCh38
rs765908995 Chr.8: 96145375-96145375 on Build GRCh38
rs773615024 Chr.8: 96145468-96145468 on Build GRCh38
rs778557602 Chr.8: 96145440-96145440 on Build GRCh38
rs372590219 Chr.8: 96145498-96145498 on Build GRCh38
rs901094674 Chr.8: 96145606-96145606 on Build GRCh38
rs1047374265 Chr.8: 96145671-96145671 on Build GRCh38
rs999492360 Chr.8: 96145320-96145320 on Build GRCh38
rs375992269 Chr.8: 96145502-96145502 on Build GRCh38
rs903826881 Chr.8: 96145308-96145308 on Build GRCh38
rs775133514 Chr.8: 96145460-96145460 on Build GRCh38
rs994022206 Chr.8: 96145337-96145337 on Build GRCh38
rs972498586 Chr.8: 96145678-96145678 on Build GRCh38
rs746556723 Chr.8: 96145463-96145463 on Build GRCh38
rs756016892 Chr.8: 96145397-96145397 on Build GRCh38
rs763275206 Chr.8: 96145377-96145377 on Build GRCh38
rs375769410 Chr.8: 96145511-96145511 on Build GRCh38
rs779900532 Chr.8: 96145530-96145530 on Build GRCh38
rs11783820 Chr.8: 96145564-96145564 on Build GRCh38
rs1030997543 Chr.8: 96145329-96145329 on Build GRCh38
rs760676378 Chr.8: 96145576-96145576 on Build GRCh38
rs766667262 Chr.8: 96145471-96145471 on Build GRCh38
rs768313069 Chr.8: 96145559-96145559 on Build GRCh38
rs200937042 Chr.8: 96145574-96145574 on Build GRCh38
rs773711684 Chr.8: 96145561-96145561 on Build GRCh38
rs767572263 Chr.8: 96145387-96145387 on Build GRCh38
rs77859767 Chr.8: 96145563-96145563 on Build GRCh38
rs771710004 Chr.8: 96145344-96145344 on Build GRCh38
rs536565151 Chr.8: 96145544-96145544 on Build GRCh38
rs758091453 Chr.8: 96145529-96145529 on Build GRCh38
rs896514917 Chr.8: 96145566-96145566 on Build GRCh38
rs764057334 Chr.8: 96145506-96145506 on Build GRCh38
rs745721870 Chr.8: 96145528-96145528 on Build GRCh38
rs548565578 Chr.8: 96145403-96145403 on Build GRCh38
rs565877093 Chr.8: 96145357-96145357 on Build GRCh38
rs749079662 Chr.8: 96145325-96145325 on Build GRCh38

View Full Product Details