Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100573800-100574311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX31
Entrez Gene ID
169166
Gene Name
sorting nexin 31
Gene Aliases
-
UniGene
Hs.567758
Gene Chromosome Location
Chr.8: 100572882-100663410 on Build GRCh38
Transcripts
NM_152628.3 XM_005250815.3 XM_011516899.1 XM_017013153.1 XM_017013154.1 XM_017013155.1 XM_017013156.1 XM_017013157.1 XM_017013158.1 XM_017013159.1 XM_017013160.1 XM_017013161.1 XM_017013162.1 XM_017013163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3778784 c.1312G>A p.E438K Chr.8: 100573876-100573876 on Build GRCh38
7091633 c.1234G>A p.D412N Chr.8: 100573954-100573954 on Build GRCh38
4406038 c.1257A>G p.R419R Chr.8: 100573931-100573931 on Build GRCh38
6179258 c.1253C>A p.S418* Chr.8: 100573935-100573935 on Build GRCh38
1453645 c.1296delT p.F432fs*4 Chr.8: 100573892-100573892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761306337 Chr.8: 100573892-100573892 on Build GRCh38
rs1043407007 Chr.8: 100574044-100574044 on Build GRCh38
rs935856851 Chr.8: 100573940-100573940 on Build GRCh38
rs779507196 Chr.8: 100573865-100573865 on Build GRCh38
rs150625089 Chr.8: 100573936-100573936 on Build GRCh38
rs765075217 Chr.8: 100573925-100573925 on Build GRCh38
rs761459950 Chr.8: 100573922-100573922 on Build GRCh38
rs758023579 Chr.8: 100573932-100573932 on Build GRCh38
rs897353320 Chr.8: 100574249-100574249 on Build GRCh38
rs774497341 Chr.8: 100573903-100573903 on Build GRCh38
rs753424240 Chr.8: 100573919-100573920 on Build GRCh38
rs1019753213 Chr.8: 100574223-100574223 on Build GRCh38
rs368048656 Chr.8: 100574255-100574256 on Build GRCh38
rs1008426274 Chr.8: 100573889-100573889 on Build GRCh38
rs372142006 Chr.8: 100573842-100573842 on Build GRCh38
rs764029476 Chr.8: 100573826-100573826 on Build GRCh38
rs757968591 Chr.8: 100573857-100573857 on Build GRCh38
rs994420997 Chr.8: 100574264-100574264 on Build GRCh38
rs147125270 Chr.8: 100574201-100574201 on Build GRCh38
rs751528411 Chr.8: 100573974-100573975 on Build GRCh38
rs746145487 Chr.8: 100573973-100573973 on Build GRCh38
rs980142442 Chr.8: 100573937-100573937 on Build GRCh38
rs752425672 Chr.8: 100573926-100573927 on Build GRCh38
rs549322527 Chr.8: 100574099-100574099 on Build GRCh38
rs771196432 Chr.8: 100573900-100573900 on Build GRCh38
rs565697594 Chr.8: 100574270-100574270 on Build GRCh38
rs138539577 Chr.8: 100574274-100574274 on Build GRCh38
rs764892779 Chr.8: 100573925-100573926 on Build GRCh38
rs765806124 Chr.8: 100573919-100573926 on Build GRCh38
rs749545695 Chr.8: 100573898-100573898 on Build GRCh38
rs913065836 Chr.8: 100574032-100574032 on Build GRCh38
rs767107523 Chr.8: 100573926-100573926 on Build GRCh38
rs547061899 Chr.8: 100574222-100574222 on Build GRCh38
rs2022923 Chr.8: 100573905-100573905 on Build GRCh38
rs563536116 Chr.8: 100574108-100574108 on Build GRCh38
rs529938376 Chr.8: 100574035-100574035 on Build GRCh38
rs753673409 Chr.8: 100573920-100573920 on Build GRCh38
rs141889730 Chr.8: 100573919-100573919 on Build GRCh38
rs772321553 Chr.8: 100573908-100573908 on Build GRCh38
rs199975268 Chr.8: 100573897-100573897 on Build GRCh38
rs569562679 Chr.8: 100574168-100574168 on Build GRCh38
rs749791336 Chr.8: 100573953-100573953 on Build GRCh38
rs370727447 Chr.8: 100574005-100574005 on Build GRCh38
rs767118643 Chr.8: 100573978-100573978 on Build GRCh38
rs759084746 Chr.8: 100573922-100573926 on Build GRCh38
rs773805331 Chr.8: 100573880-100573883 on Build GRCh38
rs141504180 Chr.8: 100574172-100574172 on Build GRCh38
rs772372255 Chr.8: 100573974-100573974 on Build GRCh38
rs368683915 Chr.8: 100573835-100573835 on Build GRCh38
rs775184373 Chr.8: 100573979-100573979 on Build GRCh38
rs748703713 Chr.8: 100573866-100573866 on Build GRCh38
rs768653129 Chr.8: 100573988-100573988 on Build GRCh38
rs1001180752 Chr.8: 100574066-100574066 on Build GRCh38
rs753527944 Chr.8: 100573828-100573828 on Build GRCh38
rs779558029 Chr.8: 100573935-100573935 on Build GRCh38
rs757284770 Chr.8: 100574009-100574010 on Build GRCh38
rs112486101 Chr.8: 100573993-100573993 on Build GRCh38
rs775693920 Chr.8: 100573909-100573909 on Build GRCh38
rs533250426 Chr.8: 100574014-100574014 on Build GRCh38
rs201004595 Chr.8: 100573959-100573959 on Build GRCh38
rs777722888 Chr.8: 100573970-100573973 on Build GRCh38
rs745518848 Chr.8: 100573850-100573850 on Build GRCh38
rs528825723 Chr.8: 100574215-100574215 on Build GRCh38
rs201163996 Chr.8: 100573892-100573892 on Build GRCh38
rs767875315 Chr.8: 100573872-100573874 on Build GRCh38

View Full Product Details