Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109575242-109575745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYBU
Entrez Gene ID
55638
Gene Name
syntabulin
Gene Aliases
GOLSYN, OCSYN, SNPHL
UniGene
Hs.390738
Gene Chromosome Location
Chr.8: 109574176-109691791 on Build GRCh38
Transcripts
NM_001099743.1 NM_001099744.1 NM_001099745.1 NM_001099746.1 NM_001099747.1 NM_001099748.1 NM_001099749.1 NM_001099750.1 NM_001099751.1 NM_001099752.1 NM_001099753.1 NM_001099754.1 NM_001099755.1 NM_001099756.1 NM_017786.5 XM_005250999.1 XM_005251000.1 XM_011517154.2 XM_017013614.1 XM_017013615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7265762 c.1390G>A p.V464I Chr.8: 109575499-109575499 on Build GRCh38
6530478 c.1534C>G p.P512A Chr.8: 109575355-109575355 on Build GRCh38
5014814 c.1567C>T p.P523S Chr.8: 109575322-109575322 on Build GRCh38
1094988 c.1628G>T p.R543I Chr.8: 109575261-109575261 on Build GRCh38
4731083 c.1497C>T p.L499L Chr.8: 109575392-109575392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779784998 Chr.8: 109575442-109575442 on Build GRCh38
rs750027113 Chr.8: 109575635-109575635 on Build GRCh38
rs772224988 Chr.8: 109575571-109575571 on Build GRCh38
rs926635461 Chr.8: 109575446-109575446 on Build GRCh38
rs775913322 Chr.8: 109575695-109575695 on Build GRCh38
rs769878772 Chr.8: 109575359-109575359 on Build GRCh38
rs770148862 Chr.8: 109575693-109575693 on Build GRCh38
rs776007436 Chr.8: 109575617-109575617 on Build GRCh38
rs773240808 Chr.8: 109575572-109575572 on Build GRCh38
rs780668031 Chr.8: 109575515-109575515 on Build GRCh38
rs751857357 Chr.8: 109575303-109575303 on Build GRCh38
rs377270994 Chr.8: 109575433-109575433 on Build GRCh38
rs777018998 Chr.8: 109575600-109575600 on Build GRCh38
rs776859500 Chr.8: 109575684-109575684 on Build GRCh38
rs373716676 Chr.8: 109575461-109575461 on Build GRCh38
rs901793182 Chr.8: 109575330-109575330 on Build GRCh38
rs746189922 Chr.8: 109575685-109575685 on Build GRCh38
rs185384946 Chr.8: 109575414-109575414 on Build GRCh38
rs941668181 Chr.8: 109575644-109575645 on Build GRCh38
rs760095575 Chr.8: 109575502-109575502 on Build GRCh38
rs376741303 Chr.8: 109575547-109575547 on Build GRCh38
rs762186378 Chr.8: 109575394-109575394 on Build GRCh38
rs749236773 Chr.8: 109575549-109575549 on Build GRCh38
rs773509419 Chr.8: 109575467-109575467 on Build GRCh38
rs754244017 Chr.8: 109575499-109575499 on Build GRCh38
rs369787649 Chr.8: 109575460-109575460 on Build GRCh38
rs746670378 Chr.8: 109575348-109575348 on Build GRCh38
rs775472677 Chr.8: 109575284-109575284 on Build GRCh38
rs769667914 Chr.8: 109575383-109575383 on Build GRCh38
rs754845803 Chr.8: 109575667-109575667 on Build GRCh38
rs772821201 Chr.8: 109575709-109575709 on Build GRCh38
rs761157517 Chr.8: 109575622-109575622 on Build GRCh38
rs749606891 Chr.8: 109575407-109575412 on Build GRCh38
rs779118117 Chr.8: 109575558-109575558 on Build GRCh38
rs760909578 Chr.8: 109575474-109575474 on Build GRCh38
rs368819508 Chr.8: 109575385-109575385 on Build GRCh38
rs756649298 Chr.8: 109575327-109575327 on Build GRCh38
rs766083751 Chr.8: 109575714-109575714 on Build GRCh38
rs35097569 Chr.8: 109575665-109575665 on Build GRCh38
rs937421054 Chr.8: 109575599-109575599 on Build GRCh38
rs778958654 Chr.8: 109575670-109575670 on Build GRCh38
rs995392568 Chr.8: 109575718-109575718 on Build GRCh38
rs746012867 Chr.8: 109575307-109575307 on Build GRCh38
rs201133211 Chr.8: 109575341-109575341 on Build GRCh38
rs771187944 Chr.8: 109575598-109575598 on Build GRCh38
rs563748833 Chr.8: 109575411-109575411 on Build GRCh38
rs370023559 Chr.8: 109575268-109575268 on Build GRCh38
rs774857289 Chr.8: 109575563-109575564 on Build GRCh38
rs769081037 Chr.8: 109575435-109575436 on Build GRCh38
rs766771826 Chr.8: 109575495-109575495 on Build GRCh38
rs748101787 Chr.8: 109575672-109575672 on Build GRCh38
rs759449078 Chr.8: 109575274-109575274 on Build GRCh38
rs765683813 Chr.8: 109575616-109575616 on Build GRCh38
rs760225917 Chr.8: 109575713-109575713 on Build GRCh38
rs865880844 Chr.8: 109575631-109575631 on Build GRCh38
rs756942073 Chr.8: 109575436-109575436 on Build GRCh38
rs372495769 Chr.8: 109575342-109575342 on Build GRCh38
rs1006398580 Chr.8: 109575358-109575359 on Build GRCh38
rs750931844 Chr.8: 109575314-109575314 on Build GRCh38
rs755016337 Chr.8: 109575646-109575646 on Build GRCh38
rs766059531 Chr.8: 109575643-109575643 on Build GRCh38
rs193166099 Chr.8: 109575298-109575298 on Build GRCh38
rs767992136 Chr.8: 109575404-109575404 on Build GRCh38
rs905942879 Chr.8: 109575272-109575272 on Build GRCh38
rs768756257 Chr.8: 109575551-109575551 on Build GRCh38
rs765353346 Chr.8: 109575281-109575281 on Build GRCh38
rs771231470 Chr.8: 109575683-109575683 on Build GRCh38
rs147762841 Chr.8: 109575386-109575386 on Build GRCh38
rs533087434 Chr.8: 109575702-109575702 on Build GRCh38
rs374617962 Chr.8: 109575395-109575395 on Build GRCh38
rs768840076 Chr.8: 109575455-109575455 on Build GRCh38
rs1050332305 Chr.8: 109575587-109575587 on Build GRCh38
rs1048020411 Chr.8: 109575722-109575722 on Build GRCh38
rs747193882 Chr.8: 109575682-109575682 on Build GRCh38
rs1054929630 Chr.8: 109575321-109575321 on Build GRCh38
rs559538024 Chr.8: 109575486-109575486 on Build GRCh38
rs779954079 Chr.8: 109575536-109575536 on Build GRCh38
rs369592030 Chr.8: 109575583-109575583 on Build GRCh38
rs202051352 Chr.8: 109575555-109575555 on Build GRCh38
rs945434764 Chr.8: 109575566-109575566 on Build GRCh38
rs374542726 Chr.8: 109575304-109575304 on Build GRCh38
rs764272355 Chr.8: 109575393-109575393 on Build GRCh38
rs543114252 Chr.8: 109575425-109575425 on Build GRCh38
rs762367987 Chr.8: 109575630-109575631 on Build GRCh38
rs778459471 Chr.8: 109575331-109575331 on Build GRCh38
rs141149968 Chr.8: 109575681-109575681 on Build GRCh38
rs760013683 Chr.8: 109575613-109575613 on Build GRCh38
rs763562789 Chr.8: 109575505-109575505 on Build GRCh38
rs766748650 Chr.8: 109575416-109575416 on Build GRCh38
rs745600122 Chr.8: 109575439-109575439 on Build GRCh38
rs772726408 Chr.8: 109575692-109575693 on Build GRCh38
rs751117748 Chr.8: 109575507-109575507 on Build GRCh38
rs748193812 Chr.8: 109575560-109575560 on Build GRCh38
rs755804798 Chr.8: 109575523-109575523 on Build GRCh38
rs774642862 Chr.8: 109575456-109575456 on Build GRCh38
rs750091115 Chr.8: 109575520-109575520 on Build GRCh38
rs754753690 Chr.8: 109575724-109575724 on Build GRCh38
rs769658598 Chr.8: 109575441-109575441 on Build GRCh38
rs751824083 Chr.8: 109575290-109575290 on Build GRCh38
rs767064145 Chr.8: 109575630-109575630 on Build GRCh38
rs913467436 Chr.8: 109575484-109575484 on Build GRCh38
rs777278797 Chr.8: 109575428-109575428 on Build GRCh38
rs754400906 Chr.8: 109575421-109575421 on Build GRCh38
rs781489210 Chr.8: 109575437-109575437 on Build GRCh38
rs753580075 Chr.8: 109575721-109575721 on Build GRCh38
rs780394698 Chr.8: 109575384-109575385 on Build GRCh38
rs374229180 Chr.8: 109575344-109575344 on Build GRCh38
rs376682836 Chr.8: 109575365-109575365 on Build GRCh38
rs140751107 Chr.8: 109575350-109575350 on Build GRCh38
rs768004422 Chr.8: 109575311-109575311 on Build GRCh38
rs201853391 Chr.8: 109575351-109575351 on Build GRCh38
rs763570052 Chr.8: 109575700-109575700 on Build GRCh38
rs766929802 Chr.8: 109575701-109575701 on Build GRCh38
rs760967757 Chr.8: 109575580-109575580 on Build GRCh38
rs199889456 Chr.8: 109575636-109575636 on Build GRCh38
rs756782697 Chr.8: 109575508-109575508 on Build GRCh38

View Full Product Details