Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123681098-123681585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA13
Entrez Gene ID
312
Gene Name
annexin A13
Gene Aliases
ANX13, ISA
UniGene
Hs.181107
Gene Chromosome Location
Chr.8: 123680794-123737426 on Build GRCh38
Transcripts
NM_001003954.2 NM_004306.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
453970 c.964C>T p.Q322* Chr.8: 123681350-123681350 on Build GRCh38
3315967 c.1014C>T p.D338D Chr.8: 123681300-123681300 on Build GRCh38
5963123 c.1021C>T p.R341C Chr.8: 123681293-123681293 on Build GRCh38
3315964 c.1035C>T p.S345S Chr.8: 123681279-123681279 on Build GRCh38
421749 c.969G>A p.G323G Chr.8: 123681345-123681345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3739283 Chr.8: 123681204-123681204 on Build GRCh38
rs745692488 Chr.8: 123681315-123681315 on Build GRCh38
rs771029692 Chr.8: 123681301-123681301 on Build GRCh38
rs747750676 Chr.8: 123681215-123681215 on Build GRCh38
rs975035129 Chr.8: 123681516-123681516 on Build GRCh38
rs371550350 Chr.8: 123681398-123681398 on Build GRCh38
rs777800831 Chr.8: 123681297-123681297 on Build GRCh38
rs908465474 Chr.8: 123681551-123681551 on Build GRCh38
rs143112749 Chr.8: 123681269-123681269 on Build GRCh38
rs999463238 Chr.8: 123681431-123681431 on Build GRCh38
rs558469036 Chr.8: 123681225-123681225 on Build GRCh38
rs199504938 Chr.8: 123681153-123681153 on Build GRCh38
rs775817458 Chr.8: 123681342-123681342 on Build GRCh38
rs767838530 Chr.8: 123681278-123681278 on Build GRCh38
rs149087670 Chr.8: 123681268-123681268 on Build GRCh38
rs201212750 Chr.8: 123681293-123681293 on Build GRCh38
rs773390233 Chr.8: 123681378-123681378 on Build GRCh38
rs201780457 Chr.8: 123681203-123681203 on Build GRCh38
rs771846238 Chr.8: 123681218-123681218 on Build GRCh38
rs562845658 Chr.8: 123681319-123681319 on Build GRCh38
rs955234967 Chr.8: 123681537-123681537 on Build GRCh38
rs560541609 Chr.8: 123681276-123681276 on Build GRCh38
rs535157130 Chr.8: 123681515-123681515 on Build GRCh38
rs773103495 Chr.8: 123681559-123681559 on Build GRCh38
rs774819153 Chr.8: 123681303-123681304 on Build GRCh38
rs752416217 Chr.8: 123681393-123681393 on Build GRCh38
rs746759219 Chr.8: 123681223-123681223 on Build GRCh38
rs1036416748 Chr.8: 123681295-123681295 on Build GRCh38
rs765388309 Chr.8: 123681503-123681503 on Build GRCh38
rs754700849 Chr.8: 123681205-123681205 on Build GRCh38
rs761103804 Chr.8: 123681383-123681383 on Build GRCh38
rs4471017 Chr.8: 123681170-123681170 on Build GRCh38
rs780092024 Chr.8: 123681309-123681309 on Build GRCh38
rs746916602 Chr.8: 123681299-123681299 on Build GRCh38
rs531775603 Chr.8: 123681353-123681353 on Build GRCh38
rs770115414 Chr.8: 123681246-123681246 on Build GRCh38
rs543175718 Chr.8: 123681302-123681302 on Build GRCh38
rs979237950 Chr.8: 123681160-123681160 on Build GRCh38
rs368842790 Chr.8: 123681292-123681292 on Build GRCh38
rs1027917811 Chr.8: 123681518-123681518 on Build GRCh38
rs759573811 Chr.8: 123681429-123681429 on Build GRCh38
rs1029014295 Chr.8: 123681434-123681435 on Build GRCh38
rs375500669 Chr.8: 123681228-123681228 on Build GRCh38
rs185422886 Chr.8: 123681394-123681394 on Build GRCh38
rs756912797 Chr.8: 123681133-123681133 on Build GRCh38
rs762896828 Chr.8: 123681345-123681345 on Build GRCh38
rs945326287 Chr.8: 123681524-123681524 on Build GRCh38
rs554928218 Chr.8: 123681157-123681157 on Build GRCh38
rs566365700 Chr.8: 123681450-123681450 on Build GRCh38
rs566253977 Chr.8: 123681130-123681130 on Build GRCh38
rs1014823367 Chr.8: 123681132-123681132 on Build GRCh38
rs141190521 Chr.8: 123681291-123681291 on Build GRCh38
rs777855966 Chr.8: 123681221-123681221 on Build GRCh38
rs578080565 Chr.8: 123681253-123681253 on Build GRCh38
rs956557585 Chr.8: 123681174-123681174 on Build GRCh38
rs1000940924 Chr.8: 123681492-123681492 on Build GRCh38
rs973942907 Chr.8: 123681554-123681554 on Build GRCh38
rs1055905792 Chr.8: 123681564-123681564 on Build GRCh38
rs374770249 Chr.8: 123681375-123681375 on Build GRCh38
rs930683820 Chr.8: 123681561-123681561 on Build GRCh38
rs752083851 Chr.8: 123681270-123681270 on Build GRCh38
rs568504159 Chr.8: 123681550-123681550 on Build GRCh38
rs369165325 Chr.8: 123681384-123681384 on Build GRCh38
rs778283233 Chr.8: 123681213-123681213 on Build GRCh38
rs781283030 Chr.8: 123681409-123681409 on Build GRCh38
rs961230642 Chr.8: 123681553-123681553 on Build GRCh38
rs768701348 Chr.8: 123681348-123681348 on Build GRCh38
rs757206497 Chr.8: 123681406-123681406 on Build GRCh38
rs368349500 Chr.8: 123681195-123681195 on Build GRCh38
rs781127320 Chr.8: 123681306-123681306 on Build GRCh38
rs930629781 Chr.8: 123681329-123681329 on Build GRCh38
rs750855704 Chr.8: 123681279-123681279 on Build GRCh38
rs770869915 Chr.8: 123681224-123681224 on Build GRCh38
rs550563682 Chr.8: 123681373-123681373 on Build GRCh38
rs535049874 Chr.8: 123681134-123681134 on Build GRCh38
rs148683632 Chr.8: 123681374-123681374 on Build GRCh38
rs4314634 Chr.8: 123681401-123681401 on Build GRCh38
rs548716263 Chr.8: 123681532-123681532 on Build GRCh38
rs867128998 Chr.8: 123681453-123681453 on Build GRCh38
rs933954044 Chr.8: 123681261-123681261 on Build GRCh38

View Full Product Details