Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123980590-123981086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L6
Entrez Gene ID
654463
Gene Name
fer-1 like family member 6
Gene Aliases
C8ORFK23
UniGene
Hs.632058
Gene Chromosome Location
Chr.8: 123851987-124120061 on Build GRCh38
Transcripts
NM_001039112.2 XM_006716618.3 XM_011517231.2 XM_011517232.2 XM_011517233.2 XM_011517235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3644878 c.1277C>T p.S426F Chr.8: 123980678-123980678 on Build GRCh38
3644879 c.1282G>A p.D428N Chr.8: 123980683-123980683 on Build GRCh38
317360 c.1368C>A p.N456K Chr.8: 123980769-123980769 on Build GRCh38
4769793 c.1226G>A p.R409Q Chr.8: 123980627-123980627 on Build GRCh38
3644880 c.1337G>A p.G446E Chr.8: 123980738-123980738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763839227 Chr.8: 123980921-123980921 on Build GRCh38
rs529243460 Chr.8: 123980866-123980866 on Build GRCh38
rs895725597 Chr.8: 123981022-123981022 on Build GRCh38
rs1032492093 Chr.8: 123980918-123980918 on Build GRCh38
rs372652726 Chr.8: 123980807-123980807 on Build GRCh38
rs34872463 Chr.8: 123981019-123981020 on Build GRCh38
rs375477178 Chr.8: 123981012-123981012 on Build GRCh38
rs752782719 Chr.8: 123981018-123981018 on Build GRCh38
rs78034602 Chr.8: 123980791-123980791 on Build GRCh38
rs760684546 Chr.8: 123980655-123980655 on Build GRCh38
rs1044587542 Chr.8: 123980957-123980957 on Build GRCh38
rs765650846 Chr.8: 123980805-123980805 on Build GRCh38
rs758857875 Chr.8: 123980686-123980686 on Build GRCh38
rs968447583 Chr.8: 123980841-123980841 on Build GRCh38
rs766165566 Chr.8: 123980835-123980835 on Build GRCh38
rs917311678 Chr.8: 123980934-123980934 on Build GRCh38
rs533520976 Chr.8: 123981060-123981060 on Build GRCh38
rs200225461 Chr.8: 123980709-123980709 on Build GRCh38
rs186787361 Chr.8: 123980832-123980832 on Build GRCh38
rs568910344 Chr.8: 123980906-123980906 on Build GRCh38
rs776542917 Chr.8: 123980798-123980798 on Build GRCh38
rs538054635 Chr.8: 123980972-123980972 on Build GRCh38
rs777414002 Chr.8: 123980839-123980839 on Build GRCh38
rs753260281 Chr.8: 123980843-123980843 on Build GRCh38
rs750103113 Chr.8: 123980699-123980699 on Build GRCh38
rs527654182 Chr.8: 123980775-123980775 on Build GRCh38
rs773368685 Chr.8: 123980792-123980792 on Build GRCh38
rs1010335853 Chr.8: 123980810-123980810 on Build GRCh38
rs549155736 Chr.8: 123980875-123980875 on Build GRCh38
rs754296474 Chr.8: 123980659-123980659 on Build GRCh38
rs886090153 Chr.8: 123980988-123980988 on Build GRCh38
rs1057028986 Chr.8: 123980630-123980630 on Build GRCh38
rs375873089 Chr.8: 123980846-123980846 on Build GRCh38
rs761706612 Chr.8: 123980629-123980629 on Build GRCh38
rs923534454 Chr.8: 123980929-123980929 on Build GRCh38
rs750047724 Chr.8: 123980808-123980808 on Build GRCh38
rs1026391512 Chr.8: 123980801-123980801 on Build GRCh38
rs143396521 Chr.8: 123981014-123981014 on Build GRCh38
rs778956217 Chr.8: 123980749-123980749 on Build GRCh38
rs778708330 Chr.8: 123980856-123980856 on Build GRCh38
rs113087741 Chr.8: 123980793-123980793 on Build GRCh38
rs950533888 Chr.8: 123980801-123980802 on Build GRCh38
rs374214825 Chr.8: 123980681-123980681 on Build GRCh38
rs964903127 Chr.8: 123980919-123980919 on Build GRCh38
rs755625470 Chr.8: 123980701-123980701 on Build GRCh38
rs934724992 Chr.8: 123980627-123980627 on Build GRCh38
rs759710529 Chr.8: 123980804-123980804 on Build GRCh38
rs756129301 Chr.8: 123981032-123981032 on Build GRCh38
rs746754207 Chr.8: 123980845-123980846 on Build GRCh38
rs765815577 Chr.8: 123980670-123980670 on Build GRCh38
rs761723061 Chr.8: 123980936-123980936 on Build GRCh38
rs576479024 Chr.8: 123980656-123980656 on Build GRCh38
rs924604024 Chr.8: 123980855-123980855 on Build GRCh38
rs749208767 Chr.8: 123980735-123980735 on Build GRCh38
rs199707703 Chr.8: 123980744-123980744 on Build GRCh38
rs539551357 Chr.8: 123980922-123980923 on Build GRCh38
rs201254129 Chr.8: 123980626-123980626 on Build GRCh38
rs764933568 Chr.8: 123980954-123980954 on Build GRCh38
rs759995131 Chr.8: 123980665-123980665 on Build GRCh38
rs190013127 Chr.8: 123980973-123980973 on Build GRCh38
rs368417039 Chr.8: 123980778-123980778 on Build GRCh38
rs923283844 Chr.8: 123980688-123980688 on Build GRCh38
rs933328316 Chr.8: 123980727-123980727 on Build GRCh38
rs762823486 Chr.8: 123981002-123981002 on Build GRCh38
rs868607365 Chr.8: 123980632-123980632 on Build GRCh38
rs201686117 Chr.8: 123980647-123980647 on Build GRCh38
rs1009871555 Chr.8: 123981030-123981030 on Build GRCh38
rs775161966 Chr.8: 123980723-123980723 on Build GRCh38
rs776361747 Chr.8: 123980932-123980932 on Build GRCh38
rs780988475 Chr.8: 123980696-123980696 on Build GRCh38
rs975528363 Chr.8: 123980830-123980830 on Build GRCh38
rs772158690 Chr.8: 123980638-123980638 on Build GRCh38
rs766267559 Chr.8: 123981011-123981011 on Build GRCh38
rs770962496 Chr.8: 123980797-123980797 on Build GRCh38
rs755861193 Chr.8: 123980822-123980822 on Build GRCh38

View Full Product Details