Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 124371479-124371974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM65
Entrez Gene ID
157378
Gene Name
transmembrane protein 65
Gene Aliases
-
UniGene
Hs.187646
Gene Chromosome Location
Chr.8: 124306189-124372699 on Build GRCh38
Transcripts
NM_194291.2 XM_011516847.2 XM_017013105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5818791 c.254G>T p.S85I Chr.8: 124371904-124371904 on Build GRCh38
5988758 c.304G>T p.E102* Chr.8: 124371854-124371854 on Build GRCh38
5964056 c.282C>T p.F94F Chr.8: 124371876-124371876 on Build GRCh38
5486649 c.188T>C p.M63T Chr.8: 124371970-124371970 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771515043 Chr.8: 124371894-124371894 on Build GRCh38
rs1008109965 Chr.8: 124371625-124371625 on Build GRCh38
rs747498258 Chr.8: 124371822-124371822 on Build GRCh38
rs973973250 Chr.8: 124371648-124371648 on Build GRCh38
rs893190852 Chr.8: 124371745-124371745 on Build GRCh38
rs17854113 Chr.8: 124371869-124371869 on Build GRCh38
rs1007810024 Chr.8: 124371608-124371608 on Build GRCh38
rs780967746 Chr.8: 124371844-124371844 on Build GRCh38
rs774630217 Chr.8: 124371569-124371569 on Build GRCh38
rs1027142995 Chr.8: 124371654-124371654 on Build GRCh38
rs868565197 Chr.8: 124371717-124371717 on Build GRCh38
rs773153140 Chr.8: 124371919-124371919 on Build GRCh38
rs532399990 Chr.8: 124371544-124371544 on Build GRCh38
rs377714284 Chr.8: 124371821-124371821 on Build GRCh38
rs540103095 Chr.8: 124371892-124371892 on Build GRCh38
rs1054656352 Chr.8: 124371744-124371744 on Build GRCh38
rs757589577 Chr.8: 124371829-124371829 on Build GRCh38
rs997239195 Chr.8: 124371605-124371605 on Build GRCh38
rs888287962 Chr.8: 124371538-124371538 on Build GRCh38
rs903366514 Chr.8: 124371519-124371519 on Build GRCh38
rs1035510180 Chr.8: 124371893-124371893 on Build GRCh38
rs998288296 Chr.8: 124371595-124371598 on Build GRCh38
rs375828904 Chr.8: 124371808-124371808 on Build GRCh38
rs897112095 Chr.8: 124371817-124371817 on Build GRCh38
rs748948229 Chr.8: 124371849-124371849 on Build GRCh38
rs575493469 Chr.8: 124371695-124371695 on Build GRCh38
rs1054952733 Chr.8: 124371592-124371592 on Build GRCh38
rs745572300 Chr.8: 124371843-124371844 on Build GRCh38
rs891132991 Chr.8: 124371792-124371792 on Build GRCh38
rs896949770 Chr.8: 124371733-124371733 on Build GRCh38
rs959442141 Chr.8: 124371912-124371912 on Build GRCh38
rs770602257 Chr.8: 124371832-124371833 on Build GRCh38
rs937976099 Chr.8: 124371710-124371710 on Build GRCh38
rs1054930192 Chr.8: 124371502-124371502 on Build GRCh38
rs530434004 Chr.8: 124371807-124371807 on Build GRCh38
rs934816395 Chr.8: 124371736-124371736 on Build GRCh38
rs1015346141 Chr.8: 124371635-124371635 on Build GRCh38
rs547881259 Chr.8: 124371716-124371716 on Build GRCh38
rs781696177 Chr.8: 124371839-124371839 on Build GRCh38
rs1045910194 Chr.8: 124371515-124371515 on Build GRCh38
rs768482902 Chr.8: 124371859-124371859 on Build GRCh38
rs1017833873 Chr.8: 124371611-124371611 on Build GRCh38
rs759739872 Chr.8: 124371952-124371952 on Build GRCh38
rs865883136 Chr.8: 124371732-124371732 on Build GRCh38
rs374732240 Chr.8: 124371943-124371943 on Build GRCh38
rs990764465 Chr.8: 124371915-124371915 on Build GRCh38
rs891016485 Chr.8: 124371616-124371616 on Build GRCh38
rs982583040 Chr.8: 124371682-124371682 on Build GRCh38
rs961974932 Chr.8: 124371760-124371760 on Build GRCh38
rs959359529 Chr.8: 124371825-124371825 on Build GRCh38
rs1003638973 Chr.8: 124371833-124371833 on Build GRCh38
rs1012012470 Chr.8: 124371884-124371884 on Build GRCh38
rs1028346540 Chr.8: 124371614-124371614 on Build GRCh38
rs371357144 Chr.8: 124371942-124371942 on Build GRCh38
rs941222815 Chr.8: 124371731-124371731 on Build GRCh38
rs1026843339 Chr.8: 124371823-124371823 on Build GRCh38
rs981700248 Chr.8: 124371724-124371724 on Build GRCh38
rs1051244590 Chr.8: 124371743-124371743 on Build GRCh38
rs765491688 Chr.8: 124371955-124371955 on Build GRCh38
rs1050161551 Chr.8: 124371781-124371781 on Build GRCh38
rs567770464 Chr.8: 124371759-124371759 on Build GRCh38
rs552117914 Chr.8: 124371680-124371680 on Build GRCh38
rs199888218 Chr.8: 124371530-124371530 on Build GRCh38
rs893262312 Chr.8: 124371593-124371593 on Build GRCh38
rs1029343036 Chr.8: 124371604-124371604 on Build GRCh38
rs528218470 Chr.8: 124371713-124371713 on Build GRCh38
rs563642391 Chr.8: 124371646-124371646 on Build GRCh38
rs950252757 Chr.8: 124371722-124371722 on Build GRCh38
rs1021080386 Chr.8: 124371675-124371675 on Build GRCh38
rs760543032 Chr.8: 124371936-124371936 on Build GRCh38
rs867221635 Chr.8: 124371911-124371911 on Build GRCh38

View Full Product Details