Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21201063-21201602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNA7
Entrez Gene ID
3444
Gene Name
interferon alpha 7
Gene Aliases
IFN-alphaJ, IFNA-J
UniGene
Hs.282274
Gene Chromosome Location
Chr.9: 21201469-21202205 on Build GRCh38
Transcripts
NM_021057.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1107795 c.566A>G p.D189G Chr.9: 21201600-21201600 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs561281305 Chr.9: 21201445-21201445 on Build GRCh38
rs969696846 Chr.9: 21201303-21201303 on Build GRCh38
rs752706041 Chr.9: 21201266-21201266 on Build GRCh38
rs151015471 Chr.9: 21201213-21201216 on Build GRCh38
rs911691974 Chr.9: 21201346-21201346 on Build GRCh38
rs190358578 Chr.9: 21201537-21201537 on Build GRCh38
rs946992571 Chr.9: 21201225-21201225 on Build GRCh38
rs574856908 Chr.9: 21201454-21201454 on Build GRCh38
rs879928898 Chr.9: 21201397-21201397 on Build GRCh38
rs556158221 Chr.9: 21201289-21201289 on Build GRCh38
rs377417687 Chr.9: 21201094-21201094 on Build GRCh38
rs185113939 Chr.9: 21201362-21201362 on Build GRCh38
rs902661923 Chr.9: 21201154-21201154 on Build GRCh38
rs969978540 Chr.9: 21201186-21201186 on Build GRCh38
rs920448969 Chr.9: 21201194-21201194 on Build GRCh38
rs1011084678 Chr.9: 21201128-21201128 on Build GRCh38
rs536565308 Chr.9: 21201274-21201274 on Build GRCh38
rs999050658 Chr.9: 21201161-21201161 on Build GRCh38
rs767132946 Chr.9: 21201578-21201578 on Build GRCh38
rs187755490 Chr.9: 21201214-21201214 on Build GRCh38
rs1043980006 Chr.9: 21201227-21201227 on Build GRCh38
rs1022913215 Chr.9: 21201143-21201143 on Build GRCh38
rs1044169599 Chr.9: 21201497-21201497 on Build GRCh38
rs944754233 Chr.9: 21201215-21201215 on Build GRCh38
rs905563749 Chr.9: 21201517-21201517 on Build GRCh38
rs973305323 Chr.9: 21201193-21201193 on Build GRCh38
rs1056646877 Chr.9: 21201451-21201451 on Build GRCh38
rs546990662 Chr.9: 21201229-21201229 on Build GRCh38
rs879174851 Chr.9: 21201324-21201326 on Build GRCh38
rs572619494 Chr.9: 21201377-21201377 on Build GRCh38
rs111710664 Chr.9: 21201088-21201088 on Build GRCh38
rs538494759 Chr.9: 21201360-21201360 on Build GRCh38
rs879436453 Chr.9: 21201396-21201396 on Build GRCh38
rs1022611823 Chr.9: 21201180-21201180 on Build GRCh38
rs193296571 Chr.9: 21201236-21201236 on Build GRCh38
rs35605417 Chr.9: 21201088-21201089 on Build GRCh38
rs761467280 Chr.9: 21201577-21201577 on Build GRCh38
rs367543507 Chr.9: 21201575-21201575 on Build GRCh38
rs994521835 Chr.9: 21201556-21201556 on Build GRCh38
rs201255229 Chr.9: 21201257-21201257 on Build GRCh38
rs1022667488 Chr.9: 21201290-21201290 on Build GRCh38
rs900767161 Chr.9: 21201121-21201121 on Build GRCh38
rs762618537 Chr.9: 21201557-21201557 on Build GRCh38
rs551912920 Chr.9: 21201408-21201408 on Build GRCh38
rs1027565543 Chr.9: 21201317-21201317 on Build GRCh38
rs1031409736 Chr.9: 21201175-21201175 on Build GRCh38
rs902764566 Chr.9: 21201398-21201398 on Build GRCh38
rs543776359 Chr.9: 21201508-21201508 on Build GRCh38
rs1038978909 Chr.9: 21201096-21201096 on Build GRCh38
rs1014038241 Chr.9: 21201479-21201479 on Build GRCh38
rs983309087 Chr.9: 21201334-21201334 on Build GRCh38
rs1044082395 Chr.9: 21201390-21201390 on Build GRCh38
rs929154739 Chr.9: 21201212-21201212 on Build GRCh38
rs74560208 Chr.9: 21201311-21201311 on Build GRCh38
rs571047974 Chr.9: 21201188-21201188 on Build GRCh38
rs950658855 Chr.9: 21201333-21201333 on Build GRCh38
rs532831048 Chr.9: 21201548-21201548 on Build GRCh38
rs999705463 Chr.9: 21201233-21201233 on Build GRCh38
rs900497008 Chr.9: 21201223-21201224 on Build GRCh38
rs944820957 Chr.9: 21201354-21201354 on Build GRCh38
rs1039029461 Chr.9: 21201219-21201220 on Build GRCh38
rs992495415 Chr.9: 21201176-21201176 on Build GRCh38
rs746817479 Chr.9: 21201499-21201499 on Build GRCh38
rs552722099 Chr.9: 21201151-21201151 on Build GRCh38
rs775380136 Chr.9: 21201553-21201553 on Build GRCh38
rs774375139 Chr.9: 21201576-21201576 on Build GRCh38

View Full Product Details