Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 32785190-32785689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM215
Entrez Gene ID
401498
Gene Name
transmembrane protein 215
Gene Aliases
-
UniGene
Hs.522063
Gene Chromosome Location
Chr.9: 32783499-32789201 on Build GRCh38
Transcripts
NM_212558.2

SNP Detail

SNP ID Chromosome Location
rs997712408 Chr.9: 32785292-32785292 on Build GRCh38
rs988173809 Chr.9: 32785624-32785624 on Build GRCh38
rs35253178 Chr.9: 32785274-32785275 on Build GRCh38
rs943312741 Chr.9: 32785573-32785573 on Build GRCh38
rs943099027 Chr.9: 32785383-32785384 on Build GRCh38
rs644337 Chr.9: 32785401-32785401 on Build GRCh38
rs866827982 Chr.9: 32785274-32785274 on Build GRCh38
rs541788379 Chr.9: 32785224-32785224 on Build GRCh38
rs1009678498 Chr.9: 32785665-32785665 on Build GRCh38
rs532919173 Chr.9: 32785444-32785444 on Build GRCh38
rs928051 Chr.9: 32785493-32785493 on Build GRCh38
rs891572825 Chr.9: 32785657-32785657 on Build GRCh38
rs563436856 Chr.9: 32785529-32785529 on Build GRCh38
rs901473439 Chr.9: 32785591-32785591 on Build GRCh38
rs939368890 Chr.9: 32785430-32785430 on Build GRCh38
rs958259340 Chr.9: 32785592-32785592 on Build GRCh38
rs929099669 Chr.9: 32785476-32785476 on Build GRCh38
rs549376413 Chr.9: 32785590-32785590 on Build GRCh38
rs375005109 Chr.9: 32785470-32785470 on Build GRCh38
rs1022753597 Chr.9: 32785533-32785533 on Build GRCh38
rs184709637 Chr.9: 32785325-32785325 on Build GRCh38
rs139619571 Chr.9: 32785564-32785564 on Build GRCh38
rs1037491687 Chr.9: 32785388-32785388 on Build GRCh38
rs992209628 Chr.9: 32785433-32785433 on Build GRCh38
rs968735144 Chr.9: 32785373-32785373 on Build GRCh38
rs77132361 Chr.9: 32785356-32785356 on Build GRCh38
rs1007206988 Chr.9: 32785243-32785243 on Build GRCh38
rs914123489 Chr.9: 32785632-32785632 on Build GRCh38
rs569327054 Chr.9: 32785259-32785259 on Build GRCh38
rs565910749 Chr.9: 32785601-32785601 on Build GRCh38
rs868643541 Chr.9: 32785620-32785620 on Build GRCh38
rs758156023 Chr.9: 32785302-32785302 on Build GRCh38
rs1017714946 Chr.9: 32785244-32785244 on Build GRCh38
rs1022485753 Chr.9: 32785368-32785368 on Build GRCh38
rs980788634 Chr.9: 32785384-32785384 on Build GRCh38
rs933447350 Chr.9: 32785324-32785324 on Build GRCh38
rs936719524 Chr.9: 32785253-32785253 on Build GRCh38
rs997154625 Chr.9: 32785647-32785647 on Build GRCh38
rs1031361060 Chr.9: 32785650-32785650 on Build GRCh38

View Full Product Details