Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 16153332-16153836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRPR
Entrez Gene ID
2925
Gene Name
gastrin releasing peptide receptor
Gene Aliases
BB2, BB2R
UniGene
Hs.567282
Gene Chromosome Location
Chr.X: 16123301-16153518 on Build GRCh38
Transcripts
NM_005314.2

SNP Detail

SNP ID Chromosome Location
rs867469009 Chr.X: 16153487-16153487 on Build GRCh38
rs908420984 Chr.X: 16153726-16153726 on Build GRCh38
rs141185951 Chr.X: 16153398-16153398 on Build GRCh38
rs751464091 Chr.X: 16153480-16153480 on Build GRCh38
rs956273642 Chr.X: 16153548-16153548 on Build GRCh38
rs996501568 Chr.X: 16153773-16153773 on Build GRCh38
rs914564277 Chr.X: 16153635-16153635 on Build GRCh38
rs758903401 Chr.X: 16153481-16153481 on Build GRCh38
rs927286388 Chr.X: 16153468-16153468 on Build GRCh38
rs983607410 Chr.X: 16153436-16153436 on Build GRCh38
rs1049864129 Chr.X: 16153782-16153782 on Build GRCh38
rs890406709 Chr.X: 16153805-16153805 on Build GRCh38
rs971672165 Chr.X: 16153443-16153443 on Build GRCh38
rs189077984 Chr.X: 16153421-16153421 on Build GRCh38
rs1054488942 Chr.X: 16153622-16153622 on Build GRCh38
rs762052527 Chr.X: 16153674-16153676 on Build GRCh38
rs181148549 Chr.X: 16153489-16153489 on Build GRCh38
rs777286330 Chr.X: 16153459-16153459 on Build GRCh38
rs776043913 Chr.X: 16153485-16153485 on Build GRCh38
rs990425183 Chr.X: 16153612-16153612 on Build GRCh38
rs913290945 Chr.X: 16153654-16153654 on Build GRCh38
rs1038277736 Chr.X: 16153755-16153755 on Build GRCh38
rs939920959 Chr.X: 16153730-16153730 on Build GRCh38
rs778864899 Chr.X: 16153362-16153362 on Build GRCh38
rs1032291438 Chr.X: 16153502-16153502 on Build GRCh38
rs1056290650 Chr.X: 16153718-16153718 on Build GRCh38
rs750486687 Chr.X: 16153736-16153736 on Build GRCh38
rs11340309 Chr.X: 16153472-16153472 on Build GRCh38
rs945212010 Chr.X: 16153682-16153682 on Build GRCh38
rs746548176 Chr.X: 16153471-16153472 on Build GRCh38
rs201413173 Chr.X: 16153527-16153530 on Build GRCh38
rs767683867 Chr.X: 16153717-16153717 on Build GRCh38
rs964466479 Chr.X: 16153357-16153357 on Build GRCh38
rs1027845138 Chr.X: 16153420-16153420 on Build GRCh38
rs12019422 Chr.X: 16153787-16153787 on Build GRCh38
rs898430706 Chr.X: 16153772-16153772 on Build GRCh38
rs980530522 Chr.X: 16153681-16153681 on Build GRCh38
rs774580957 Chr.X: 16153653-16153653 on Build GRCh38
rs202120714 Chr.X: 16153472-16153472 on Build GRCh38
rs774962356 Chr.X: 16153356-16153356 on Build GRCh38
rs917495058 Chr.X: 16153437-16153437 on Build GRCh38
rs980546739 Chr.X: 16153446-16153446 on Build GRCh38

View Full Product Details