Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21972456-21973003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMS
Entrez Gene ID
6611
Gene Name
spermine synthase
Gene Aliases
MRSR, SPMSY, SRS, SpS
UniGene
Hs.724874
Gene Chromosome Location
Chr.X: 21940573-21994837 on Build GRCh38
Transcripts
NM_001258423.1 NM_004595.4 XM_005274582.2 XM_011545568.2 XM_017029753.1 XM_017029754.1 XM_017029755.1

SNP Detail

SNP ID Chromosome Location
rs915491995 Chr.X: 21972867-21972867 on Build GRCh38
rs969851824 Chr.X: 21972805-21972805 on Build GRCh38
rs397515381 Chr.X: 21972576-21972576 on Build GRCh38
rs776471342 Chr.X: 21972648-21972648 on Build GRCh38
rs185759750 Chr.X: 21972607-21972607 on Build GRCh38
rs774131287 Chr.X: 21972543-21972543 on Build GRCh38
rs948202798 Chr.X: 21972628-21972628 on Build GRCh38
rs767100593 Chr.X: 21972552-21972552 on Build GRCh38
rs898818351 Chr.X: 21972583-21972583 on Build GRCh38
rs956911093 Chr.X: 21972773-21972773 on Build GRCh38
rs140953114 Chr.X: 21972635-21972635 on Build GRCh38
rs188103923 Chr.X: 21972657-21972657 on Build GRCh38
rs1050016021 Chr.X: 21972893-21972893 on Build GRCh38
rs752713559 Chr.X: 21972560-21972560 on Build GRCh38
rs756990950 Chr.X: 21972580-21972580 on Build GRCh38
rs76666762 Chr.X: 21972941-21972941 on Build GRCh38
rs981181175 Chr.X: 21972917-21972923 on Build GRCh38
rs140373740 Chr.X: 21972748-21972748 on Build GRCh38
rs753045110 Chr.X: 21972559-21972561 on Build GRCh38
rs778410949 Chr.X: 21972813-21972813 on Build GRCh38
rs946388494 Chr.X: 21972873-21972873 on Build GRCh38
rs1023730468 Chr.X: 21972801-21972801 on Build GRCh38
rs759353187 Chr.X: 21972819-21972819 on Build GRCh38
rs988834062 Chr.X: 21972857-21972857 on Build GRCh38
rs775940570 Chr.X: 21972713-21972713 on Build GRCh38
rs59033470 Chr.X: 21972724-21972724 on Build GRCh38
rs759163057 Chr.X: 21972551-21972551 on Build GRCh38
rs993849779 Chr.X: 21972738-21972738 on Build GRCh38
rs764246500 Chr.X: 21972947-21972947 on Build GRCh38
rs766638145 Chr.X: 21972581-21972581 on Build GRCh38
rs766665397 Chr.X: 21972684-21972684 on Build GRCh38
rs1004799847 Chr.X: 21972512-21972512 on Build GRCh38
rs774370390 Chr.X: 21972818-21972818 on Build GRCh38
rs749724022 Chr.X: 21972832-21972832 on Build GRCh38
rs755082527 Chr.X: 21972618-21972618 on Build GRCh38
rs984081258 Chr.X: 21972784-21972784 on Build GRCh38
rs866381068 Chr.X: 21972916-21972916 on Build GRCh38
rs770681620 Chr.X: 21972542-21972542 on Build GRCh38
rs772998822 Chr.X: 21972737-21972737 on Build GRCh38
rs944477076 Chr.X: 21972964-21972964 on Build GRCh38
rs969390687 Chr.X: 21972879-21972879 on Build GRCh38
rs112405183 Chr.X: 21972722-21972722 on Build GRCh38
rs746297985 Chr.X: 21972561-21972561 on Build GRCh38
rs1050172608 Chr.X: 21972695-21972695 on Build GRCh38
rs151144042 Chr.X: 21972746-21972746 on Build GRCh38
rs982651698 Chr.X: 21972634-21972634 on Build GRCh38
rs888826833 Chr.X: 21972700-21972700 on Build GRCh38
rs1009033660 Chr.X: 21972656-21972656 on Build GRCh38
rs1046364018 Chr.X: 21972945-21972945 on Build GRCh38
rs747422871 Chr.X: 21972637-21972637 on Build GRCh38
rs751710136 Chr.X: 21972690-21972690 on Build GRCh38
rs1022515074 Chr.X: 21972877-21972877 on Build GRCh38
rs757527743 Chr.X: 21972833-21972833 on Build GRCh38
rs753600774 Chr.X: 21972579-21972579 on Build GRCh38
rs910320736 Chr.X: 21972931-21972931 on Build GRCh38
rs765694930 Chr.X: 21972481-21972481 on Build GRCh38

View Full Product Details