Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 35955839-35956338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP47
Entrez Gene ID
286464
Gene Name
cilia and flagella associated protein 47
Gene Aliases
CHDC2, CXorf22, CXorf30, CXorf59
UniGene
Hs.376425
Gene Chromosome Location
Chr.X: 35919734-36385319 on Build GRCh38
Transcripts
NM_001304548.1 NM_152632.3 XM_017029452.1 XM_017029453.1

SNP Detail

SNP ID Chromosome Location
rs367971605 Chr.X: 35956216-35956216 on Build GRCh38
rs978397752 Chr.X: 35955926-35955926 on Build GRCh38
rs774554548 Chr.X: 35955868-35955868 on Build GRCh38
rs779697282 Chr.X: 35955928-35955928 on Build GRCh38
rs753438645 Chr.X: 35956235-35956235 on Build GRCh38
rs776735297 Chr.X: 35955949-35955949 on Build GRCh38
rs192233472 Chr.X: 35956123-35956123 on Build GRCh38
rs752975745 Chr.X: 35956055-35956055 on Build GRCh38
rs757261016 Chr.X: 35956103-35956103 on Build GRCh38
rs779702508 Chr.X: 35955883-35955883 on Build GRCh38
rs771838651 Chr.X: 35955912-35955912 on Build GRCh38
rs748742312 Chr.X: 35956190-35956190 on Build GRCh38
rs1000079907 Chr.X: 35956261-35956261 on Build GRCh38
rs1019980942 Chr.X: 35956217-35956217 on Build GRCh38
rs772595042 Chr.X: 35956146-35956146 on Build GRCh38
rs748110121 Chr.X: 35956143-35956143 on Build GRCh38
rs761074842 Chr.X: 35956071-35956071 on Build GRCh38
rs1036337924 Chr.X: 35955995-35955995 on Build GRCh38
rs374417101 Chr.X: 35955937-35955937 on Build GRCh38
rs371807838 Chr.X: 35956072-35956072 on Build GRCh38
rs750944152 Chr.X: 35956085-35956085 on Build GRCh38
rs750570659 Chr.X: 35956105-35956105 on Build GRCh38
rs747345059 Chr.X: 35955903-35955903 on Build GRCh38
rs748871201 Chr.X: 35956157-35956157 on Build GRCh38
rs148122184 Chr.X: 35956262-35956262 on Build GRCh38
rs746754951 Chr.X: 35956108-35956108 on Build GRCh38
rs769829112 Chr.X: 35956144-35956144 on Build GRCh38
rs1050087423 Chr.X: 35956139-35956139 on Build GRCh38
rs767870539 Chr.X: 35956043-35956043 on Build GRCh38
rs961280514 Chr.X: 35956312-35956312 on Build GRCh38
rs772864004 Chr.X: 35956020-35956020 on Build GRCh38
rs142572048 Chr.X: 35955968-35955968 on Build GRCh38
rs766123614 Chr.X: 35956052-35956052 on Build GRCh38
rs762291354 Chr.X: 35956206-35956206 on Build GRCh38
rs758530475 Chr.X: 35956245-35956245 on Build GRCh38
rs960758381 Chr.X: 35955935-35955935 on Build GRCh38
rs754373831 Chr.X: 35956100-35956100 on Build GRCh38
rs200890203 Chr.X: 35956223-35956223 on Build GRCh38
rs373129355 Chr.X: 35956124-35956124 on Build GRCh38
rs746078040 Chr.X: 35955910-35955910 on Build GRCh38
rs770466227 Chr.X: 35956172-35956172 on Build GRCh38
rs139640560 Chr.X: 35956174-35956174 on Build GRCh38
rs762258408 Chr.X: 35955907-35955907 on Build GRCh38
rs150580241 Chr.X: 35956104-35956104 on Build GRCh38
rs201028860 Chr.X: 35955967-35955967 on Build GRCh38
rs760986914 Chr.X: 35956187-35956187 on Build GRCh38
rs758542190 Chr.X: 35956106-35956106 on Build GRCh38
rs371126181 Chr.X: 35955932-35955932 on Build GRCh38
rs780374404 Chr.X: 35956107-35956107 on Build GRCh38
rs777060079 Chr.X: 35956203-35956203 on Build GRCh38

View Full Product Details