Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37441892-37442394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRRG1
Entrez Gene ID
5638
Gene Name
proline rich and Gla domain 1
Gene Aliases
PRGP1
UniGene
Hs.190341
Gene Chromosome Location
Chr.X: 37349275-37457295 on Build GRCh38
Transcripts
NM_000950.2 NM_001142395.1 NM_001173486.1 NM_001173489.1 NM_001173490.1

SNP Detail

SNP ID Chromosome Location
rs141395433 Chr.X: 37442321-37442321 on Build GRCh38
rs923258494 Chr.X: 37442092-37442092 on Build GRCh38
rs1039091461 Chr.X: 37442085-37442085 on Build GRCh38
rs992310641 Chr.X: 37441915-37441915 on Build GRCh38
rs891983363 Chr.X: 37442104-37442104 on Build GRCh38
rs782190507 Chr.X: 37442060-37442060 on Build GRCh38
rs41309725 Chr.X: 37442098-37442098 on Build GRCh38
rs1001739232 Chr.X: 37442237-37442237 on Build GRCh38
rs919099057 Chr.X: 37442005-37442005 on Build GRCh38
rs933426774 Chr.X: 37442095-37442095 on Build GRCh38
rs188356334 Chr.X: 37442131-37442131 on Build GRCh38
rs781868135 Chr.X: 37442053-37442053 on Build GRCh38
rs185914413 Chr.X: 37442280-37442280 on Build GRCh38
rs782096068 Chr.X: 37441912-37441912 on Build GRCh38
rs782085309 Chr.X: 37442135-37442135 on Build GRCh38
rs984737854 Chr.X: 37442018-37442018 on Build GRCh38
rs781972773 Chr.X: 37442132-37442132 on Build GRCh38
rs782759326 Chr.X: 37442030-37442030 on Build GRCh38
rs782818991 Chr.X: 37442121-37442121 on Build GRCh38
rs782506416 Chr.X: 37442291-37442291 on Build GRCh38
rs950581093 Chr.X: 37442016-37442016 on Build GRCh38
rs181074545 Chr.X: 37442156-37442156 on Build GRCh38

View Full Product Details