Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 44844695-44845204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP21
Entrez Gene ID
63904
Gene Name
dual specificity phosphatase 21
Gene Aliases
LMWDSP21
UniGene
Hs.534478
Gene Chromosome Location
Chr.X: 44844003-44844888 on Build GRCh38
Transcripts
NM_022076.3

SNP Detail

SNP ID Chromosome Location
rs374977948 Chr.X: 44844737-44844737 on Build GRCh38
rs770084342 Chr.X: 44844810-44844810 on Build GRCh38
rs761933280 Chr.X: 44845073-44845073 on Build GRCh38
rs186695660 Chr.X: 44845167-44845167 on Build GRCh38
rs370301001 Chr.X: 44844963-44844963 on Build GRCh38
rs190373105 Chr.X: 44844923-44844923 on Build GRCh38
rs968746592 Chr.X: 44844945-44844945 on Build GRCh38
rs182564153 Chr.X: 44844926-44844926 on Build GRCh38
rs917685304 Chr.X: 44845110-44845110 on Build GRCh38
rs919486728 Chr.X: 44845149-44845149 on Build GRCh38
rs979564319 Chr.X: 44845000-44845000 on Build GRCh38
rs527510923 Chr.X: 44845101-44845101 on Build GRCh38
rs1029770933 Chr.X: 44844782-44844782 on Build GRCh38
rs766807491 Chr.X: 44845040-44845040 on Build GRCh38
rs955373685 Chr.X: 44844792-44844792 on Build GRCh38
rs764170409 Chr.X: 44844739-44844739 on Build GRCh38
rs760652493 Chr.X: 44844738-44844738 on Build GRCh38
rs149043782 Chr.X: 44844729-44844729 on Build GRCh38
rs142059967 Chr.X: 44845009-44845009 on Build GRCh38
rs959548458 Chr.X: 44845038-44845038 on Build GRCh38
rs911981900 Chr.X: 44844875-44844875 on Build GRCh38
rs774503508 Chr.X: 44844725-44844725 on Build GRCh38
rs753381856 Chr.X: 44844752-44844752 on Build GRCh38
rs1010013795 Chr.X: 44844846-44844846 on Build GRCh38
rs745500570 Chr.X: 44844789-44844789 on Build GRCh38
rs771130317 Chr.X: 44844722-44844722 on Build GRCh38
rs772113148 Chr.X: 44845146-44845146 on Build GRCh38
rs186102846 Chr.X: 44844879-44844879 on Build GRCh38

View Full Product Details