Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49076406-49076914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR45
Entrez Gene ID
11152
Gene Name
WD repeat domain 45
Gene Aliases
JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
UniGene
Hs.632807
Gene Chromosome Location
Chr.X: 49074433-49101121 on Build GRCh38
Transcripts
NM_001029896.1 NM_007075.3

SNP Detail

SNP ID Chromosome Location
rs797046101 Chr.X: 49076469-49076469 on Build GRCh38
rs782245609 Chr.X: 49076775-49076775 on Build GRCh38
rs983618032 Chr.X: 49076843-49076843 on Build GRCh38
rs782350058 Chr.X: 49076544-49076544 on Build GRCh38
rs782643042 Chr.X: 49076502-49076502 on Build GRCh38
rs782473996 Chr.X: 49076768-49076768 on Build GRCh38
rs782302447 Chr.X: 49076514-49076514 on Build GRCh38
rs1027902150 Chr.X: 49076829-49076829 on Build GRCh38
rs387907331 Chr.X: 49076432-49076433 on Build GRCh38
rs377212055 Chr.X: 49076601-49076601 on Build GRCh38
rs782399651 Chr.X: 49076491-49076491 on Build GRCh38
rs142809324 Chr.X: 49076736-49076736 on Build GRCh38
rs35492870 Chr.X: 49076710-49076711 on Build GRCh38
rs782211949 Chr.X: 49076478-49076478 on Build GRCh38
rs782674726 Chr.X: 49076840-49076840 on Build GRCh38
rs782701302 Chr.X: 49076569-49076569 on Build GRCh38
rs372347765 Chr.X: 49076567-49076567 on Build GRCh38
rs782660155 Chr.X: 49076623-49076623 on Build GRCh38
rs782183202 Chr.X: 49076845-49076845 on Build GRCh38
rs782069953 Chr.X: 49076559-49076559 on Build GRCh38
rs41310595 Chr.X: 49076732-49076732 on Build GRCh38
rs782052651 Chr.X: 49076875-49076875 on Build GRCh38
rs782511044 Chr.X: 49076579-49076579 on Build GRCh38
rs782395953 Chr.X: 49076654-49076654 on Build GRCh38
rs140596058 Chr.X: 49076515-49076515 on Build GRCh38
rs782259993 Chr.X: 49076866-49076866 on Build GRCh38
rs373218492 Chr.X: 49076765-49076765 on Build GRCh38
rs948285795 Chr.X: 49076564-49076564 on Build GRCh38
rs372437815 Chr.X: 49076657-49076657 on Build GRCh38
rs782666101 Chr.X: 49076472-49076472 on Build GRCh38
rs375257631 Chr.X: 49076748-49076748 on Build GRCh38
rs782290088 Chr.X: 49076658-49076658 on Build GRCh38
rs1052492642 Chr.X: 49076615-49076615 on Build GRCh38
rs147437546 Chr.X: 49076729-49076729 on Build GRCh38
rs782442652 Chr.X: 49076846-49076846 on Build GRCh38
rs782171399 Chr.X: 49076521-49076521 on Build GRCh38
rs373717091 Chr.X: 49076445-49076445 on Build GRCh38
rs367609966 Chr.X: 49076551-49076551 on Build GRCh38
rs781805068 Chr.X: 49076474-49076474 on Build GRCh38
rs142923330 Chr.X: 49076665-49076665 on Build GRCh38
rs369672076 Chr.X: 49076626-49076626 on Build GRCh38
rs782723092 Chr.X: 49076477-49076477 on Build GRCh38
rs782434987 Chr.X: 49076476-49076476 on Build GRCh38
rs781840546 Chr.X: 49076574-49076574 on Build GRCh38
rs781951274 Chr.X: 49076548-49076548 on Build GRCh38
rs200325764 Chr.X: 49076558-49076558 on Build GRCh38
rs782794242 Chr.X: 49076798-49076798 on Build GRCh38
rs781930776 Chr.X: 49076520-49076520 on Build GRCh38
rs188389490 Chr.X: 49076789-49076789 on Build GRCh38
rs960384812 Chr.X: 49076888-49076888 on Build GRCh38
rs782558946 Chr.X: 49076609-49076609 on Build GRCh38
rs781985024 Chr.X: 49076655-49076655 on Build GRCh38
rs782760682 Chr.X: 49076553-49076553 on Build GRCh38
rs782725867 Chr.X: 49076670-49076670 on Build GRCh38
rs376936511 Chr.X: 49076649-49076649 on Build GRCh38
rs139837168 Chr.X: 49076730-49076730 on Build GRCh38
rs372370133 Chr.X: 49076627-49076627 on Build GRCh38
rs907563420 Chr.X: 49076435-49076435 on Build GRCh38
rs782329150 Chr.X: 49076660-49076660 on Build GRCh38
rs781869774 Chr.X: 49076463-49076463 on Build GRCh38
rs782643743 Chr.X: 49076770-49076770 on Build GRCh38
rs369310756 Chr.X: 49076661-49076661 on Build GRCh38
rs144081442 Chr.X: 49076468-49076468 on Build GRCh38
rs782706648 Chr.X: 49076737-49076737 on Build GRCh38
rs782631059 Chr.X: 49076629-49076629 on Build GRCh38
rs782598566 Chr.X: 49076853-49076853 on Build GRCh38
rs781998959 Chr.X: 49076694-49076694 on Build GRCh38
rs782485581 Chr.X: 49076596-49076596 on Build GRCh38

View Full Product Details