Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107981974-107982478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX13B
Entrez Gene ID
56156
Gene Name
testis expressed 13B
Gene Aliases
TGC3B, TSGA5
UniGene
Hs.333130
Gene Chromosome Location
Chr.X: 107980864-107982370 on Build GRCh38
Transcripts
NM_031273.2

SNP Detail

SNP ID Chromosome Location
rs924674998 Chr.X: 107982298-107982298 on Build GRCh38
rs1050143027 Chr.X: 107982404-107982404 on Build GRCh38
rs1008986803 Chr.X: 107981996-107981996 on Build GRCh38
rs111779279 Chr.X: 107982187-107982187 on Build GRCh38
rs755196323 Chr.X: 107982197-107982197 on Build GRCh38
rs1053003056 Chr.X: 107982455-107982455 on Build GRCh38
rs866825302 Chr.X: 107982091-107982091 on Build GRCh38
rs761872189 Chr.X: 107982131-107982131 on Build GRCh38
rs376883010 Chr.X: 107982086-107982086 on Build GRCh38
rs757268120 Chr.X: 107982031-107982031 on Build GRCh38
rs758061329 Chr.X: 107982014-107982014 on Build GRCh38
rs548824099 Chr.X: 107982062-107982062 on Build GRCh38
rs762208184 Chr.X: 107982080-107982080 on Build GRCh38
rs181132670 Chr.X: 107982330-107982330 on Build GRCh38
rs763773978 Chr.X: 107982136-107982136 on Build GRCh38
rs766461736 Chr.X: 107982110-107982110 on Build GRCh38
rs762655640 Chr.X: 107982201-107982201 on Build GRCh38
rs760698818 Chr.X: 107982109-107982109 on Build GRCh38
rs772187067 Chr.X: 107982417-107982417 on Build GRCh38
rs961977397 Chr.X: 107982383-107982383 on Build GRCh38
rs751410554 Chr.X: 107982139-107982139 on Build GRCh38
rs935937445 Chr.X: 107982416-107982416 on Build GRCh38
rs768900707 Chr.X: 107982073-107982073 on Build GRCh38
rs769727918 Chr.X: 107982427-107982427 on Build GRCh38
rs386826825 Chr.X: 107982349-107982350 on Build GRCh38
rs6616672 Chr.X: 107982350-107982350 on Build GRCh38
rs977471775 Chr.X: 107982285-107982285 on Build GRCh38
rs974799693 Chr.X: 107982386-107982386 on Build GRCh38
rs1010011952 Chr.X: 107982121-107982121 on Build GRCh38
rs751438016 Chr.X: 107982028-107982028 on Build GRCh38
rs1022277002 Chr.X: 107982132-107982132 on Build GRCh38
rs892710332 Chr.X: 107982064-107982064 on Build GRCh38
rs958529390 Chr.X: 107982190-107982190 on Build GRCh38
rs147421606 Chr.X: 107982061-107982061 on Build GRCh38
rs748387662 Chr.X: 107982227-107982227 on Build GRCh38
rs781222097 Chr.X: 107982164-107982164 on Build GRCh38
rs764785807 Chr.X: 107982181-107982181 on Build GRCh38
rs754000945 Chr.X: 107982124-107982124 on Build GRCh38
rs370276150 Chr.X: 107982166-107982166 on Build GRCh38
rs41312560 Chr.X: 107982270-107982270 on Build GRCh38
rs6622278 Chr.X: 107982349-107982349 on Build GRCh38
rs199958232 Chr.X: 107982085-107982085 on Build GRCh38
rs186016130 Chr.X: 107982400-107982400 on Build GRCh38
rs920279633 Chr.X: 107982394-107982394 on Build GRCh38
rs779267340 Chr.X: 107982220-107982220 on Build GRCh38
rs147223587 Chr.X: 107982217-107982217 on Build GRCh38
rs748977461 Chr.X: 107982066-107982066 on Build GRCh38
rs777689678 Chr.X: 107982017-107982017 on Build GRCh38
rs917189881 Chr.X: 107982347-107982347 on Build GRCh38
rs972166349 Chr.X: 107982392-107982392 on Build GRCh38
rs774841881 Chr.X: 107982079-107982079 on Build GRCh38
rs780814728 Chr.X: 107982059-107982059 on Build GRCh38
rs36125529 Chr.X: 107982282-107982282 on Build GRCh38
rs755614844 Chr.X: 107982173-107982173 on Build GRCh38
rs910299402 Chr.X: 107982415-107982415 on Build GRCh38
rs935876213 Chr.X: 107982042-107982042 on Build GRCh38
rs968890962 Chr.X: 107982276-107982276 on Build GRCh38
rs968015141 Chr.X: 107982138-107982138 on Build GRCh38

View Full Product Details