Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 182383119-182383618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLUL
Entrez Gene ID
2752
Gene Name
glutamate-ammonia ligase
Gene Aliases
GLNS, GS, PIG43, PIG59
UniGene
Hs.132016
Gene Chromosome Location
Chr.1: 182378093-182392206 on Build GRCh38
Transcripts
NM_001033044.3 NM_001033056.3 NM_002065.6 XM_006711278.1

SNP Detail

SNP ID Chromosome Location
rs764305199 Chr.1: 182383286-182383286 on Build GRCh38
rs549158693 Chr.1: 182383189-182383189 on Build GRCh38
rs571517535 Chr.1: 182383348-182383348 on Build GRCh38
rs886045610 Chr.1: 182383555-182383555 on Build GRCh38
rs1041043204 Chr.1: 182383492-182383492 on Build GRCh38
rs12082312 Chr.1: 182383577-182383577 on Build GRCh38
rs571173521 Chr.1: 182383268-182383268 on Build GRCh38
rs898457031 Chr.1: 182383481-182383481 on Build GRCh38
rs1041110746 Chr.1: 182383280-182383280 on Build GRCh38
rs376234655 Chr.1: 182383250-182383250 on Build GRCh38
rs146631739 Chr.1: 182383347-182383347 on Build GRCh38
rs538361884 Chr.1: 182383425-182383425 on Build GRCh38
rs550233273 Chr.1: 182383594-182383594 on Build GRCh38
rs1023782538 Chr.1: 182383523-182383523 on Build GRCh38
rs973582858 Chr.1: 182383415-182383415 on Build GRCh38
rs775797329 Chr.1: 182383213-182383213 on Build GRCh38
rs573813239 Chr.1: 182383511-182383511 on Build GRCh38
rs528761679 Chr.1: 182383327-182383327 on Build GRCh38
rs1047693460 Chr.1: 182383234-182383234 on Build GRCh38
rs899859074 Chr.1: 182383505-182383505 on Build GRCh38
rs561577706 Chr.1: 182383294-182383294 on Build GRCh38
rs929135445 Chr.1: 182383478-182383478 on Build GRCh38
rs182932991 Chr.1: 182383549-182383549 on Build GRCh38
rs577169145 Chr.1: 182383556-182383556 on Build GRCh38
rs768055902 Chr.1: 182383458-182383458 on Build GRCh38
rs992452640 Chr.1: 182383373-182383373 on Build GRCh38
rs7734 Chr.1: 182383486-182383486 on Build GRCh38
rs988914355 Chr.1: 182383308-182383311 on Build GRCh38
rs566248330 Chr.1: 182383503-182383503 on Build GRCh38
rs905349532 Chr.1: 182383524-182383524 on Build GRCh38
rs957044989 Chr.1: 182383288-182383288 on Build GRCh38
rs191590401 Chr.1: 182383487-182383487 on Build GRCh38
rs887075015 Chr.1: 182383252-182383252 on Build GRCh38
rs994599606 Chr.1: 182383147-182383147 on Build GRCh38
rs886045609 Chr.1: 182383424-182383424 on Build GRCh38
rs899911178 Chr.1: 182383289-182383289 on Build GRCh38
rs1043547767 Chr.1: 182383545-182383545 on Build GRCh38
rs1021778402 Chr.1: 182383313-182383313 on Build GRCh38
rs534579885 Chr.1: 182383525-182383525 on Build GRCh38
rs1023813661 Chr.1: 182383170-182383170 on Build GRCh38
rs912945206 Chr.1: 182383332-182383332 on Build GRCh38
rs762460063 Chr.1: 182383423-182383423 on Build GRCh38
rs929187878 Chr.1: 182383233-182383233 on Build GRCh38
rs1037534751 Chr.1: 182383231-182383231 on Build GRCh38
rs763220897 Chr.1: 182383253-182383253 on Build GRCh38
rs1014106655 Chr.1: 182383520-182383520 on Build GRCh38
rs186333925 Chr.1: 182383372-182383372 on Build GRCh38
rs915597774 Chr.1: 182383402-182383402 on Build GRCh38
rs777566947 Chr.1: 182383430-182383430 on Build GRCh38
rs1031892649 Chr.1: 182383364-182383364 on Build GRCh38
rs1013722306 Chr.1: 182383167-182383167 on Build GRCh38
rs8527 Chr.1: 182383585-182383585 on Build GRCh38
rs8762 Chr.1: 182383483-182383483 on Build GRCh38
rs969821818 Chr.1: 182383413-182383413 on Build GRCh38
rs996918168 Chr.1: 182383527-182383527 on Build GRCh38
rs920450618 Chr.1: 182383232-182383232 on Build GRCh38
rs961110777 Chr.1: 182383160-182383160 on Build GRCh38
rs774094634 Chr.1: 182383436-182383437 on Build GRCh38
rs7543200 Chr.1: 182383439-182383439 on Build GRCh38
rs181238669 Chr.1: 182383159-182383159 on Build GRCh38
rs549506269 Chr.1: 182383187-182383187 on Build GRCh38
rs1008239774 Chr.1: 182383259-182383259 on Build GRCh38

View Full Product Details