Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 182858907-182859437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX9
Entrez Gene ID
1660
Gene Name
DEAH-box helicase 9
Gene Aliases
DDX9, LKP, NDH2, NDHII, RHA
UniGene
Hs.191518
Gene Chromosome Location
Chr.1: 182839304-182887982 on Build GRCh38
Transcripts
NM_001357.4 NR_033302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1250066 c.1140+1G>A p.? Chr.1: 182859118-182859118 on Build GRCh38
2097215 c.1125T>C p.D375D Chr.1: 182859102-182859102 on Build GRCh38
6569411 c.1099G>T p.E367* Chr.1: 182859076-182859076 on Build GRCh38
7400767 c.1063-3C>T p.? Chr.1: 182859037-182859037 on Build GRCh38
7307962 c.1067C>A p.T356N Chr.1: 182859044-182859044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559199115 Chr.1: 182859176-182859176 on Build GRCh38
rs913176702 Chr.1: 182859392-182859392 on Build GRCh38
rs540390697 Chr.1: 182859258-182859261 on Build GRCh38
rs756019684 Chr.1: 182858990-182858990 on Build GRCh38
rs945892272 Chr.1: 182859098-182859098 on Build GRCh38
rs754538011 Chr.1: 182859145-182859145 on Build GRCh38
rs754846908 Chr.1: 182859136-182859136 on Build GRCh38
rs906657433 Chr.1: 182859103-182859103 on Build GRCh38
rs945862845 Chr.1: 182859403-182859403 on Build GRCh38
rs961081360 Chr.1: 182858939-182858939 on Build GRCh38
rs769975360 Chr.1: 182858940-182858941 on Build GRCh38
rs368787874 Chr.1: 182859048-182859048 on Build GRCh38
rs1006228360 Chr.1: 182859187-182859187 on Build GRCh38
rs772383576 Chr.1: 182859151-182859151 on Build GRCh38
rs556240261 Chr.1: 182859317-182859317 on Build GRCh38
rs750206452 Chr.1: 182858969-182858969 on Build GRCh38
rs371598256 Chr.1: 182859140-182859140 on Build GRCh38
rs895375833 Chr.1: 182859175-182859175 on Build GRCh38
rs557181740 Chr.1: 182859409-182859409 on Build GRCh38
rs752937147 Chr.1: 182859067-182859067 on Build GRCh38
rs935919794 Chr.1: 182859021-182859021 on Build GRCh38
rs1049734302 Chr.1: 182859365-182859365 on Build GRCh38
rs376992047 Chr.1: 182859132-182859132 on Build GRCh38
rs767056538 Chr.1: 182859091-182859091 on Build GRCh38
rs12090342 Chr.1: 182859373-182859373 on Build GRCh38
rs376734640 Chr.1: 182859082-182859082 on Build GRCh38
rs546891147 Chr.1: 182859171-182859171 on Build GRCh38
rs1022598436 Chr.1: 182859158-182859158 on Build GRCh38
rs544552335 Chr.1: 182859217-182859217 on Build GRCh38
rs932736742 Chr.1: 182859339-182859339 on Build GRCh38
rs935905676 Chr.1: 182859354-182859354 on Build GRCh38
rs747184284 Chr.1: 182859163-182859163 on Build GRCh38
rs768462016 Chr.1: 182858933-182858933 on Build GRCh38
rs16859747 Chr.1: 182859204-182859204 on Build GRCh38
rs778837594 Chr.1: 182859137-182859137 on Build GRCh38
rs1054272901 Chr.1: 182859071-182859071 on Build GRCh38
rs878940730 Chr.1: 182859159-182859160 on Build GRCh38
rs777305991 Chr.1: 182859044-182859044 on Build GRCh38
rs922379690 Chr.1: 182858986-182858986 on Build GRCh38
rs373551086 Chr.1: 182859102-182859102 on Build GRCh38
rs772858703 Chr.1: 182859001-182859001 on Build GRCh38
rs779858668 Chr.1: 182859019-182859019 on Build GRCh38
rs972441035 Chr.1: 182858960-182858960 on Build GRCh38
rs373938897 Chr.1: 182859065-182859065 on Build GRCh38
rs772183206 Chr.1: 182858996-182858996 on Build GRCh38
rs773452295 Chr.1: 182859006-182859009 on Build GRCh38
rs777824420 Chr.1: 182859154-182859154 on Build GRCh38
rs955307221 Chr.1: 182859276-182859276 on Build GRCh38
rs985103749 Chr.1: 182859328-182859328 on Build GRCh38
rs747834694 Chr.1: 182859233-182859233 on Build GRCh38
rs771154094 Chr.1: 182859169-182859169 on Build GRCh38
rs910899048 Chr.1: 182859340-182859340 on Build GRCh38
rs747886156 Chr.1: 182859148-182859148 on Build GRCh38
rs1029358697 Chr.1: 182859262-182859262 on Build GRCh38
rs771603690 Chr.1: 182859020-182859020 on Build GRCh38
rs542426463 Chr.1: 182859375-182859375 on Build GRCh38
rs969015473 Chr.1: 182859191-182859191 on Build GRCh38
rs985562949 Chr.1: 182859002-182859002 on Build GRCh38
rs779490663 Chr.1: 182859115-182859115 on Build GRCh38
rs369582240 Chr.1: 182859100-182859100 on Build GRCh38
rs567830092 Chr.1: 182859018-182859018 on Build GRCh38

View Full Product Details