Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 115640268-115640776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLS3
Entrez Gene ID
5358
Gene Name
plastin 3
Gene Aliases
BMND18, T-plastin
UniGene
Hs.496622
Gene Chromosome Location
Chr.X: 115560850-115650861 on Build GRCh38
Transcripts
NM_001136025.4 NM_001172335.2 NM_001282337.1 NM_001282338.1 NM_005032.6 XM_011537534.1

SNP Detail

SNP ID Chromosome Location
rs782708632 Chr.X: 115640502-115640502 on Build GRCh38
rs912266536 Chr.X: 115640350-115640350 on Build GRCh38
rs202076515 Chr.X: 115640408-115640408 on Build GRCh38
rs782546232 Chr.X: 115640542-115640542 on Build GRCh38
rs782089574 Chr.X: 115640523-115640523 on Build GRCh38
rs142569974 Chr.X: 115640490-115640490 on Build GRCh38
rs183898055 Chr.X: 115640534-115640534 on Build GRCh38
rs944940390 Chr.X: 115640434-115640434 on Build GRCh38
rs782208780 Chr.X: 115640726-115640726 on Build GRCh38
rs781988960 Chr.X: 115640444-115640444 on Build GRCh38
rs891879522 Chr.X: 115640703-115640703 on Build GRCh38
rs782279384 Chr.X: 115640381-115640381 on Build GRCh38
rs140968059 Chr.X: 115640441-115640441 on Build GRCh38
rs199599805 Chr.X: 115640390-115640390 on Build GRCh38
rs191317026 Chr.X: 115640720-115640720 on Build GRCh38
rs137917062 Chr.X: 115640474-115640474 on Build GRCh38
rs782779243 Chr.X: 115640585-115640585 on Build GRCh38
rs782599491 Chr.X: 115640736-115640736 on Build GRCh38
rs187910012 Chr.X: 115640671-115640671 on Build GRCh38
rs1010364749 Chr.X: 115640718-115640718 on Build GRCh38
rs151060244 Chr.X: 115640300-115640300 on Build GRCh38
rs782408161 Chr.X: 115640477-115640477 on Build GRCh38
rs1034656962 Chr.X: 115640750-115640750 on Build GRCh38
rs782574315 Chr.X: 115640673-115640673 on Build GRCh38
rs782042501 Chr.X: 115640479-115640479 on Build GRCh38
rs1039720584 Chr.X: 115640438-115640438 on Build GRCh38
rs782362372 Chr.X: 115640443-115640443 on Build GRCh38
rs782586628 Chr.X: 115640419-115640419 on Build GRCh38
rs193086079 Chr.X: 115640342-115640342 on Build GRCh38
rs986421613 Chr.X: 115640327-115640327 on Build GRCh38
rs781865507 Chr.X: 115640540-115640540 on Build GRCh38
rs781788248 Chr.X: 115640685-115640685 on Build GRCh38
rs951142212 Chr.X: 115640301-115640301 on Build GRCh38
rs782649661 Chr.X: 115640374-115640374 on Build GRCh38
rs782804353 Chr.X: 115640333-115640333 on Build GRCh38
rs781820311 Chr.X: 115640516-115640516 on Build GRCh38
rs781900145 Chr.X: 115640634-115640634 on Build GRCh38

View Full Product Details