Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 117898119-117898629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL13
Entrez Gene ID
90293
Gene Name
kelch like family member 13
Gene Aliases
BKLHD2
UniGene
Hs.348262
Gene Chromosome Location
Chr.X: 117897813-118117340 on Build GRCh38
Transcripts
NM_001168299.1 NM_001168300.1 NM_001168301.1 NM_001168302.1 NM_001168303.1 NM_033495.3 XM_011531409.2 XM_011531410.2 XM_011531411.1 XM_017029950.1

SNP Detail

SNP ID Chromosome Location
rs765508381 Chr.X: 117898187-117898187 on Build GRCh38
rs201342313 Chr.X: 117898186-117898187 on Build GRCh38
rs6645413 Chr.X: 117898214-117898214 on Build GRCh38
rs947633791 Chr.X: 117898465-117898465 on Build GRCh38
rs906975052 Chr.X: 117898275-117898275 on Build GRCh38
rs139164455 Chr.X: 117898327-117898327 on Build GRCh38
rs908888203 Chr.X: 117898181-117898182 on Build GRCh38
rs905990228 Chr.X: 117898560-117898560 on Build GRCh38
rs762785116 Chr.X: 117898397-117898397 on Build GRCh38
rs371718391 Chr.X: 117898428-117898428 on Build GRCh38
rs1001625616 Chr.X: 117898561-117898561 on Build GRCh38
rs950653064 Chr.X: 117898179-117898179 on Build GRCh38
rs751478895 Chr.X: 117898526-117898526 on Build GRCh38
rs940338657 Chr.X: 117898205-117898205 on Build GRCh38
rs1035807781 Chr.X: 117898583-117898583 on Build GRCh38
rs923016815 Chr.X: 117898362-117898362 on Build GRCh38
rs931563127 Chr.X: 117898175-117898178 on Build GRCh38
rs779586573 Chr.X: 117898186-117898186 on Build GRCh38
rs903087227 Chr.X: 117898533-117898533 on Build GRCh38
rs1052767798 Chr.X: 117898410-117898410 on Build GRCh38
rs1036016425 Chr.X: 117898267-117898267 on Build GRCh38
rs140487028 Chr.X: 117898458-117898458 on Build GRCh38
rs938405018 Chr.X: 117898330-117898330 on Build GRCh38
rs747491528 Chr.X: 117898396-117898396 on Build GRCh38

View Full Product Details