Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120430737-120431272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMP2
Entrez Gene ID
3920
Gene Name
lysosomal associated membrane protein 2
Gene Aliases
CD107b, LAMP-2, LAMPB, LGP110
UniGene
Hs.496684
Gene Chromosome Location
Chr.X: 120426148-120469349 on Build GRCh38
Transcripts
NM_001122606.1 NM_002294.2

SNP Detail

SNP ID Chromosome Location
rs916829066 Chr.X: 120430766-120430766 on Build GRCh38
rs542576305 Chr.X: 120431171-120431171 on Build GRCh38
rs891114974 Chr.X: 120430768-120430768 on Build GRCh38
rs758434357 Chr.X: 120430973-120430973 on Build GRCh38
rs908795462 Chr.X: 120431087-120431087 on Build GRCh38
rs1028350037 Chr.X: 120430763-120430763 on Build GRCh38
rs1801135 Chr.X: 120431027-120431027 on Build GRCh38
rs386827301 Chr.X: 120430837-120430838 on Build GRCh38
rs750939378 Chr.X: 120430846-120430846 on Build GRCh38
rs917617678 Chr.X: 120431056-120431056 on Build GRCh38
rs756938982 Chr.X: 120430773-120430773 on Build GRCh38
rs1050285762 Chr.X: 120431227-120431227 on Build GRCh38
rs755520529 Chr.X: 120431009-120431009 on Build GRCh38
rs757173953 Chr.X: 120431200-120431203 on Build GRCh38
rs778938403 Chr.X: 120431211-120431211 on Build GRCh38
rs753133641 Chr.X: 120431063-120431063 on Build GRCh38
rs1008155913 Chr.X: 120430828-120430828 on Build GRCh38
rs185972753 Chr.X: 120430838-120430838 on Build GRCh38
rs766396944 Chr.X: 120431003-120431008 on Build GRCh38
rs749885888 Chr.X: 120430893-120430893 on Build GRCh38
rs1032582981 Chr.X: 120430923-120430923 on Build GRCh38
rs143070918 Chr.X: 120431086-120431086 on Build GRCh38
rs768475790 Chr.X: 120431248-120431248 on Build GRCh38
rs959191331 Chr.X: 120431013-120431013 on Build GRCh38
rs8160 Chr.X: 120431026-120431026 on Build GRCh38
rs749090092 Chr.X: 120431199-120431199 on Build GRCh38
rs41300191 Chr.X: 120431118-120431118 on Build GRCh38
rs371031457 Chr.X: 120431066-120431066 on Build GRCh38
rs5957383 Chr.X: 120430837-120430837 on Build GRCh38
rs898643725 Chr.X: 120431173-120431173 on Build GRCh38
rs1038575879 Chr.X: 120431168-120431168 on Build GRCh38

View Full Product Details