Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202600267-202600775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT2
Entrez Gene ID
127833
Gene Name
synaptotagmin 2
Gene Aliases
CMS7, MYSPC, SytII
UniGene
Hs.25422
Gene Chromosome Location
Chr.1: 202590596-202710444 on Build GRCh38
Transcripts
NM_001136504.1 NM_177402.4 XM_011509192.2 XM_017000309.1 XM_017000310.1 XM_017000311.1 XM_017000312.1 XM_017000313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6009731 c.906G>A p.V302V Chr.1: 202600370-202600370 on Build GRCh38
4027086 c.835C>T p.R279C Chr.1: 202600441-202600441 on Build GRCh38
4608251 c.850G>T p.A284S Chr.1: 202600426-202600426 on Build GRCh38
164763 c.806A>G p.E269G Chr.1: 202600470-202600470 on Build GRCh38
5616693 c.912C>A p.G304G Chr.1: 202600364-202600364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779602046 Chr.1: 202600347-202600347 on Build GRCh38
rs1035406478 Chr.1: 202600587-202600587 on Build GRCh38
rs953745027 Chr.1: 202600690-202600691 on Build GRCh38
rs115267501 Chr.1: 202600472-202600472 on Build GRCh38
rs565352841 Chr.1: 202600652-202600652 on Build GRCh38
rs907699 Chr.1: 202600427-202600427 on Build GRCh38
rs752537599 Chr.1: 202600352-202600352 on Build GRCh38
rs368659613 Chr.1: 202600506-202600506 on Build GRCh38
rs904322687 Chr.1: 202600407-202600407 on Build GRCh38
rs968166238 Chr.1: 202600671-202600671 on Build GRCh38
rs12047787 Chr.1: 202600703-202600703 on Build GRCh38
rs775229774 Chr.1: 202600491-202600491 on Build GRCh38
rs547612799 Chr.1: 202600728-202600728 on Build GRCh38
rs545437677 Chr.1: 202600603-202600603 on Build GRCh38
rs761719749 Chr.1: 202600515-202600515 on Build GRCh38
rs776277945 Chr.1: 202600331-202600331 on Build GRCh38
rs945738997 Chr.1: 202600705-202600705 on Build GRCh38
rs577574499 Chr.1: 202600748-202600748 on Build GRCh38
rs531725391 Chr.1: 202600570-202600570 on Build GRCh38
rs1014199356 Chr.1: 202600572-202600572 on Build GRCh38
rs769477916 Chr.1: 202600481-202600481 on Build GRCh38
rs201527461 Chr.1: 202600317-202600317 on Build GRCh38
rs932292649 Chr.1: 202600303-202600303 on Build GRCh38
rs199542449 Chr.1: 202600428-202600428 on Build GRCh38
rs977118348 Chr.1: 202600723-202600723 on Build GRCh38
rs905377408 Chr.1: 202600739-202600739 on Build GRCh38
rs887519724 Chr.1: 202600710-202600710 on Build GRCh38
rs1467120 Chr.1: 202600749-202600749 on Build GRCh38
rs910394788 Chr.1: 202600685-202600685 on Build GRCh38
rs368709290 Chr.1: 202600321-202600321 on Build GRCh38
rs376917986 Chr.1: 202600509-202600509 on Build GRCh38
rs780359132 Chr.1: 202600373-202600373 on Build GRCh38
rs563203327 Chr.1: 202600551-202600551 on Build GRCh38
rs1005672399 Chr.1: 202600519-202600519 on Build GRCh38
rs923219832 Chr.1: 202600693-202600693 on Build GRCh38
rs963466455 Chr.1: 202600648-202600648 on Build GRCh38
rs756234494 Chr.1: 202600372-202600372 on Build GRCh38
rs1040310459 Chr.1: 202600689-202600689 on Build GRCh38
rs1002837863 Chr.1: 202600747-202600747 on Build GRCh38
rs750425155 Chr.1: 202600367-202600367 on Build GRCh38
rs988171465 Chr.1: 202600616-202600616 on Build GRCh38
rs71635588 Chr.1: 202600364-202600364 on Build GRCh38
rs979668127 Chr.1: 202600682-202600682 on Build GRCh38
rs1038903345 Chr.1: 202600730-202600730 on Build GRCh38
rs913971232 Chr.1: 202600634-202600634 on Build GRCh38
rs763981285 Chr.1: 202600657-202600657 on Build GRCh38
rs372628488 Chr.1: 202600400-202600400 on Build GRCh38
rs906551659 Chr.1: 202600599-202600599 on Build GRCh38
rs755306709 Chr.1: 202600413-202600413 on Build GRCh38
rs943242352 Chr.1: 202600688-202600688 on Build GRCh38
rs943335288 Chr.1: 202600424-202600424 on Build GRCh38
rs370255354 Chr.1: 202600316-202600316 on Build GRCh38
rs374302028 Chr.1: 202600430-202600430 on Build GRCh38
rs946521673 Chr.1: 202600338-202600338 on Build GRCh38
rs566653846 Chr.1: 202600729-202600729 on Build GRCh38
rs765896219 Chr.1: 202600326-202600326 on Build GRCh38
rs200553744 Chr.1: 202600353-202600353 on Build GRCh38
rs530739760 Chr.1: 202600602-202600602 on Build GRCh38
rs778365654 Chr.1: 202600451-202600451 on Build GRCh38
rs759167957 Chr.1: 202600477-202600477 on Build GRCh38
rs746445612 Chr.1: 202600460-202600460 on Build GRCh38
rs765004983 Chr.1: 202600350-202600350 on Build GRCh38
rs1002460363 Chr.1: 202600556-202600556 on Build GRCh38
rs776179209 Chr.1: 202600473-202600473 on Build GRCh38
rs1017135532 Chr.1: 202600527-202600527 on Build GRCh38
rs751093350 Chr.1: 202600702-202600702 on Build GRCh38
rs543303662 Chr.1: 202600496-202600496 on Build GRCh38
rs973541302 Chr.1: 202600653-202600653 on Build GRCh38

View Full Product Details