Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202896613-202897120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL12
Entrez Gene ID
59349
Gene Name
kelch like family member 12
Gene Aliases
C3IP1, DKIR
UniGene
Hs.706793
Gene Chromosome Location
Chr.1: 202891096-202928644 on Build GRCh38
Transcripts
NM_001303051.1 NM_001303109.1 NM_021633.3 XM_011509835.1 XM_011509836.1 XM_011509837.1 XM_017001995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3385630 c.892G>A p.V298M Chr.1: 202896901-202896901 on Build GRCh38
4421682 c.872G>T p.S291I Chr.1: 202896921-202896921 on Build GRCh38
4421683 c.986G>T p.S329I Chr.1: 202896921-202896921 on Build GRCh38
4414716 c.873C>T p.S291S Chr.1: 202896920-202896920 on Build GRCh38
4414717 c.987C>T p.S329S Chr.1: 202896920-202896920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200356503 Chr.1: 202896997-202896997 on Build GRCh38
rs778779595 Chr.1: 202896867-202896867 on Build GRCh38
rs950461494 Chr.1: 202896882-202896882 on Build GRCh38
rs959930768 Chr.1: 202897092-202897092 on Build GRCh38
rs199552913 Chr.1: 202897000-202897000 on Build GRCh38
rs746023146 Chr.1: 202896935-202896935 on Build GRCh38
rs1009717549 Chr.1: 202896706-202896706 on Build GRCh38
rs114132831 Chr.1: 202897011-202897011 on Build GRCh38
rs764679282 Chr.1: 202896803-202896803 on Build GRCh38
rs1001436984 Chr.1: 202896781-202896781 on Build GRCh38
rs191227918 Chr.1: 202896831-202896831 on Build GRCh38
rs759257866 Chr.1: 202897012-202897012 on Build GRCh38
rs1043164740 Chr.1: 202896647-202896647 on Build GRCh38
rs769177103 Chr.1: 202896964-202896964 on Build GRCh38
rs765898152 Chr.1: 202897006-202897006 on Build GRCh38
rs748080354 Chr.1: 202896869-202896869 on Build GRCh38
rs980335833 Chr.1: 202897075-202897075 on Build GRCh38
rs768192806 Chr.1: 202896972-202896972 on Build GRCh38
rs182910818 Chr.1: 202897024-202897024 on Build GRCh38
rs775023096 Chr.1: 202896808-202896808 on Build GRCh38
rs761368712 Chr.1: 202896809-202896809 on Build GRCh38
rs1002411805 Chr.1: 202896791-202896791 on Build GRCh38
rs777162279 Chr.1: 202896996-202896996 on Build GRCh38
rs900740055 Chr.1: 202896934-202896934 on Build GRCh38
rs781115585 Chr.1: 202896905-202896905 on Build GRCh38
rs760281955 Chr.1: 202897001-202897001 on Build GRCh38
rs370985224 Chr.1: 202897072-202897072 on Build GRCh38
rs116804728 Chr.1: 202896766-202896766 on Build GRCh38
rs201540872 Chr.1: 202896988-202896988 on Build GRCh38
rs548897081 Chr.1: 202896663-202896663 on Build GRCh38
rs556229646 Chr.1: 202896890-202896890 on Build GRCh38
rs923212124 Chr.1: 202897085-202897085 on Build GRCh38
rs1022430293 Chr.1: 202896719-202896719 on Build GRCh38
rs534589877 Chr.1: 202896790-202896790 on Build GRCh38
rs201795698 Chr.1: 202896932-202896932 on Build GRCh38
rs200154578 Chr.1: 202896832-202896832 on Build GRCh38
rs775908332 Chr.1: 202896949-202896949 on Build GRCh38
rs757301002 Chr.1: 202896884-202896884 on Build GRCh38
rs552777463 Chr.1: 202896876-202896876 on Build GRCh38
rs769829965 Chr.1: 202897086-202897086 on Build GRCh38
rs111452276 Chr.1: 202896768-202896768 on Build GRCh38
rs1016724511 Chr.1: 202897026-202897026 on Build GRCh38
rs766094982 Chr.1: 202896837-202896837 on Build GRCh38
rs769880715 Chr.1: 202896944-202896944 on Build GRCh38
rs754787664 Chr.1: 202896859-202896859 on Build GRCh38
rs904005450 Chr.1: 202896780-202896780 on Build GRCh38
rs960685530 Chr.1: 202896821-202896821 on Build GRCh38
rs904022852 Chr.1: 202896722-202896722 on Build GRCh38
rs753607460 Chr.1: 202896843-202896843 on Build GRCh38
rs1026512575 Chr.1: 202896875-202896875 on Build GRCh38

View Full Product Details