Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203308587-203309083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG2
Entrez Gene ID
7832
Gene Name
BTG anti-proliferation factor 2
Gene Aliases
APRO1, PC3, TIS21
UniGene
Hs.519162
Gene Chromosome Location
Chr.1: 203305536-203309602 on Build GRCh38
Transcripts
NM_006763.2

SNP Detail

SNP ID Chromosome Location
rs534900693 Chr.1: 203308745-203308745 on Build GRCh38
rs935433642 Chr.1: 203308840-203308840 on Build GRCh38
rs1046540553 Chr.1: 203308800-203308800 on Build GRCh38
rs935417653 Chr.1: 203308841-203308841 on Build GRCh38
rs764567961 Chr.1: 203308990-203308990 on Build GRCh38
rs780900419 Chr.1: 203308836-203308836 on Build GRCh38
rs961818211 Chr.1: 203309046-203309046 on Build GRCh38
rs924422721 Chr.1: 203308836-203308836 on Build GRCh38
rs973096507 Chr.1: 203308669-203308669 on Build GRCh38
rs575103998 Chr.1: 203308908-203308908 on Build GRCh38
rs16851280 Chr.1: 203308615-203308615 on Build GRCh38
rs924082329 Chr.1: 203308817-203308817 on Build GRCh38
rs764873286 Chr.1: 203308725-203308725 on Build GRCh38
rs989693507 Chr.1: 203308688-203308688 on Build GRCh38
rs11586635 Chr.1: 203308649-203308649 on Build GRCh38
rs916830474 Chr.1: 203308751-203308751 on Build GRCh38
rs965952000 Chr.1: 203309057-203309057 on Build GRCh38
rs1014771219 Chr.1: 203309035-203309035 on Build GRCh38
rs1034798235 Chr.1: 203308629-203308629 on Build GRCh38
rs373141560 Chr.1: 203308774-203308774 on Build GRCh38
rs367892990 Chr.1: 203308916-203308916 on Build GRCh38
rs879162712 Chr.1: 203308677-203308677 on Build GRCh38
rs1045367507 Chr.1: 203308906-203308906 on Build GRCh38
rs557364252 Chr.1: 203308980-203308980 on Build GRCh38
rs572194545 Chr.1: 203309023-203309023 on Build GRCh38
rs538887336 Chr.1: 203308769-203308769 on Build GRCh38
rs949610709 Chr.1: 203308772-203308772 on Build GRCh38
rs893841088 Chr.1: 203308869-203308869 on Build GRCh38
rs565394831 Chr.1: 203308696-203308696 on Build GRCh38
rs961837311 Chr.1: 203308662-203308662 on Build GRCh38
rs994546782 Chr.1: 203309049-203309049 on Build GRCh38
rs905432979 Chr.1: 203308924-203308924 on Build GRCh38
rs751564630 Chr.1: 203309018-203309018 on Build GRCh38
rs537329168 Chr.1: 203308946-203308946 on Build GRCh38
rs920279956 Chr.1: 203308670-203308670 on Build GRCh38
rs763532512 Chr.1: 203308931-203308931 on Build GRCh38
rs1009172053 Chr.1: 203309025-203309025 on Build GRCh38
rs1011322751 Chr.1: 203308899-203308899 on Build GRCh38
rs374313138 Chr.1: 203308646-203308646 on Build GRCh38
rs78959463 Chr.1: 203309042-203309042 on Build GRCh38
rs577164404 Chr.1: 203308992-203308992 on Build GRCh38
rs150236063 Chr.1: 203308930-203308930 on Build GRCh38
rs60342861 Chr.1: 203308747-203308747 on Build GRCh38
rs910451465 Chr.1: 203308700-203308700 on Build GRCh38
rs993294068 Chr.1: 203308958-203308958 on Build GRCh38
rs745825418 Chr.1: 203308953-203308953 on Build GRCh38
rs1035206494 Chr.1: 203308952-203308953 on Build GRCh38
rs751952792 Chr.1: 203309040-203309040 on Build GRCh38

View Full Product Details