Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206588881-206589418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2D
Entrez Gene ID
1939
Gene Name
eukaryotic translation initiation factor 2D
Gene Aliases
HCA56, LGTN
UniGene
Hs.497581
Gene Chromosome Location
Chr.1: 206572485-206612551 on Build GRCh38
Transcripts
XM_011509257.2 NM_182663.3 NM_182664.3 NM_182665.3

Gene Symbol
RASSF5
Entrez Gene ID
83593
Gene Name
Ras association domain family member 5
Gene Aliases
Maxp1, NORE1, NORE1A, NORE1B, RAPL, RASSF3
UniGene
Hs.497579
Gene Chromosome Location
Chr.1: 206507530-206589284 on Build GRCh38
Transcripts
XM_011509257.2 NM_182663.3 NM_182664.3 NM_182665.3

SNP Detail

SNP ID Chromosome Location
rs543389600 Chr.1: 206588939-206588939 on Build GRCh38
rs973825517 Chr.1: 206588984-206588984 on Build GRCh38
rs77214751 Chr.1: 206589328-206589328 on Build GRCh38
rs950631286 Chr.1: 206589290-206589290 on Build GRCh38
rs11539427 Chr.1: 206588950-206588950 on Build GRCh38
rs1051515255 Chr.1: 206589012-206589012 on Build GRCh38
rs1025144012 Chr.1: 206589153-206589153 on Build GRCh38
rs545447203 Chr.1: 206588993-206588993 on Build GRCh38
rs943122191 Chr.1: 206589025-206589025 on Build GRCh38
rs904282214 Chr.1: 206589063-206589063 on Build GRCh38
rs184742000 Chr.1: 206589233-206589233 on Build GRCh38
rs942071474 Chr.1: 206588965-206588965 on Build GRCh38
rs1053199580 Chr.1: 206589109-206589109 on Build GRCh38
rs919269878 Chr.1: 206589283-206589283 on Build GRCh38
rs1142795 Chr.1: 206588949-206588949 on Build GRCh38
rs1009855874 Chr.1: 206589133-206589133 on Build GRCh38
rs183041409 Chr.1: 206588924-206588924 on Build GRCh38
rs556372314 Chr.1: 206589329-206589329 on Build GRCh38
rs554324155 Chr.1: 206589315-206589315 on Build GRCh38
rs782437185 Chr.1: 206588969-206588969 on Build GRCh38
rs530452467 Chr.1: 206589303-206589303 on Build GRCh38
rs11119102 Chr.1: 206589352-206589352 on Build GRCh38
rs192770805 Chr.1: 206589086-206589086 on Build GRCh38
rs11539428 Chr.1: 206589123-206589123 on Build GRCh38
rs1039295697 Chr.1: 206589341-206589341 on Build GRCh38
rs188566971 Chr.1: 206589023-206589023 on Build GRCh38
rs1002686763 Chr.1: 206589170-206589170 on Build GRCh38
rs574114990 Chr.1: 206589316-206589316 on Build GRCh38
rs970908135 Chr.1: 206589164-206589164 on Build GRCh38
rs563181716 Chr.1: 206589011-206589011 on Build GRCh38
rs553958087 Chr.1: 206589088-206589088 on Build GRCh38
rs115802123 Chr.1: 206589343-206589343 on Build GRCh38
rs1033740957 Chr.1: 206589182-206589182 on Build GRCh38
rs782804293 Chr.1: 206588983-206588983 on Build GRCh38
rs367583228 Chr.1: 206589066-206589066 on Build GRCh38
rs919159458 Chr.1: 206588990-206588990 on Build GRCh38
rs187667048 Chr.1: 206589276-206589276 on Build GRCh38
rs146697160 Chr.1: 206589335-206589335 on Build GRCh38
rs77534575 Chr.1: 206588948-206588948 on Build GRCh38
rs569347839 Chr.1: 206589082-206589082 on Build GRCh38
rs1038808933 Chr.1: 206589026-206589026 on Build GRCh38
rs571671993 Chr.1: 206589242-206589242 on Build GRCh38
rs782084488 Chr.1: 206589261-206589261 on Build GRCh38

View Full Product Details