Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212796185-212796721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSL1
Entrez Gene ID
25936
Gene Name
NSL1, MIS12 kinetochore complex component
Gene Aliases
C1orf48, DC8, MIS14
UniGene
Hs.497692
Gene Chromosome Location
Chr.1: 212726153-212798900 on Build GRCh38
Transcripts
XM_017000968.1 XM_017000969.1 NM_001042552.2 NM_001042553.2 NM_001146169.1 NM_001146170.1 NM_001146171.1 XM_011509202.2 XM_017000328.1 XM_017000329.1 XM_017000330.1 XM_017000331.1

Gene Symbol
TATDN3
Entrez Gene ID
128387
Gene Name
TatD DNase domain containing 3
Gene Aliases
-
UniGene
Hs.530538
Gene Chromosome Location
Chr.1: 212791828-212816825 on Build GRCh38
Transcripts
XM_017000968.1 XM_017000969.1 NM_001042552.2 NM_001042553.2 NM_001146169.1 NM_001146170.1 NM_001146171.1 XM_011509202.2 XM_017000328.1 XM_017000329.1 XM_017000330.1 XM_017000331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3483176 c.113C>T p.A38V Chr.1: 212796530-212796530 on Build GRCh38
3785146 c.173+2T>G p.? Chr.1: 212796592-212796592 on Build GRCh38
678698 c.123A>C p.A41A Chr.1: 212796540-212796540 on Build GRCh38
903611 c.102C>A p.A34A Chr.1: 212796519-212796519 on Build GRCh38
4633556 c.173G>T p.R58M Chr.1: 212796590-212796590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779826722 Chr.1: 212796551-212796551 on Build GRCh38
rs760318925 Chr.1: 212796501-212796501 on Build GRCh38
rs201088735 Chr.1: 212796521-212796521 on Build GRCh38
rs771482771 Chr.1: 212796575-212796575 on Build GRCh38
rs949720068 Chr.1: 212796360-212796360 on Build GRCh38
rs1004383642 Chr.1: 212796469-212796469 on Build GRCh38
rs1045376993 Chr.1: 212796396-212796396 on Build GRCh38
rs141517277 Chr.1: 212796265-212796265 on Build GRCh38
rs141185416 Chr.1: 212796540-212796541 on Build GRCh38
rs190012466 Chr.1: 212796233-212796233 on Build GRCh38
rs901642505 Chr.1: 212796389-212796389 on Build GRCh38
rs773572343 Chr.1: 212796476-212796476 on Build GRCh38
rs777062648 Chr.1: 212796586-212796586 on Build GRCh38
rs867068125 Chr.1: 212796539-212796539 on Build GRCh38
rs765432299 Chr.1: 212796496-212796496 on Build GRCh38
rs564552830 Chr.1: 212796223-212796223 on Build GRCh38
rs115150941 Chr.1: 212796564-212796564 on Build GRCh38
rs11421046 Chr.1: 212796501-212796502 on Build GRCh38
rs1040230182 Chr.1: 212796368-212796368 on Build GRCh38
rs980728603 Chr.1: 212796231-212796231 on Build GRCh38
rs11120029 Chr.1: 212796238-212796238 on Build GRCh38
rs1054234920 Chr.1: 212796291-212796291 on Build GRCh38
rs373682891 Chr.1: 212796614-212796614 on Build GRCh38
rs762713698 Chr.1: 212796525-212796525 on Build GRCh38
rs761269571 Chr.1: 212796478-212796478 on Build GRCh38
rs766876496 Chr.1: 212796546-212796546 on Build GRCh38
rs769909996 Chr.1: 212796601-212796601 on Build GRCh38
rs776778828 Chr.1: 212796487-212796487 on Build GRCh38
rs568073763 Chr.1: 212796256-212796256 on Build GRCh38
rs913026205 Chr.1: 212796645-212796645 on Build GRCh38
rs996120609 Chr.1: 212796447-212796447 on Build GRCh38
rs1050746513 Chr.1: 212796448-212796448 on Build GRCh38
rs980400946 Chr.1: 212796559-212796559 on Build GRCh38
rs763898688 Chr.1: 212796500-212796501 on Build GRCh38
rs759697077 Chr.1: 212796489-212796489 on Build GRCh38
rs6684313 Chr.1: 212796654-212796654 on Build GRCh38
rs889816505 Chr.1: 212796449-212796450 on Build GRCh38
rs961437287 Chr.1: 212796504-212796504 on Build GRCh38
rs755756183 Chr.1: 212796550-212796550 on Build GRCh38
rs765749949 Chr.1: 212796501-212796503 on Build GRCh38
rs917536770 Chr.1: 212796212-212796212 on Build GRCh38
rs994172180 Chr.1: 212796509-212796510 on Build GRCh38
rs565819581 Chr.1: 212796337-212796337 on Build GRCh38
rs763726725 Chr.1: 212796626-212796626 on Build GRCh38
rs886828542 Chr.1: 212796411-212796411 on Build GRCh38
rs745916371 Chr.1: 212796597-212796597 on Build GRCh38
rs774267239 Chr.1: 212796257-212796257 on Build GRCh38
rs776003590 Chr.1: 212796453-212796453 on Build GRCh38
rs923116472 Chr.1: 212796657-212796657 on Build GRCh38
rs370651128 Chr.1: 212796553-212796553 on Build GRCh38
rs796869022 Chr.1: 212796501-212796502 on Build GRCh38
rs547426310 Chr.1: 212796319-212796319 on Build GRCh38
rs763192657 Chr.1: 212796624-212796624 on Build GRCh38
rs770877357 Chr.1: 212796484-212796484 on Build GRCh38
rs397785164 Chr.1: 212796513-212796514 on Build GRCh38
rs763589291 Chr.1: 212796530-212796530 on Build GRCh38
rs987507693 Chr.1: 212796222-212796222 on Build GRCh38
rs747410442 Chr.1: 212796570-212796570 on Build GRCh38
rs537309844 Chr.1: 212796558-212796558 on Build GRCh38
rs745917599 Chr.1: 212796333-212796333 on Build GRCh38
rs774207387 Chr.1: 212796628-212796628 on Build GRCh38
rs55709122 Chr.1: 212796547-212796547 on Build GRCh38
rs557521772 Chr.1: 212796349-212796349 on Build GRCh38
rs376271458 Chr.1: 212796529-212796529 on Build GRCh38
rs946128825 Chr.1: 212796246-212796246 on Build GRCh38
rs757221228 Chr.1: 212796542-212796542 on Build GRCh38
rs751411204 Chr.1: 212796531-212796531 on Build GRCh38
rs533729772 Chr.1: 212796502-212796503 on Build GRCh38
rs538484576 Chr.1: 212796274-212796274 on Build GRCh38
rs866277522 Chr.1: 212796272-212796272 on Build GRCh38
rs1055627329 Chr.1: 212796309-212796309 on Build GRCh38
rs761566527 Chr.1: 212796641-212796641 on Build GRCh38
rs181229957 Chr.1: 212796307-212796307 on Build GRCh38
rs989807148 Chr.1: 212796617-212796617 on Build GRCh38

View Full Product Details