Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212802523-212803053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TATDN3
Entrez Gene ID
128387
Gene Name
TatD DNase domain containing 3
Gene Aliases
-
UniGene
Hs.530538
Gene Chromosome Location
Chr.1: 212791828-212816825 on Build GRCh38
Transcripts
NM_001042552.2 NM_001042553.2 NM_001146169.1 NM_001146170.1 NM_001146171.1 XM_011509202.2 XM_017000328.1 XM_017000329.1 XM_017000330.1 XM_017000331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5775381 c.308T>C p.L103S Chr.1: 212802750-212802750 on Build GRCh38
6244585 c.276G>T p.L92F Chr.1: 212802718-212802718 on Build GRCh38
6245370 c.279C>A p.P93P Chr.1: 212802721-212802721 on Build GRCh38
6269962 c.271G>T p.A91S Chr.1: 212802713-212802713 on Build GRCh38
6061380 c.298G>A p.D100N Chr.1: 212802740-212802740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774875403 Chr.1: 212802794-212802794 on Build GRCh38
rs1014953372 Chr.1: 212802912-212802912 on Build GRCh38
rs759056037 Chr.1: 212802661-212802661 on Build GRCh38
rs372819741 Chr.1: 212802689-212802689 on Build GRCh38
rs746798032 Chr.1: 212802745-212802745 on Build GRCh38
rs550259408 Chr.1: 212802901-212802901 on Build GRCh38
rs369697680 Chr.1: 212802739-212802739 on Build GRCh38
rs998246182 Chr.1: 212802869-212802869 on Build GRCh38
rs949852413 Chr.1: 212802631-212802631 on Build GRCh38
rs756533878 Chr.1: 212802708-212802708 on Build GRCh38
rs1005671056 Chr.1: 212802902-212802902 on Build GRCh38
rs367546111 Chr.1: 212802685-212802685 on Build GRCh38
rs372314001 Chr.1: 212802786-212802786 on Build GRCh38
rs763727852 Chr.1: 212802809-212802809 on Build GRCh38
rs1025609580 Chr.1: 212802831-212802831 on Build GRCh38
rs762504134 Chr.1: 212802802-212802802 on Build GRCh38
rs750769818 Chr.1: 212802707-212802707 on Build GRCh38
rs778937211 Chr.1: 212802585-212802585 on Build GRCh38
rs770978650 Chr.1: 212802650-212802650 on Build GRCh38
rs970976720 Chr.1: 212802871-212802871 on Build GRCh38
rs778894572 Chr.1: 212802732-212802732 on Build GRCh38
rs76001450 Chr.1: 212802564-212802564 on Build GRCh38
rs776173385 Chr.1: 212802658-212802658 on Build GRCh38
rs755039378 Chr.1: 212802714-212802714 on Build GRCh38
rs372764628 Chr.1: 212802900-212802900 on Build GRCh38
rs140267863 Chr.1: 212802743-212802743 on Build GRCh38
rs754354697 Chr.1: 212802713-212802713 on Build GRCh38
rs762516786 Chr.1: 212802692-212802692 on Build GRCh38
rs916887643 Chr.1: 212802607-212802607 on Build GRCh38
rs1024409572 Chr.1: 212803001-212803001 on Build GRCh38
rs745822787 Chr.1: 212802782-212802782 on Build GRCh38
rs570779083 Chr.1: 212803012-212803012 on Build GRCh38
rs558792186 Chr.1: 212802963-212802963 on Build GRCh38
rs904257976 Chr.1: 212802687-212802687 on Build GRCh38
rs973211858 Chr.1: 212802552-212802552 on Build GRCh38
rs927135409 Chr.1: 212802664-212802666 on Build GRCh38
rs776581874 Chr.1: 212802770-212802770 on Build GRCh38
rs770890786 Chr.1: 212802753-212802753 on Build GRCh38
rs752352931 Chr.1: 212802686-212802686 on Build GRCh38
rs778487739 Chr.1: 212802612-212802612 on Build GRCh38
rs377183459 Chr.1: 212802699-212802699 on Build GRCh38
rs1041376108 Chr.1: 212802664-212802664 on Build GRCh38
rs778200176 Chr.1: 212802744-212802744 on Build GRCh38
rs994401156 Chr.1: 212802974-212802974 on Build GRCh38
rs931865079 Chr.1: 212802570-212802570 on Build GRCh38
rs901147310 Chr.1: 212802790-212802790 on Build GRCh38
rs911813783 Chr.1: 212802645-212802645 on Build GRCh38
rs1017061920 Chr.1: 212802914-212802914 on Build GRCh38
rs1012457401 Chr.1: 212802778-212802778 on Build GRCh38
rs111955952 Chr.1: 212802863-212802863 on Build GRCh38
rs767811172 Chr.1: 212802703-212802703 on Build GRCh38
rs780505374 Chr.1: 212802710-212802710 on Build GRCh38
rs1028349483 Chr.1: 212802894-212802894 on Build GRCh38
rs971032504 Chr.1: 212803009-212803009 on Build GRCh38
rs961938015 Chr.1: 212802946-212802946 on Build GRCh38
rs979823913 Chr.1: 212802634-212802634 on Build GRCh38

View Full Product Details