Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236215899-236216435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERO1B
Entrez Gene ID
56605
Gene Name
endoplasmic reticulum oxidoreductase 1 beta
Gene Aliases
ERO1LB, Ero1beta
UniGene
Hs.558519
Gene Chromosome Location
Chr.1: 236215122-236282039 on Build GRCh38
Transcripts
NM_019891.3

SNP Detail

SNP ID Chromosome Location
rs150012347 Chr.1: 236215966-236215970 on Build GRCh38
rs950652537 Chr.1: 236216304-236216308 on Build GRCh38
rs748264997 Chr.1: 236216327-236216327 on Build GRCh38
rs750244223 Chr.1: 236215944-236215944 on Build GRCh38
rs1054622715 Chr.1: 236216130-236216130 on Build GRCh38
rs1027566785 Chr.1: 236216297-236216297 on Build GRCh38
rs143051410 Chr.1: 236216365-236216365 on Build GRCh38
rs961010819 Chr.1: 236216072-236216072 on Build GRCh38
rs967285047 Chr.1: 236216298-236216298 on Build GRCh38
rs575651583 Chr.1: 236216006-236216006 on Build GRCh38
rs754037155 Chr.1: 236216347-236216347 on Build GRCh38
rs918486505 Chr.1: 236216344-236216344 on Build GRCh38
rs1051306168 Chr.1: 236216203-236216203 on Build GRCh38
rs766480112 Chr.1: 236216132-236216132 on Build GRCh38
rs71698326 Chr.1: 236215970-236215974 on Build GRCh38
rs916575687 Chr.1: 236216331-236216331 on Build GRCh38
rs778256984 Chr.1: 236216133-236216133 on Build GRCh38
rs964915446 Chr.1: 236215961-236215961 on Build GRCh38
rs559702948 Chr.1: 236216196-236216196 on Build GRCh38
rs760769782 Chr.1: 236216064-236216064 on Build GRCh38
rs890165914 Chr.1: 236216153-236216158 on Build GRCh38
rs144963757 Chr.1: 236216317-236216317 on Build GRCh38
rs3208069 Chr.1: 236215925-236215925 on Build GRCh38
rs973988563 Chr.1: 236216338-236216338 on Build GRCh38
rs892813174 Chr.1: 236216412-236216412 on Build GRCh38
rs1011107851 Chr.1: 236216265-236216265 on Build GRCh38
rs577674958 Chr.1: 236216230-236216230 on Build GRCh38
rs149030117 Chr.1: 236216325-236216325 on Build GRCh38
rs757537043 Chr.1: 236216389-236216392 on Build GRCh38
rs941255715 Chr.1: 236216119-236216119 on Build GRCh38
rs540943841 Chr.1: 236216173-236216173 on Build GRCh38
rs918423301 Chr.1: 236216134-236216134 on Build GRCh38
rs754976779 Chr.1: 236216322-236216322 on Build GRCh38
rs1008659034 Chr.1: 236216210-236216210 on Build GRCh38
rs539964750 Chr.1: 236216409-236216409 on Build GRCh38
rs555262280 Chr.1: 236216138-236216138 on Build GRCh38
rs1050932815 Chr.1: 236216357-236216357 on Build GRCh38
rs990791614 Chr.1: 236216074-236216074 on Build GRCh38
rs868707479 Chr.1: 236216015-236216015 on Build GRCh38
rs571618225 Chr.1: 236216404-236216404 on Build GRCh38
rs913445173 Chr.1: 236216374-236216374 on Build GRCh38
rs1022418763 Chr.1: 236215976-236215976 on Build GRCh38
rs150317762 Chr.1: 236215995-236215995 on Build GRCh38
rs1039324520 Chr.1: 236216246-236216246 on Build GRCh38
rs1017834248 Chr.1: 236216229-236216229 on Build GRCh38
rs942449554 Chr.1: 236216369-236216369 on Build GRCh38
rs920536492 Chr.1: 236215938-236215938 on Build GRCh38
rs545029185 Chr.1: 236216261-236216261 on Build GRCh38
rs138528327 Chr.1: 236216367-236216367 on Build GRCh38
rs753387040 Chr.1: 236215965-236215966 on Build GRCh38
rs758402792 Chr.1: 236215987-236215987 on Build GRCh38
rs1022470946 Chr.1: 236216285-236216285 on Build GRCh38
rs536799641 Chr.1: 236216061-236216061 on Build GRCh38
rs962668203 Chr.1: 236216337-236216337 on Build GRCh38
rs890849622 Chr.1: 236216222-236216222 on Build GRCh38
rs548105590 Chr.1: 236216149-236216149 on Build GRCh38
rs573813327 Chr.1: 236216145-236216145 on Build GRCh38
rs916529880 Chr.1: 236216105-236216105 on Build GRCh38
rs928033096 Chr.1: 236216038-236216038 on Build GRCh38
rs964525071 Chr.1: 236216248-236216248 on Build GRCh38
rs999676345 Chr.1: 236216308-236216308 on Build GRCh38
rs2463193 Chr.1: 236216323-236216323 on Build GRCh38
rs538665014 Chr.1: 236215975-236215975 on Build GRCh38
rs929809328 Chr.1: 236216346-236216346 on Build GRCh38
rs563321309 Chr.1: 236216270-236216270 on Build GRCh38
rs1017823647 Chr.1: 236215939-236215939 on Build GRCh38

View Full Product Details