Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 242124333-242124880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLD5
Entrez Gene ID
200150
Gene Name
phospholipase D family member 5
Gene Aliases
PLDC
UniGene
Hs.672452
Gene Chromosome Location
Chr.1: 242085051-242530546 on Build GRCh38
Transcripts
NM_001195811.1 NM_001195812.1 NM_001320272.1 NM_152666.2 XM_006711752.2 XM_011544115.2 XM_011544116.2 XM_011544119.2 XM_011544120.2 XM_011544121.2 XM_011544122.2 XM_017000567.1 XM_017000568.1 XM_017000569.1 XM_017000570.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3486221 c.604G>A p.E202K Chr.1: 242124521-242124521 on Build GRCh38
5701811 c.838_839CC>AA p.P280N Chr.1: 242124562-242124563 on Build GRCh38
5448480 c.643A>C p.S215R Chr.1: 242124572-242124572 on Build GRCh38
6125597 c.832A>C p.R278R Chr.1: 242124569-242124569 on Build GRCh38
6042431 c.657+6T>C p.? Chr.1: 242124462-242124462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967938870 Chr.1: 242124469-242124469 on Build GRCh38
rs1040186013 Chr.1: 242124732-242124732 on Build GRCh38
rs558332566 Chr.1: 242124654-242124654 on Build GRCh38
rs955599469 Chr.1: 242124385-242124385 on Build GRCh38
rs759082075 Chr.1: 242124638-242124638 on Build GRCh38
rs201173621 Chr.1: 242124675-242124675 on Build GRCh38
rs950086287 Chr.1: 242124585-242124585 on Build GRCh38
rs772481464 Chr.1: 242124420-242124420 on Build GRCh38
rs767140158 Chr.1: 242124506-242124506 on Build GRCh38
rs764369508 Chr.1: 242124668-242124668 on Build GRCh38
rs746013947 Chr.1: 242124584-242124584 on Build GRCh38
rs965703423 Chr.1: 242124394-242124394 on Build GRCh38
rs758573625 Chr.1: 242124711-242124711 on Build GRCh38
rs1020569355 Chr.1: 242124460-242124460 on Build GRCh38
rs561468899 Chr.1: 242124754-242124754 on Build GRCh38
rs760759650 Chr.1: 242124721-242124721 on Build GRCh38
rs769511896 Chr.1: 242124592-242124592 on Build GRCh38
rs544065464 Chr.1: 242124688-242124688 on Build GRCh38
rs770909623 Chr.1: 242124606-242124606 on Build GRCh38
rs1018587279 Chr.1: 242124384-242124384 on Build GRCh38
rs10926623 Chr.1: 242124382-242124382 on Build GRCh38
rs771730067 Chr.1: 242124643-242124643 on Build GRCh38
rs775026704 Chr.1: 242124653-242124653 on Build GRCh38
rs780286397 Chr.1: 242124713-242124713 on Build GRCh38
rs780377710 Chr.1: 242124431-242124432 on Build GRCh38
rs758663255 Chr.1: 242124589-242124589 on Build GRCh38
rs746091574 Chr.1: 242124417-242124417 on Build GRCh38
rs372647511 Chr.1: 242124427-242124427 on Build GRCh38
rs148872073 Chr.1: 242124537-242124537 on Build GRCh38
rs181551327 Chr.1: 242124773-242124773 on Build GRCh38
rs765880795 Chr.1: 242124472-242124472 on Build GRCh38
rs759172020 Chr.1: 242124489-242124489 on Build GRCh38
rs764111369 Chr.1: 242124793-242124793 on Build GRCh38
rs538684831 Chr.1: 242124387-242124387 on Build GRCh38
rs991533360 Chr.1: 242124800-242124800 on Build GRCh38
rs1000870534 Chr.1: 242124792-242124792 on Build GRCh38
rs762414075 Chr.1: 242124456-242124456 on Build GRCh38
rs878865333 Chr.1: 242124440-242124440 on Build GRCh38
rs919202192 Chr.1: 242124618-242124618 on Build GRCh38
rs757338742 Chr.1: 242124564-242124564 on Build GRCh38
rs765886579 Chr.1: 242124691-242124691 on Build GRCh38
rs779124256 Chr.1: 242124569-242124569 on Build GRCh38
rs917872181 Chr.1: 242124687-242124687 on Build GRCh38
rs538505826 Chr.1: 242124596-242124596 on Build GRCh38
rs376737481 Chr.1: 242124437-242124437 on Build GRCh38
rs888955295 Chr.1: 242124708-242124708 on Build GRCh38
rs750509646 Chr.1: 242124710-242124710 on Build GRCh38
rs917210751 Chr.1: 242124558-242124558 on Build GRCh38
rs1020937027 Chr.1: 242124774-242124774 on Build GRCh38
rs200034961 Chr.1: 242124539-242124539 on Build GRCh38
rs1027796668 Chr.1: 242124365-242124365 on Build GRCh38
rs991543214 Chr.1: 242124556-242124556 on Build GRCh38
rs769749511 Chr.1: 242124436-242124436 on Build GRCh38
rs978977041 Chr.1: 242124490-242124490 on Build GRCh38
rs917358393 Chr.1: 242124842-242124842 on Build GRCh38
rs530417017 Chr.1: 242124779-242124779 on Build GRCh38
rs780188956 Chr.1: 242124590-242124590 on Build GRCh38
rs747933843 Chr.1: 242124428-242124428 on Build GRCh38
rs367577812 Chr.1: 242124487-242124487 on Build GRCh38
rs754331875 Chr.1: 242124555-242124555 on Build GRCh38
rs753070481 Chr.1: 242124511-242124511 on Build GRCh38
rs773004331 Chr.1: 242124380-242124380 on Build GRCh38
rs371417144 Chr.1: 242124591-242124591 on Build GRCh38
rs901501910 Chr.1: 242124736-242124736 on Build GRCh38
rs958869916 Chr.1: 242124794-242124794 on Build GRCh38
rs773844950 Chr.1: 242124627-242124627 on Build GRCh38
rs375910427 Chr.1: 242124446-242124446 on Build GRCh38
rs949266103 Chr.1: 242124699-242124699 on Build GRCh38
rs958791634 Chr.1: 242124540-242124540 on Build GRCh38
rs371090789 Chr.1: 242124750-242124750 on Build GRCh38
rs749240389 Chr.1: 242124599-242124599 on Build GRCh38

View Full Product Details