Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244451901-244452412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
UniGene
Hs.498313
Gene Chromosome Location
Chr.1: 244408492-244452134 on Build GRCh38
Transcripts
NM_001126.3 XM_011544138.2 XM_011544142.2 XM_017000943.1 XM_017000946.1 XM_017000947.1 XM_017000949.1 XM_017000950.1 XM_017000951.1 XM_017000952.1 XM_017000953.1

Gene Symbol
C1orf101
Entrez Gene ID
257044
Gene Name
chromosome 1 open reading frame 101
Gene Aliases
-
UniGene
Hs.459534
Gene Chromosome Location
Chr.1: 244452014-244640362 on Build GRCh38
Transcripts
NM_001126.3 XM_011544138.2 XM_011544142.2 XM_017000943.1 XM_017000946.1 XM_017000947.1 XM_017000949.1 XM_017000950.1 XM_017000951.1 XM_017000952.1 XM_017000953.1

SNP Detail

SNP ID Chromosome Location
rs944944049 Chr.1: 244452244-244452244 on Build GRCh38
rs148749464 Chr.1: 244452156-244452156 on Build GRCh38
rs1054331687 Chr.1: 244452225-244452225 on Build GRCh38
rs924190694 Chr.1: 244452186-244452186 on Build GRCh38
rs916850141 Chr.1: 244452007-244452007 on Build GRCh38
rs1050165456 Chr.1: 244452261-244452261 on Build GRCh38
rs946913983 Chr.1: 244452053-244452053 on Build GRCh38
rs951832536 Chr.1: 244452095-244452095 on Build GRCh38
rs995910307 Chr.1: 244452078-244452078 on Build GRCh38
rs916590922 Chr.1: 244452134-244452134 on Build GRCh38
rs74151394 Chr.1: 244452307-244452307 on Build GRCh38
rs902716712 Chr.1: 244452047-244452047 on Build GRCh38
rs761496921 Chr.1: 244452386-244452387 on Build GRCh38
rs771309961 Chr.1: 244452131-244452131 on Build GRCh38
rs902781451 Chr.1: 244452055-244452055 on Build GRCh38
rs982361775 Chr.1: 244452185-244452185 on Build GRCh38
rs564510210 Chr.1: 244452293-244452293 on Build GRCh38
rs201407724 Chr.1: 244452301-244452301 on Build GRCh38
rs113254054 Chr.1: 244452097-244452097 on Build GRCh38
rs1042951644 Chr.1: 244452213-244452213 on Build GRCh38
rs1023300774 Chr.1: 244452085-244452085 on Build GRCh38
rs940014305 Chr.1: 244452003-244452003 on Build GRCh38
rs971728197 Chr.1: 244452116-244452116 on Build GRCh38
rs1046612461 Chr.1: 244452038-244452038 on Build GRCh38
rs954670927 Chr.1: 244452069-244452069 on Build GRCh38
rs959685573 Chr.1: 244451957-244451957 on Build GRCh38
rs1019232191 Chr.1: 244451938-244451957 on Build GRCh38
rs949644482 Chr.1: 244452033-244452033 on Build GRCh38
rs893957836 Chr.1: 244452042-244452042 on Build GRCh38
rs556630544 Chr.1: 244451962-244451962 on Build GRCh38
rs1008843469 Chr.1: 244452071-244452076 on Build GRCh38
rs1048821528 Chr.1: 244452323-244452323 on Build GRCh38
rs893570997 Chr.1: 244452239-244452239 on Build GRCh38
rs907798704 Chr.1: 244451977-244451977 on Build GRCh38
rs928037740 Chr.1: 244452005-244452005 on Build GRCh38
rs920029705 Chr.1: 244452101-244452101 on Build GRCh38
rs564269064 Chr.1: 244452312-244452312 on Build GRCh38
rs1014837217 Chr.1: 244452074-244452074 on Build GRCh38
rs981417485 Chr.1: 244452118-244452118 on Build GRCh38
rs1027876125 Chr.1: 244452353-244452353 on Build GRCh38
rs991339775 Chr.1: 244452368-244452368 on Build GRCh38
rs367782386 Chr.1: 244452054-244452054 on Build GRCh38
rs990853347 Chr.1: 244452133-244452133 on Build GRCh38
rs535116031 Chr.1: 244452037-244452037 on Build GRCh38
rs1027435916 Chr.1: 244452082-244452082 on Build GRCh38
rs186209993 Chr.1: 244452356-244452356 on Build GRCh38
rs1055501344 Chr.1: 244452002-244452003 on Build GRCh38
rs1040450135 Chr.1: 244452064-244452064 on Build GRCh38
rs959389244 Chr.1: 244452146-244452146 on Build GRCh38
rs748845200 Chr.1: 244451987-244451987 on Build GRCh38
rs938089459 Chr.1: 244452036-244452036 on Build GRCh38
rs900649275 Chr.1: 244452069-244452076 on Build GRCh38
rs946861549 Chr.1: 244452208-244452208 on Build GRCh38
rs1012618470 Chr.1: 244451961-244451961 on Build GRCh38
rs1008560430 Chr.1: 244452304-244452304 on Build GRCh38
rs951885013 Chr.1: 244451947-244451947 on Build GRCh38
rs1003184343 Chr.1: 244452381-244452381 on Build GRCh38
rs1004685297 Chr.1: 244452345-244452345 on Build GRCh38
rs1027489760 Chr.1: 244451933-244451933 on Build GRCh38
rs900207147 Chr.1: 244452077-244452077 on Build GRCh38
rs915338510 Chr.1: 244452181-244452181 on Build GRCh38
rs577803098 Chr.1: 244452346-244452346 on Build GRCh38
rs897711520 Chr.1: 244452331-244452331 on Build GRCh38
rs971294258 Chr.1: 244452358-244452358 on Build GRCh38
rs1014805568 Chr.1: 244452103-244452103 on Build GRCh38
rs995866835 Chr.1: 244452067-244452067 on Build GRCh38
rs887528999 Chr.1: 244452334-244452334 on Build GRCh38
rs924180127 Chr.1: 244452229-244452229 on Build GRCh38
rs931702204 Chr.1: 244452257-244452257 on Build GRCh38
rs865994647 Chr.1: 244452091-244452091 on Build GRCh38
rs554958832 Chr.1: 244452062-244452062 on Build GRCh38
rs935506869 Chr.1: 244452214-244452214 on Build GRCh38
rs1040655352 Chr.1: 244452290-244452290 on Build GRCh38
rs575969038 Chr.1: 244452004-244452004 on Build GRCh38

View Full Product Details