Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 998906-999445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP4
Entrez Gene ID
23560
Gene Name
GTP binding protein 4
Gene Aliases
CRFG, NGB, NOG1
UniGene
Hs.215766
Gene Chromosome Location
Chr.10: 988409-1017768 on Build GRCh38
Transcripts
NM_012341.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5016976 c.577G>A p.V193M Chr.10: 999018-999018 on Build GRCh38
4689244 c.576C>T p.D192D Chr.10: 999017-999017 on Build GRCh38
3806360 c.597G>A p.A199A Chr.10: 999038-999038 on Build GRCh38
6676946 c.596C>T p.A199V Chr.10: 999037-999037 on Build GRCh38
307680 c.654G>T p.Q218H Chr.10: 999095-999095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904344555 Chr.10: 999288-999288 on Build GRCh38
rs529872363 Chr.10: 999080-999080 on Build GRCh38
rs537409728 Chr.10: 999199-999199 on Build GRCh38
rs969262191 Chr.10: 998948-998948 on Build GRCh38
rs982998653 Chr.10: 999165-999165 on Build GRCh38
rs1000290729 Chr.10: 999386-999386 on Build GRCh38
rs527995239 Chr.10: 999290-999291 on Build GRCh38
rs765719251 Chr.10: 998954-998954 on Build GRCh38
rs964241278 Chr.10: 999369-999369 on Build GRCh38
rs3763720 Chr.10: 999159-999159 on Build GRCh38
rs184361372 Chr.10: 999420-999420 on Build GRCh38
rs772063679 Chr.10: 999110-999111 on Build GRCh38
rs987314875 Chr.10: 999349-999349 on Build GRCh38
rs1041589917 Chr.10: 999279-999279 on Build GRCh38
rs770948893 Chr.10: 999021-999021 on Build GRCh38
rs995538425 Chr.10: 999304-999305 on Build GRCh38
rs1020524639 Chr.10: 999031-999031 on Build GRCh38
rs772394354 Chr.10: 999014-999014 on Build GRCh38
rs951483748 Chr.10: 999312-999312 on Build GRCh38
rs770869739 Chr.10: 999377-999377 on Build GRCh38
rs974529464 Chr.10: 998976-998976 on Build GRCh38
rs936984954 Chr.10: 999233-999233 on Build GRCh38
rs568496150 Chr.10: 999180-999180 on Build GRCh38
rs567535488 Chr.10: 999038-999038 on Build GRCh38
rs973553280 Chr.10: 999191-999191 on Build GRCh38
rs895765335 Chr.10: 999270-999270 on Build GRCh38
rs936360174 Chr.10: 999119-999119 on Build GRCh38
rs151220214 Chr.10: 999008-999008 on Build GRCh38
rs892236075 Chr.10: 999172-999172 on Build GRCh38
rs774838074 Chr.10: 999169-999169 on Build GRCh38
rs763859284 Chr.10: 999059-999059 on Build GRCh38
rs376110021 Chr.10: 999037-999037 on Build GRCh38
rs1024817285 Chr.10: 998947-998947 on Build GRCh38
rs926986778 Chr.10: 999227-999227 on Build GRCh38
rs753057229 Chr.10: 998955-998955 on Build GRCh38
rs878972374 Chr.10: 999060-999060 on Build GRCh38
rs1037394913 Chr.10: 998932-998932 on Build GRCh38
rs201529735 Chr.10: 999022-999022 on Build GRCh38
rs778732676 Chr.10: 998962-998963 on Build GRCh38
rs554327450 Chr.10: 999338-999338 on Build GRCh38
rs761476712 Chr.10: 999069-999069 on Build GRCh38
rs749865001 Chr.10: 999085-999085 on Build GRCh38
rs945948568 Chr.10: 999274-999274 on Build GRCh38
rs1002067873 Chr.10: 999262-999262 on Build GRCh38
rs905522548 Chr.10: 998933-998933 on Build GRCh38
rs771221481 Chr.10: 999110-999110 on Build GRCh38
rs749415149 Chr.10: 999307-999307 on Build GRCh38
rs760080491 Chr.10: 998953-998953 on Build GRCh38
rs373574581 Chr.10: 999033-999033 on Build GRCh38
rs751142766 Chr.10: 998972-998972 on Build GRCh38
rs748108407 Chr.10: 999091-999092 on Build GRCh38
rs747218501 Chr.10: 999102-999102 on Build GRCh38
rs574764467 Chr.10: 999275-999275 on Build GRCh38
rs1009996576 Chr.10: 999018-999018 on Build GRCh38
rs1000688320 Chr.10: 999053-999053 on Build GRCh38
rs746844431 Chr.10: 999368-999368 on Build GRCh38
rs977002484 Chr.10: 999107-999107 on Build GRCh38
rs779828418 Chr.10: 999007-999007 on Build GRCh38
rs192302912 Chr.10: 999203-999203 on Build GRCh38
rs748376784 Chr.10: 999036-999036 on Build GRCh38
rs763653035 Chr.10: 998961-998961 on Build GRCh38
rs755984227 Chr.10: 998990-998990 on Build GRCh38
rs780638455 Chr.10: 998986-998986 on Build GRCh38
rs745824292 Chr.10: 998989-998989 on Build GRCh38
rs763215962 Chr.10: 999054-999054 on Build GRCh38
rs554846318 Chr.10: 999244-999244 on Build GRCh38
rs368183005 Chr.10: 998962-998962 on Build GRCh38
rs912733839 Chr.10: 999367-999367 on Build GRCh38
rs779953150 Chr.10: 999087-999087 on Build GRCh38
rs1029894569 Chr.10: 999114-999114 on Build GRCh38
rs141450164 Chr.10: 999095-999095 on Build GRCh38
rs910022076 Chr.10: 999179-999179 on Build GRCh38
rs369810568 Chr.10: 999028-999028 on Build GRCh38
rs773282249 Chr.10: 999017-999017 on Build GRCh38
rs540486022 Chr.10: 999278-999278 on Build GRCh38
rs566484717 Chr.10: 999088-999088 on Build GRCh38
rs546730627 Chr.10: 999086-999086 on Build GRCh38
rs754754673 Chr.10: 998943-998943 on Build GRCh38
rs60965306 Chr.10: 998973-998973 on Build GRCh38

View Full Product Details