Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72007524-72008014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST3
Entrez Gene ID
9469
Gene Name
carbohydrate sulfotransferase 3
Gene Aliases
C6ST, C6ST1, HSD
UniGene
Hs.158304
Gene Chromosome Location
Chr.10: 71964362-72013564 on Build GRCh38
Transcripts
NM_004273.4 XM_006718075.3 XM_011540369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3382989 c.886G>A p.V296I Chr.10: 72007917-72007917 on Build GRCh38
4598679 c.793C>A p.R265S Chr.10: 72007824-72007824 on Build GRCh38
3375526 c.768C>T p.N256N Chr.10: 72007799-72007799 on Build GRCh38
4596154 c.532G>A p.G178S Chr.10: 72007563-72007563 on Build GRCh38
2161366 c.567C>T p.R189R Chr.10: 72007598-72007598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752600315 Chr.10: 72007857-72007857 on Build GRCh38
rs767914471 Chr.10: 72007748-72007748 on Build GRCh38
rs762277214 Chr.10: 72007831-72007831 on Build GRCh38
rs751961615 Chr.10: 72007549-72007549 on Build GRCh38
rs974101244 Chr.10: 72007619-72007619 on Build GRCh38
rs893919771 Chr.10: 72007901-72007901 on Build GRCh38
rs772237363 Chr.10: 72007876-72007876 on Build GRCh38
rs570484661 Chr.10: 72007703-72007703 on Build GRCh38
rs749239221 Chr.10: 72007868-72007868 on Build GRCh38
rs1050116997 Chr.10: 72007742-72007742 on Build GRCh38
rs145440508 Chr.10: 72007655-72007655 on Build GRCh38
rs762224143 Chr.10: 72007917-72007917 on Build GRCh38
rs763485176 Chr.10: 72007994-72007994 on Build GRCh38
rs576130316 Chr.10: 72007800-72007800 on Build GRCh38
rs567771631 Chr.10: 72007544-72007544 on Build GRCh38
rs201963050 Chr.10: 72007558-72007558 on Build GRCh38
rs992053096 Chr.10: 72007565-72007565 on Build GRCh38
rs969298186 Chr.10: 72007626-72007626 on Build GRCh38
rs768853633 Chr.10: 72007828-72007828 on Build GRCh38
rs200988207 Chr.10: 72007788-72007788 on Build GRCh38
rs760887436 Chr.10: 72007752-72007752 on Build GRCh38
rs757896210 Chr.10: 72007798-72007798 on Build GRCh38
rs373546394 Chr.10: 72007916-72007916 on Build GRCh38
rs759796765 Chr.10: 72007735-72007735 on Build GRCh38
rs762393741 Chr.10: 72007847-72007847 on Build GRCh38
rs121908617 Chr.10: 72007695-72007695 on Build GRCh38
rs747175788 Chr.10: 72007886-72007886 on Build GRCh38
rs769540174 Chr.10: 72007559-72007559 on Build GRCh38
rs565957683 Chr.10: 72007664-72007664 on Build GRCh38
rs121908616 Chr.10: 72007807-72007807 on Build GRCh38
rs754258548 Chr.10: 72007760-72007760 on Build GRCh38
rs780204389 Chr.10: 72007885-72007885 on Build GRCh38
rs775287488 Chr.10: 72007583-72007583 on Build GRCh38
rs771814734 Chr.10: 72007580-72007580 on Build GRCh38
rs772288774 Chr.10: 72007812-72007812 on Build GRCh38
rs1023609671 Chr.10: 72007984-72007984 on Build GRCh38
rs981969686 Chr.10: 72007649-72007649 on Build GRCh38
rs121908619 Chr.10: 72007634-72007634 on Build GRCh38
rs866470808 Chr.10: 72007726-72007726 on Build GRCh38
rs746848315 Chr.10: 72007692-72007692 on Build GRCh38
rs756623618 Chr.10: 72007566-72007566 on Build GRCh38
rs779740819 Chr.10: 72007779-72007779 on Build GRCh38
rs754684832 Chr.10: 72007853-72007853 on Build GRCh38
rs768963465 Chr.10: 72007900-72007900 on Build GRCh38
rs28937593 Chr.10: 72007942-72007942 on Build GRCh38
rs768626435 Chr.10: 72007697-72007697 on Build GRCh38
rs749977155 Chr.10: 72007768-72007768 on Build GRCh38
rs367857089 Chr.10: 72007601-72007601 on Build GRCh38
rs764522356 Chr.10: 72007757-72007757 on Build GRCh38
rs749924021 Chr.10: 72007837-72007837 on Build GRCh38
rs121908618 Chr.10: 72007951-72007951 on Build GRCh38
rs150965438 Chr.10: 72007562-72007562 on Build GRCh38
rs764565664 Chr.10: 72007658-72007658 on Build GRCh38
rs748124800 Chr.10: 72007700-72007700 on Build GRCh38
rs371650865 Chr.10: 72007665-72007665 on Build GRCh38
rs903606184 Chr.10: 72007893-72007893 on Build GRCh38
rs999197667 Chr.10: 72007941-72007941 on Build GRCh38
rs775980360 Chr.10: 72007751-72007751 on Build GRCh38
rs774652828 Chr.10: 72007602-72007602 on Build GRCh38
rs758043795 Chr.10: 72007769-72007769 on Build GRCh38
rs868146865 Chr.10: 72007884-72007884 on Build GRCh38
rs774599785 Chr.10: 72007719-72007719 on Build GRCh38
rs759873741 Chr.10: 72007613-72007613 on Build GRCh38
rs774572727 Chr.10: 72007558-72007559 on Build GRCh38
rs748812857 Chr.10: 72007571-72007571 on Build GRCh38
rs764704926 Chr.10: 72007762-72007762 on Build GRCh38
rs1013851768 Chr.10: 72007958-72007958 on Build GRCh38
rs754665395 Chr.10: 72007790-72007790 on Build GRCh38
rs760987231 Chr.10: 72007824-72007824 on Build GRCh38
rs756637194 Chr.10: 72007657-72007657 on Build GRCh38
rs1031171690 Chr.10: 72007946-72007946 on Build GRCh38
rs896297940 Chr.10: 72007950-72007950 on Build GRCh38
rs768518290 Chr.10: 72007584-72007584 on Build GRCh38
rs781075355 Chr.10: 72007791-72007791 on Build GRCh38
rs745523940 Chr.10: 72007575-72007575 on Build GRCh38
rs867366138 Chr.10: 72007974-72007974 on Build GRCh38
rs777792605 Chr.10: 72007867-72007867 on Build GRCh38
rs1041291103 Chr.10: 72007827-72007827 on Build GRCh38
rs369711355 Chr.10: 72007856-72007856 on Build GRCh38
rs779793827 Chr.10: 72007686-72007686 on Build GRCh38
rs121908620 Chr.10: 72007888-72007888 on Build GRCh38
rs930383426 Chr.10: 72007699-72007699 on Build GRCh38
rs535309495 Chr.10: 72007990-72007990 on Build GRCh38
rs751135376 Chr.10: 72007849-72007849 on Build GRCh38
rs373748456 Chr.10: 72007559-72007559 on Build GRCh38
rs775505653 Chr.10: 72007818-72007818 on Build GRCh38
rs777302566 Chr.10: 72007568-72007568 on Build GRCh38
rs147804585 Chr.10: 72007592-72007592 on Build GRCh38
rs754380752 Chr.10: 72007659-72007659 on Build GRCh38
rs1053022692 Chr.10: 72007864-72007864 on Build GRCh38
rs770159255 Chr.10: 72007952-72007952 on Build GRCh38
rs778478785 Chr.10: 72007574-72007574 on Build GRCh38
rs1010927640 Chr.10: 72007973-72007973 on Build GRCh38
rs140547825 Chr.10: 72007859-72007859 on Build GRCh38
rs765880880 Chr.10: 72007848-72007848 on Build GRCh38
rs758095258 Chr.10: 72007672-72007672 on Build GRCh38
rs762274724 Chr.10: 72007571-72007573 on Build GRCh38
rs149671252 Chr.10: 72007628-72007628 on Build GRCh38
rs942951728 Chr.10: 72007822-72007822 on Build GRCh38
rs776855980 Chr.10: 72007902-72007902 on Build GRCh38
rs745999727 Chr.10: 72007806-72007806 on Build GRCh38
rs765800675 Chr.10: 72007832-72007832 on Build GRCh38
rs771098871 Chr.10: 72007706-72007706 on Build GRCh38
rs539386405 Chr.10: 72007721-72007721 on Build GRCh38

View Full Product Details