Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72061742-72062240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPOCK2
Entrez Gene ID
9806
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2
Gene Aliases
testican-2
UniGene
Hs.523009
Gene Chromosome Location
Chr.10: 72059034-72089032 on Build GRCh38
Transcripts
NM_001244950.1 NM_014767.2 XM_011540404.2

SNP Detail

SNP ID Chromosome Location
rs576550926 Chr.10: 72062124-72062124 on Build GRCh38
rs539922519 Chr.10: 72062047-72062047 on Build GRCh38
rs1026070505 Chr.10: 72062127-72062127 on Build GRCh38
rs138566541 Chr.10: 72061864-72061864 on Build GRCh38
rs556288922 Chr.10: 72061938-72061938 on Build GRCh38
rs75548271 Chr.10: 72061910-72061910 on Build GRCh38
rs796503426 Chr.10: 72062117-72062117 on Build GRCh38
rs991953859 Chr.10: 72061822-72061822 on Build GRCh38
rs569043550 Chr.10: 72061981-72061981 on Build GRCh38
rs575457188 Chr.10: 72061973-72061973 on Build GRCh38
rs797002269 Chr.10: 72061763-72061764 on Build GRCh38
rs979795618 Chr.10: 72062019-72062019 on Build GRCh38
rs879773142 Chr.10: 72061880-72061880 on Build GRCh38
rs1668174 Chr.10: 72061905-72061905 on Build GRCh38
rs969791069 Chr.10: 72062011-72062012 on Build GRCh38
rs1678623 Chr.10: 72061875-72061875 on Build GRCh38
rs552739963 Chr.10: 72061884-72061884 on Build GRCh38
rs910155359 Chr.10: 72061893-72061893 on Build GRCh38
rs564601948 Chr.10: 72062101-72062101 on Build GRCh38
rs968169610 Chr.10: 72061911-72061911 on Build GRCh38
rs536026633 Chr.10: 72062128-72062128 on Build GRCh38
rs192647964 Chr.10: 72062013-72062013 on Build GRCh38
rs1047599010 Chr.10: 72061816-72061816 on Build GRCh38
rs960808987 Chr.10: 72061790-72061790 on Build GRCh38
rs920431542 Chr.10: 72062026-72062028 on Build GRCh38
rs3740131 Chr.10: 72061763-72061763 on Build GRCh38
rs1031706599 Chr.10: 72061925-72061925 on Build GRCh38
rs996265617 Chr.10: 72062195-72062195 on Build GRCh38
rs1043721912 Chr.10: 72062021-72062021 on Build GRCh38
rs960609003 Chr.10: 72062118-72062118 on Build GRCh38
rs184884256 Chr.10: 72061984-72061984 on Build GRCh38
rs140524322 Chr.10: 72061990-72061991 on Build GRCh38
rs999790208 Chr.10: 72061916-72061916 on Build GRCh38
rs964341402 Chr.10: 72062188-72062188 on Build GRCh38
rs751331187 Chr.10: 72061894-72061894 on Build GRCh38
rs975675827 Chr.10: 72062213-72062213 on Build GRCh38
rs111444916 Chr.10: 72061992-72061992 on Build GRCh38
rs779334907 Chr.10: 72062132-72062132 on Build GRCh38
rs908913772 Chr.10: 72062171-72062171 on Build GRCh38
rs1051192264 Chr.10: 72061982-72061982 on Build GRCh38
rs1033585364 Chr.10: 72062113-72062113 on Build GRCh38
rs562352979 Chr.10: 72062163-72062163 on Build GRCh38
rs903837145 Chr.10: 72062029-72062029 on Build GRCh38
rs921477294 Chr.10: 72061934-72061934 on Build GRCh38

View Full Product Details