Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88815230-88815745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPM
Entrez Gene ID
340654
Gene Name
lipase family member M
Gene Aliases
LIPL3, bA304I5.1
UniGene
Hs.55118
Gene Chromosome Location
Chr.10: 88802730-88820857 on Build GRCh38
Transcripts
NM_001128215.1 XM_011539748.2 XM_011539751.2 XM_011539752.2 XM_011539753.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6007639 c.823C>A p.L275I Chr.10: 88815468-88815468 on Build GRCh38
4559208 c.796G>A p.D266N Chr.10: 88815441-88815441 on Build GRCh38
7283344 c.724delA p.E244fs*20 Chr.10: 88815369-88815369 on Build GRCh38
7329754 c.837C>G p.F279L Chr.10: 88815482-88815482 on Build GRCh38
4440745 c.858+6T>A p.? Chr.10: 88815509-88815509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750911666 Chr.10: 88815548-88815548 on Build GRCh38
rs151149351 Chr.10: 88815289-88815289 on Build GRCh38
rs3910680 Chr.10: 88815466-88815466 on Build GRCh38
rs1004371178 Chr.10: 88815505-88815505 on Build GRCh38
rs905753283 Chr.10: 88815570-88815570 on Build GRCh38
rs534491414 Chr.10: 88815370-88815370 on Build GRCh38
rs61737049 Chr.10: 88815494-88815494 on Build GRCh38
rs117451088 Chr.10: 88815605-88815605 on Build GRCh38
rs902422599 Chr.10: 88815496-88815496 on Build GRCh38
rs965830498 Chr.10: 88815518-88815518 on Build GRCh38
rs748511022 Chr.10: 88815371-88815371 on Build GRCh38
rs771820380 Chr.10: 88815332-88815332 on Build GRCh38
rs952335496 Chr.10: 88815553-88815553 on Build GRCh38
rs770351112 Chr.10: 88815413-88815413 on Build GRCh38
rs984982329 Chr.10: 88815278-88815278 on Build GRCh38
rs771800221 Chr.10: 88815664-88815664 on Build GRCh38
rs558852322 Chr.10: 88815315-88815315 on Build GRCh38
rs373741061 Chr.10: 88815517-88815517 on Build GRCh38
rs756463502 Chr.10: 88815600-88815600 on Build GRCh38
rs937390573 Chr.10: 88815451-88815451 on Build GRCh38
rs184848760 Chr.10: 88815415-88815415 on Build GRCh38
rs371872718 Chr.10: 88815256-88815256 on Build GRCh38
rs1045707854 Chr.10: 88815368-88815368 on Build GRCh38
rs61737047 Chr.10: 88815500-88815500 on Build GRCh38
rs763528896 Chr.10: 88815420-88815420 on Build GRCh38
rs989076108 Chr.10: 88815632-88815632 on Build GRCh38
rs984909007 Chr.10: 88815562-88815562 on Build GRCh38
rs898598292 Chr.10: 88815537-88815537 on Build GRCh38
rs928740656 Chr.10: 88815374-88815374 on Build GRCh38
rs556686565 Chr.10: 88815481-88815481 on Build GRCh38
rs996298262 Chr.10: 88815533-88815533 on Build GRCh38
rs574582328 Chr.10: 88815440-88815440 on Build GRCh38
rs565207277 Chr.10: 88815704-88815704 on Build GRCh38
rs932923772 Chr.10: 88815471-88815471 on Build GRCh38
rs1021364059 Chr.10: 88815698-88815698 on Build GRCh38
rs1029179229 Chr.10: 88815545-88815545 on Build GRCh38
rs754883952 Chr.10: 88815257-88815257 on Build GRCh38
rs764404510 Chr.10: 88815518-88815528 on Build GRCh38
rs894948265 Chr.10: 88815459-88815459 on Build GRCh38
rs995505458 Chr.10: 88815549-88815549 on Build GRCh38
rs928599863 Chr.10: 88815610-88815610 on Build GRCh38
rs570763647 Chr.10: 88815326-88815326 on Build GRCh38
rs1023399142 Chr.10: 88815567-88815567 on Build GRCh38
rs999229964 Chr.10: 88815511-88815511 on Build GRCh38
rs937460459 Chr.10: 88815655-88815655 on Build GRCh38
rs962231186 Chr.10: 88815602-88815602 on Build GRCh38
rs914347971 Chr.10: 88815351-88815351 on Build GRCh38
rs368948657 Chr.10: 88815561-88815561 on Build GRCh38
rs752975955 Chr.10: 88815550-88815550 on Build GRCh38
rs778700708 Chr.10: 88815566-88815566 on Build GRCh38
rs952341751 Chr.10: 88815265-88815265 on Build GRCh38
rs955857661 Chr.10: 88815630-88815630 on Build GRCh38
rs1054981086 Chr.10: 88815453-88815453 on Build GRCh38
rs368743898 Chr.10: 88815690-88815690 on Build GRCh38
rs775332279 Chr.10: 88815343-88815343 on Build GRCh38
rs766319379 Chr.10: 88815431-88815431 on Build GRCh38
rs968886646 Chr.10: 88815722-88815722 on Build GRCh38

View Full Product Details