Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96949054-96949551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCOR
Entrez Gene ID
84458
Gene Name
ligand dependent nuclear receptor corepressor
Gene Aliases
MLR2
UniGene
Hs.745068
Gene Chromosome Location
Chr.10: 96832250-96964441 on Build GRCh38
Transcripts
NM_001170765.1 NM_001170766.1 NM_032440.3 XM_006718033.3 XM_011540277.2 XM_017016784.1 XM_017016785.1 XM_017016786.1 XM_017016787.1 XM_017016788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1492331 c.4C>T p.Q2* Chr.10: 96949061-96949061 on Build GRCh38
6874177 c.107C>T p.A36V Chr.10: 96949164-96949164 on Build GRCh38
7090112 c.129C>A p.P43P Chr.10: 96949186-96949186 on Build GRCh38
6194150 c.98T>G p.L33R Chr.10: 96949155-96949155 on Build GRCh38
6874176 c.107C>T p.A36V Chr.10: 96949164-96949164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771711824 Chr.10: 96949284-96949284 on Build GRCh38
rs752853388 Chr.10: 96949342-96949342 on Build GRCh38
rs745504616 Chr.10: 96949098-96949098 on Build GRCh38
rs764456104 Chr.10: 96949388-96949388 on Build GRCh38
rs753103078 Chr.10: 96949304-96949304 on Build GRCh38
rs749236880 Chr.10: 96949245-96949245 on Build GRCh38
rs763362938 Chr.10: 96949158-96949158 on Build GRCh38
rs913660387 Chr.10: 96949523-96949523 on Build GRCh38
rs926339849 Chr.10: 96949429-96949429 on Build GRCh38
rs745686634 Chr.10: 96949331-96949331 on Build GRCh38
rs766754480 Chr.10: 96949316-96949316 on Build GRCh38
rs756133931 Chr.10: 96949209-96949209 on Build GRCh38
rs370335700 Chr.10: 96949123-96949123 on Build GRCh38
rs766722071 Chr.10: 96949172-96949172 on Build GRCh38
rs1055157432 Chr.10: 96949351-96949351 on Build GRCh38
rs757161422 Chr.10: 96949273-96949273 on Build GRCh38
rs562229779 Chr.10: 96949352-96949352 on Build GRCh38
rs762257326 Chr.10: 96949125-96949125 on Build GRCh38
rs201502124 Chr.10: 96949076-96949076 on Build GRCh38
rs941057433 Chr.10: 96949153-96949153 on Build GRCh38
rs749034031 Chr.10: 96949085-96949085 on Build GRCh38
rs765665641 Chr.10: 96949133-96949133 on Build GRCh38
rs773575518 Chr.10: 96949142-96949142 on Build GRCh38
rs904122680 Chr.10: 96949327-96949327 on Build GRCh38
rs746469489 Chr.10: 96949289-96949289 on Build GRCh38
rs920730544 Chr.10: 96949202-96949202 on Build GRCh38
rs899705652 Chr.10: 96949345-96949345 on Build GRCh38
rs772606143 Chr.10: 96949213-96949213 on Build GRCh38
rs1015231464 Chr.10: 96949382-96949382 on Build GRCh38
rs113302221 Chr.10: 96949333-96949333 on Build GRCh38
rs759785318 Chr.10: 96949164-96949164 on Build GRCh38
rs976205751 Chr.10: 96949184-96949184 on Build GRCh38
rs765437692 Chr.10: 96949225-96949225 on Build GRCh38
rs770837388 Chr.10: 96949090-96949090 on Build GRCh38
rs199619146 Chr.10: 96949189-96949189 on Build GRCh38
rs778826068 Chr.10: 96949275-96949275 on Build GRCh38
rs151183176 Chr.10: 96949138-96949138 on Build GRCh38
rs965766975 Chr.10: 96949496-96949496 on Build GRCh38
rs1027962735 Chr.10: 96949415-96949415 on Build GRCh38
rs955311564 Chr.10: 96949352-96949355 on Build GRCh38
rs767722121 Chr.10: 96949340-96949340 on Build GRCh38
rs569313645 Chr.10: 96949455-96949455 on Build GRCh38
rs545981191 Chr.10: 96949314-96949314 on Build GRCh38
rs557977659 Chr.10: 96949374-96949374 on Build GRCh38
rs572274617 Chr.10: 96949310-96949310 on Build GRCh38
rs937979258 Chr.10: 96949347-96949347 on Build GRCh38
rs751767388 Chr.10: 96949175-96949175 on Build GRCh38
rs996675213 Chr.10: 96949351-96949352 on Build GRCh38
rs771696930 Chr.10: 96949448-96949449 on Build GRCh38
rs1017249780 Chr.10: 96949485-96949485 on Build GRCh38
rs745526548 Chr.10: 96949282-96949282 on Build GRCh38
rs1007167543 Chr.10: 96949456-96949456 on Build GRCh38
rs117243916 Chr.10: 96949381-96949381 on Build GRCh38
rs61752727 Chr.10: 96949130-96949130 on Build GRCh38
rs771529352 Chr.10: 96949107-96949107 on Build GRCh38
rs767528630 Chr.10: 96949190-96949190 on Build GRCh38
rs373150652 Chr.10: 96949321-96949321 on Build GRCh38
rs376163076 Chr.10: 96949323-96949323 on Build GRCh38
rs752755858 Chr.10: 96949199-96949199 on Build GRCh38
rs1013716062 Chr.10: 96949354-96949354 on Build GRCh38
rs186186209 Chr.10: 96949092-96949092 on Build GRCh38
rs952009463 Chr.10: 96949425-96949425 on Build GRCh38
rs937530448 Chr.10: 96949493-96949493 on Build GRCh38
rs777919365 Chr.10: 96949219-96949219 on Build GRCh38
rs777719136 Chr.10: 96949078-96949078 on Build GRCh38
rs140995155 Chr.10: 96949188-96949188 on Build GRCh38
rs896553759 Chr.10: 96949352-96949352 on Build GRCh38
rs144102057 Chr.10: 96949503-96949503 on Build GRCh38
rs993348201 Chr.10: 96949409-96949409 on Build GRCh38
rs774863095 Chr.10: 96949288-96949288 on Build GRCh38
rs543683601 Chr.10: 96949477-96949477 on Build GRCh38

View Full Product Details