Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97714443-97714977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARVELD1
Entrez Gene ID
83742
Gene Name
MARVEL domain containing 1
Gene Aliases
GB14, MARVD1, MRVLDC1, bA548K23.8
UniGene
Hs.744073
Gene Chromosome Location
Chr.10: 97713708-97718152 on Build GRCh38
Transcripts
NM_031484.3

SNP Detail

SNP ID Chromosome Location
rs970074159 Chr.10: 97714643-97714643 on Build GRCh38
rs955451030 Chr.10: 97714855-97714855 on Build GRCh38
rs934090178 Chr.10: 97714585-97714585 on Build GRCh38
rs940996057 Chr.10: 97714667-97714667 on Build GRCh38
rs541505121 Chr.10: 97714723-97714723 on Build GRCh38
rs962598434 Chr.10: 97714900-97714900 on Build GRCh38
rs1051233681 Chr.10: 97714586-97714586 on Build GRCh38
rs1022746148 Chr.10: 97714590-97714590 on Build GRCh38
rs767360098 Chr.10: 97714536-97714536 on Build GRCh38
rs545804643 Chr.10: 97714546-97714546 on Build GRCh38
rs1006120057 Chr.10: 97714826-97714826 on Build GRCh38
rs1043611793 Chr.10: 97714787-97714787 on Build GRCh38
rs139003280 Chr.10: 97714554-97714554 on Build GRCh38
rs567514376 Chr.10: 97714912-97714912 on Build GRCh38
rs896600459 Chr.10: 97714825-97714825 on Build GRCh38
rs1026582998 Chr.10: 97714477-97714477 on Build GRCh38
rs941122268 Chr.10: 97714469-97714469 on Build GRCh38
rs987324901 Chr.10: 97714873-97714873 on Build GRCh38
rs950621891 Chr.10: 97714523-97714523 on Build GRCh38
rs1000022448 Chr.10: 97714616-97714616 on Build GRCh38
rs1012617597 Chr.10: 97714639-97714639 on Build GRCh38
rs149362453 Chr.10: 97714823-97714823 on Build GRCh38
rs921322287 Chr.10: 97714570-97714570 on Build GRCh38
rs1031012039 Chr.10: 97714624-97714624 on Build GRCh38
rs889614000 Chr.10: 97714775-97714775 on Build GRCh38
rs975457010 Chr.10: 97714564-97714564 on Build GRCh38
rs903860532 Chr.10: 97714809-97714810 on Build GRCh38
rs781666839 Chr.10: 97714503-97714503 on Build GRCh38
rs758687146 Chr.10: 97714813-97714813 on Build GRCh38
rs1050938709 Chr.10: 97714761-97714761 on Build GRCh38
rs1006885176 Chr.10: 97714858-97714858 on Build GRCh38
rs964614059 Chr.10: 97714844-97714844 on Build GRCh38
rs909555237 Chr.10: 97714659-97714659 on Build GRCh38
rs766987627 Chr.10: 97714847-97714847 on Build GRCh38
rs549214138 Chr.10: 97714853-97714853 on Build GRCh38
rs551673767 Chr.10: 97714740-97714740 on Build GRCh38
rs896991596 Chr.10: 97714527-97714527 on Build GRCh38
rs924782103 Chr.10: 97714592-97714592 on Build GRCh38
rs188406722 Chr.10: 97714784-97714784 on Build GRCh38
rs562165077 Chr.10: 97714633-97714633 on Build GRCh38
rs112941381 Chr.10: 97714474-97714474 on Build GRCh38
rs558053682 Chr.10: 97714547-97714547 on Build GRCh38
rs1016166085 Chr.10: 97714828-97714828 on Build GRCh38
rs761701746 Chr.10: 97714524-97714524 on Build GRCh38
rs528494697 Chr.10: 97714727-97714727 on Build GRCh38
rs934847901 Chr.10: 97714921-97714921 on Build GRCh38
rs934922443 Chr.10: 97714631-97714631 on Build GRCh38
rs1013612162 Chr.10: 97714845-97714849 on Build GRCh38
rs960770539 Chr.10: 97714627-97714627 on Build GRCh38
rs41290460 Chr.10: 97714726-97714726 on Build GRCh38
rs924801558 Chr.10: 97714920-97714920 on Build GRCh38
rs576317560 Chr.10: 97714550-97714550 on Build GRCh38
rs972482298 Chr.10: 97714724-97714724 on Build GRCh38
rs1016480040 Chr.10: 97714892-97714893 on Build GRCh38
rs182901208 Chr.10: 97714646-97714646 on Build GRCh38
rs975363622 Chr.10: 97714846-97714846 on Build GRCh38
rs999516853 Chr.10: 97714821-97714821 on Build GRCh38
rs892786667 Chr.10: 97714636-97714636 on Build GRCh38
rs990387209 Chr.10: 97714945-97714945 on Build GRCh38
rs954756126 Chr.10: 97714591-97714603 on Build GRCh38
rs889746729 Chr.10: 97714569-97714569 on Build GRCh38

View Full Product Details