Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 115170708-115171209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRNL1
Entrez Gene ID
26033
Gene Name
attractin like 1
Gene Aliases
ALP, bA338L11.1, bA454H24.1
UniGene
Hs.501127
Gene Chromosome Location
Chr.10: 115093365-115948999 on Build GRCh38
Transcripts
NM_001276282.3 NM_207303.4 NR_074088.2 XM_011539587.1 XM_011539588.2 XM_011539589.2 XM_011539590.2 XM_011539591.2 XM_017016035.1 XM_017016036.1 XM_017016037.1 XM_017016038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5703455 c.1138A>C p.N380H Chr.10: 115171082-115171082 on Build GRCh38
6065073 c.1234G>A p.E412K Chr.10: 115171178-115171178 on Build GRCh38
7256018 c.1252A>G p.I418V Chr.10: 115171196-115171196 on Build GRCh38
6065072 c.1249C>T p.H417Y Chr.10: 115171193-115171193 on Build GRCh38
340321 c.1257G>T p.M419I Chr.10: 115171201-115171201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570594572 Chr.10: 115171025-115171025 on Build GRCh38
rs961661560 Chr.10: 115170787-115170787 on Build GRCh38
rs782812123 Chr.10: 115171002-115171002 on Build GRCh38
rs373819068 Chr.10: 115171125-115171125 on Build GRCh38
rs1030034701 Chr.10: 115170817-115170817 on Build GRCh38
rs1015865283 Chr.10: 115170784-115170784 on Build GRCh38
rs782272274 Chr.10: 115171092-115171092 on Build GRCh38
rs1005732306 Chr.10: 115171143-115171143 on Build GRCh38
rs954495090 Chr.10: 115170846-115170846 on Build GRCh38
rs782249375 Chr.10: 115171083-115171083 on Build GRCh38
rs548558010 Chr.10: 115170840-115170840 on Build GRCh38
rs979055210 Chr.10: 115170898-115170898 on Build GRCh38
rs558935409 Chr.10: 115170983-115170983 on Build GRCh38
rs782444504 Chr.10: 115171103-115171103 on Build GRCh38
rs782070859 Chr.10: 115171166-115171166 on Build GRCh38
rs151311210 Chr.10: 115170771-115170771 on Build GRCh38
rs985905288 Chr.10: 115170848-115170848 on Build GRCh38
rs782567165 Chr.10: 115171108-115171108 on Build GRCh38
rs782673866 Chr.10: 115171085-115171085 on Build GRCh38
rs558181788 Chr.10: 115170855-115170855 on Build GRCh38
rs570343516 Chr.10: 115170892-115170892 on Build GRCh38
rs781868602 Chr.10: 115171034-115171034 on Build GRCh38
rs917331607 Chr.10: 115171012-115171012 on Build GRCh38
rs141967868 Chr.10: 115171112-115171112 on Build GRCh38
rs782514148 Chr.10: 115171087-115171087 on Build GRCh38
rs782441637 Chr.10: 115171158-115171158 on Build GRCh38
rs948782638 Chr.10: 115171050-115171050 on Build GRCh38
rs782726678 Chr.10: 115171028-115171028 on Build GRCh38
rs781810351 Chr.10: 115171003-115171003 on Build GRCh38
rs537021858 Chr.10: 115170960-115170960 on Build GRCh38
rs782347663 Chr.10: 115171116-115171116 on Build GRCh38
rs368688754 Chr.10: 115171175-115171175 on Build GRCh38
rs545393284 Chr.10: 115170946-115170946 on Build GRCh38
rs910336586 Chr.10: 115170889-115170889 on Build GRCh38
rs534829009 Chr.10: 115171044-115171044 on Build GRCh38
rs782022587 Chr.10: 115171165-115171165 on Build GRCh38
rs782696015 Chr.10: 115171168-115171168 on Build GRCh38
rs188137804 Chr.10: 115170793-115170793 on Build GRCh38
rs782242181 Chr.10: 115171145-115171145 on Build GRCh38
rs566319595 Chr.10: 115170783-115170783 on Build GRCh38
rs782126949 Chr.10: 115171182-115171182 on Build GRCh38

View Full Product Details