Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125852450-125852998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCCIP
Entrez Gene ID
56647
Gene Name
BRCA2 and CDKN1A interacting protein
Gene Aliases
TOK-1, TOK1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125823535-125853695 on Build GRCh38
Transcripts
NM_016567.3 NM_018180.2 XM_017016404.1 XM_017016405.1

Gene Symbol
DHX32
Entrez Gene ID
55760
Gene Name
DEAH-box helicase 32 (putative)
Gene Aliases
DDX32, DHLP1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125836337-125896483 on Build GRCh38
Transcripts
NM_016567.3 NM_018180.2 XM_017016404.1 XM_017016405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6643522 c.1121C>T p.S374L Chr.10: 125852614-125852614 on Build GRCh38
244027 c.1096T>C p.Y366H Chr.10: 125852639-125852639 on Build GRCh38
6866811 c.1093G>A p.V365M Chr.10: 125852642-125852642 on Build GRCh38
3978221 c.1152G>A p.Q384Q Chr.10: 125852583-125852583 on Build GRCh38
4677380 c.1188_1190delTTC p.S397delS Chr.10: 125852545-125852547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766216612 Chr.10: 125852669-125852669 on Build GRCh38
rs201263763 Chr.10: 125852617-125852617 on Build GRCh38
rs529609690 Chr.10: 125852949-125852949 on Build GRCh38
rs1007104520 Chr.10: 125852692-125852692 on Build GRCh38
rs1006477455 Chr.10: 125852889-125852889 on Build GRCh38
rs188682116 Chr.10: 125852932-125852932 on Build GRCh38
rs758759796 Chr.10: 125852691-125852692 on Build GRCh38
rs772989683 Chr.10: 125852652-125852652 on Build GRCh38
rs753837269 Chr.10: 125852567-125852567 on Build GRCh38
rs1018123970 Chr.10: 125852718-125852718 on Build GRCh38
rs911423180 Chr.10: 125852633-125852633 on Build GRCh38
rs961381093 Chr.10: 125852573-125852573 on Build GRCh38
rs776137704 Chr.10: 125852614-125852614 on Build GRCh38
rs572182875 Chr.10: 125852861-125852861 on Build GRCh38
rs750220644 Chr.10: 125852556-125852556 on Build GRCh38
rs760827622 Chr.10: 125852950-125852950 on Build GRCh38
rs762558209 Chr.10: 125852521-125852521 on Build GRCh38
rs924067428 Chr.10: 125852708-125852708 on Build GRCh38
rs61757586 Chr.10: 125852572-125852572 on Build GRCh38
rs558546654 Chr.10: 125852905-125852905 on Build GRCh38
rs935385456 Chr.10: 125852723-125852723 on Build GRCh38
rs1018483616 Chr.10: 125852899-125852899 on Build GRCh38
rs896857974 Chr.10: 125852662-125852662 on Build GRCh38
rs376082110 Chr.10: 125852503-125852503 on Build GRCh38
rs374705004 Chr.10: 125852671-125852671 on Build GRCh38
rs766609594 Chr.10: 125852955-125852955 on Build GRCh38
rs758457394 Chr.10: 125852570-125852570 on Build GRCh38
rs776227847 Chr.10: 125852683-125852683 on Build GRCh38
rs558317433 Chr.10: 125852540-125852540 on Build GRCh38
rs367837068 Chr.10: 125852684-125852684 on Build GRCh38
rs574680190 Chr.10: 125852724-125852724 on Build GRCh38
rs375726805 Chr.10: 125852564-125852564 on Build GRCh38
rs573673829 Chr.10: 125852632-125852632 on Build GRCh38
rs150144867 Chr.10: 125852923-125852923 on Build GRCh38
rs1044330755 Chr.10: 125852827-125852827 on Build GRCh38
rs544231546 Chr.10: 125852879-125852879 on Build GRCh38
rs747199233 Chr.10: 125852586-125852586 on Build GRCh38
rs905742998 Chr.10: 125852844-125852844 on Build GRCh38
rs751613860 Chr.10: 125852686-125852686 on Build GRCh38
rs1035900904 Chr.10: 125852856-125852856 on Build GRCh38
rs772213357 Chr.10: 125852487-125852487 on Build GRCh38
rs942263596 Chr.10: 125852651-125852651 on Build GRCh38
rs543883941 Chr.10: 125852599-125852599 on Build GRCh38
rs769250866 Chr.10: 125852761-125852761 on Build GRCh38
rs146463146 Chr.10: 125852542-125852542 on Build GRCh38
rs751246260 Chr.10: 125852533-125852533 on Build GRCh38
rs748237705 Chr.10: 125852476-125852476 on Build GRCh38
rs560312431 Chr.10: 125852843-125852843 on Build GRCh38
rs888005894 Chr.10: 125852644-125852644 on Build GRCh38
rs758264288 Chr.10: 125852607-125852607 on Build GRCh38
rs140992376 Chr.10: 125852903-125852903 on Build GRCh38
rs370666697 Chr.10: 125852649-125852649 on Build GRCh38
rs141841599 Chr.10: 125852631-125852631 on Build GRCh38
rs981974319 Chr.10: 125852554-125852554 on Build GRCh38
rs777280901 Chr.10: 125852620-125852620 on Build GRCh38
rs111356244 Chr.10: 125852918-125852918 on Build GRCh38
rs371312612 Chr.10: 125852563-125852563 on Build GRCh38
rs753053597 Chr.10: 125852657-125852658 on Build GRCh38
rs947327431 Chr.10: 125852775-125852775 on Build GRCh38
rs773451806 Chr.10: 125852934-125852934 on Build GRCh38
rs759292712 Chr.10: 125852968-125852968 on Build GRCh38
rs747865883 Chr.10: 125852482-125852482 on Build GRCh38
rs367911803 Chr.10: 125852609-125852609 on Build GRCh38
rs944227273 Chr.10: 125852677-125852677 on Build GRCh38
rs182575602 Chr.10: 125852774-125852774 on Build GRCh38
rs992028017 Chr.10: 125852936-125852936 on Build GRCh38
rs766010296 Chr.10: 125852545-125852547 on Build GRCh38
rs368990177 Chr.10: 125852867-125852867 on Build GRCh38
rs760048971 Chr.10: 125852514-125852514 on Build GRCh38
rs768262026 Chr.10: 125852622-125852622 on Build GRCh38
rs549243975 Chr.10: 125852615-125852615 on Build GRCh38
rs965989918 Chr.10: 125852902-125852902 on Build GRCh38
rs775082542 Chr.10: 125852518-125852518 on Build GRCh38
rs761425156 Chr.10: 125852538-125852538 on Build GRCh38
rs760379407 Chr.10: 125852658-125852658 on Build GRCh38
rs148099794 Chr.10: 125852598-125852598 on Build GRCh38
rs896929616 Chr.10: 125852860-125852860 on Build GRCh38
rs905694382 Chr.10: 125852500-125852500 on Build GRCh38
rs776899102 Chr.10: 125852504-125852504 on Build GRCh38
rs1002713974 Chr.10: 125852855-125852855 on Build GRCh38
rs747287125 Chr.10: 125852502-125852502 on Build GRCh38
rs755766924 Chr.10: 125852516-125852522 on Build GRCh38
rs186738718 Chr.10: 125852493-125852493 on Build GRCh38
rs1026645598 Chr.10: 125852874-125852874 on Build GRCh38
rs909389265 Chr.10: 125852966-125852966 on Build GRCh38
rs763765334 Chr.10: 125852523-125852523 on Build GRCh38
rs893794024 Chr.10: 125852743-125852743 on Build GRCh38

View Full Product Details