Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1630061-1630571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP5-5
Entrez Gene ID
439915
Gene Name
keratin associated protein 5-5
Gene Aliases
KRTAP5-11, KRTAP5.5
UniGene
Hs.653467
Gene Chromosome Location
Chr.11: 1629803-1630930 on Build GRCh38
Transcripts
NM_001001480.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5787158 c.495C>T p.C165C Chr.11: 1630335-1630335 on Build GRCh38
7374517 c.701A>T p.Q234L Chr.11: 1630541-1630541 on Build GRCh38
1293353 c.498C>T p.S166S Chr.11: 1630338-1630338 on Build GRCh38
4145588 c.576C>T p.Y192Y Chr.11: 1630416-1630416 on Build GRCh38
321323 c.553C>A p.Q185K Chr.11: 1630393-1630393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754409154 Chr.11: 1630199-1630200 on Build GRCh38
rs776684652 Chr.11: 1630147-1630147 on Build GRCh38
rs569811286 Chr.11: 1630182-1630182 on Build GRCh38
rs879102866 Chr.11: 1630376-1630376 on Build GRCh38
rs751875530 Chr.11: 1630365-1630368 on Build GRCh38
rs758128143 Chr.11: 1630314-1630314 on Build GRCh38
rs199980437 Chr.11: 1630427-1630427 on Build GRCh38
rs776952241 Chr.11: 1630244-1630267 on Build GRCh38
rs991068877 Chr.11: 1630245-1630245 on Build GRCh38
rs372594896 Chr.11: 1630498-1630498 on Build GRCh38
rs200864103 Chr.11: 1630223-1630223 on Build GRCh38
rs750436776 Chr.11: 1630133-1630133 on Build GRCh38
rs773439922 Chr.11: 1630346-1630346 on Build GRCh38
rs917504603 Chr.11: 1630149-1630149 on Build GRCh38
rs377559006 Chr.11: 1630514-1630514 on Build GRCh38
rs372072203 Chr.11: 1630137-1630137 on Build GRCh38
rs766608240 Chr.11: 1630356-1630385 on Build GRCh38
rs199686073 Chr.11: 1630392-1630392 on Build GRCh38
rs750531750 Chr.11: 1630460-1630461 on Build GRCh38
rs1040301547 Chr.11: 1630451-1630451 on Build GRCh38
rs747067865 Chr.11: 1630211-1630213 on Build GRCh38
rs765654566 Chr.11: 1630202-1630229 on Build GRCh38
rs926063975 Chr.11: 1630353-1630353 on Build GRCh38
rs767651575 Chr.11: 1630343-1630343 on Build GRCh38
rs751231399 Chr.11: 1630475-1630475 on Build GRCh38
rs766171125 Chr.11: 1630358-1630358 on Build GRCh38
rs747778624 Chr.11: 1630154-1630154 on Build GRCh38
rs373726678 Chr.11: 1630362-1630362 on Build GRCh38
rs756932573 Chr.11: 1630476-1630476 on Build GRCh38
rs375782833 Chr.11: 1630526-1630526 on Build GRCh38
rs754960909 Chr.11: 1630363-1630363 on Build GRCh38
rs756206513 Chr.11: 1630464-1630465 on Build GRCh38
rs776201445 Chr.11: 1630411-1630413 on Build GRCh38
rs74396270 Chr.11: 1630385-1630385 on Build GRCh38
rs559627884 Chr.11: 1630407-1630407 on Build GRCh38
rs755291491 Chr.11: 1630494-1630494 on Build GRCh38
rs774414710 Chr.11: 1630334-1630334 on Build GRCh38
rs759345977 Chr.11: 1630449-1630449 on Build GRCh38
rs771479002 Chr.11: 1630159-1630159 on Build GRCh38
rs774048905 Chr.11: 1630218-1630248 on Build GRCh38
rs755885136 Chr.11: 1630297-1630297 on Build GRCh38
rs552263240 Chr.11: 1630199-1630199 on Build GRCh38
rs777243096 Chr.11: 1630097-1630097 on Build GRCh38
rs749070033 Chr.11: 1630384-1630411 on Build GRCh38
rs759900883 Chr.11: 1630400-1630428 on Build GRCh38
rs80015637 Chr.11: 1630181-1630181 on Build GRCh38
rs779433206 Chr.11: 1630497-1630497 on Build GRCh38
rs750195884 Chr.11: 1630219-1630219 on Build GRCh38
rs764129117 Chr.11: 1630194-1630194 on Build GRCh38
rs760937140 Chr.11: 1630438-1630438 on Build GRCh38
rs769674711 Chr.11: 1630177-1630182 on Build GRCh38
rs201337556 Chr.11: 1630252-1630252 on Build GRCh38
rs772157333 Chr.11: 1630423-1630423 on Build GRCh38
rs780760925 Chr.11: 1630481-1630481 on Build GRCh38
rs778744373 Chr.11: 1630152-1630152 on Build GRCh38
rs537457356 Chr.11: 1630191-1630191 on Build GRCh38
rs755275335 Chr.11: 1630201-1630229 on Build GRCh38
rs762619274 Chr.11: 1630188-1630188 on Build GRCh38
rs377326691 Chr.11: 1630382-1630382 on Build GRCh38
rs754218603 Chr.11: 1630487-1630487 on Build GRCh38
rs147101544 Chr.11: 1630325-1630325 on Build GRCh38
rs757429622 Chr.11: 1630213-1630213 on Build GRCh38
rs767417591 Chr.11: 1630218-1630218 on Build GRCh38
rs267602795 Chr.11: 1630262-1630262 on Build GRCh38
rs772782123 Chr.11: 1630397-1630423 on Build GRCh38
rs760424500 Chr.11: 1630351-1630351 on Build GRCh38
rs765288570 Chr.11: 1630455-1630455 on Build GRCh38
rs773857331 Chr.11: 1630190-1630215 on Build GRCh38
rs1027710141 Chr.11: 1630531-1630531 on Build GRCh38
rs201941213 Chr.11: 1630205-1630205 on Build GRCh38
rs756005403 Chr.11: 1630134-1630134 on Build GRCh38
rs61867582 Chr.11: 1630320-1630320 on Build GRCh38
rs748765927 Chr.11: 1630139-1630139 on Build GRCh38
rs745870353 Chr.11: 1630215-1630241 on Build GRCh38
rs766151666 Chr.11: 1630439-1630439 on Build GRCh38
rs751741965 Chr.11: 1630117-1630117 on Build GRCh38
rs559565463 Chr.11: 1630131-1630131 on Build GRCh38
rs764605776 Chr.11: 1630414-1630415 on Build GRCh38
rs565769824 Chr.11: 1630216-1630216 on Build GRCh38
rs760118517 Chr.11: 1630247-1630252 on Build GRCh38
rs780456065 Chr.11: 1630391-1630391 on Build GRCh38
rs765424276 Chr.11: 1630110-1630110 on Build GRCh38
rs193086302 Chr.11: 1630317-1630317 on Build GRCh38
rs761113610 Chr.11: 1630192-1630215 on Build GRCh38
rs747858698 Chr.11: 1630390-1630391 on Build GRCh38
rs76164438 Chr.11: 1630193-1630193 on Build GRCh38
rs781358661 Chr.11: 1630380-1630395 on Build GRCh38
rs753122575 Chr.11: 1630358-1630385 on Build GRCh38
rs763382896 Chr.11: 1630413-1630414 on Build GRCh38
rs370405113 Chr.11: 1630468-1630468 on Build GRCh38
rs761281204 Chr.11: 1630222-1630245 on Build GRCh38
rs777903431 Chr.11: 1630500-1630500 on Build GRCh38
rs565013524 Chr.11: 1630113-1630113 on Build GRCh38
rs71025765 Chr.11: 1630415-1630444 on Build GRCh38
rs769159514 Chr.11: 1630411-1630411 on Build GRCh38
rs543002232 Chr.11: 1630085-1630085 on Build GRCh38
rs768785002 Chr.11: 1630329-1630329 on Build GRCh38
rs75391124 Chr.11: 1630122-1630122 on Build GRCh38
rs746644909 Chr.11: 1630390-1630390 on Build GRCh38
rs535632539 Chr.11: 1630288-1630288 on Build GRCh38
rs201404581 Chr.11: 1630536-1630536 on Build GRCh38
rs201208697 Chr.11: 1630289-1630297 on Build GRCh38
rs77039648 Chr.11: 1630415-1630415 on Build GRCh38
rs61867581 Chr.11: 1630310-1630310 on Build GRCh38
rs774985887 Chr.11: 1630188-1630218 on Build GRCh38
rs759947752 Chr.11: 1630109-1630109 on Build GRCh38
rs376896030 Chr.11: 1630328-1630328 on Build GRCh38
rs762129293 Chr.11: 1630445-1630447 on Build GRCh38
rs757087387 Chr.11: 1630121-1630121 on Build GRCh38
rs878940471 Chr.11: 1630377-1630377 on Build GRCh38
rs563625149 Chr.11: 1630471-1630471 on Build GRCh38
rs765450166 Chr.11: 1630355-1630356 on Build GRCh38
rs758695857 Chr.11: 1630208-1630236 on Build GRCh38
rs768310752 Chr.11: 1630217-1630222 on Build GRCh38
rs200368281 Chr.11: 1630283-1630283 on Build GRCh38
rs780964318 Chr.11: 1630214-1630241 on Build GRCh38
rs751087858 Chr.11: 1630540-1630540 on Build GRCh38
rs200756510 Chr.11: 1630395-1630395 on Build GRCh38
rs111318317 Chr.11: 1630422-1630422 on Build GRCh38
rs369404931 Chr.11: 1630254-1630254 on Build GRCh38
rs761818697 Chr.11: 1630337-1630337 on Build GRCh38
rs576867883 Chr.11: 1630384-1630413 on Build GRCh38
rs375860990 Chr.11: 1630441-1630441 on Build GRCh38
rs777937937 Chr.11: 1630210-1630215 on Build GRCh38
rs200516743 Chr.11: 1630380-1630380 on Build GRCh38
rs757266560 Chr.11: 1630370-1630395 on Build GRCh38
rs771441561 Chr.11: 1630393-1630395 on Build GRCh38
rs547644527 Chr.11: 1630129-1630129 on Build GRCh38
rs751786174 Chr.11: 1630416-1630417 on Build GRCh38
rs767430798 Chr.11: 1630446-1630472 on Build GRCh38
rs534780728 Chr.11: 1630207-1630207 on Build GRCh38
rs763556754 Chr.11: 1630189-1630217 on Build GRCh38
rs4752771 Chr.11: 1630413-1630413 on Build GRCh38
rs76143925 Chr.11: 1630383-1630383 on Build GRCh38
rs201456157 Chr.11: 1630302-1630302 on Build GRCh38
rs375846662 Chr.11: 1630461-1630461 on Build GRCh38
rs77602976 Chr.11: 1630169-1630169 on Build GRCh38
rs771198452 Chr.11: 1630510-1630510 on Build GRCh38
rs771001169 Chr.11: 1630088-1630102 on Build GRCh38
rs138562325 Chr.11: 1630365-1630365 on Build GRCh38
rs767102941 Chr.11: 1630237-1630242 on Build GRCh38
rs756083923 Chr.11: 1630381-1630382 on Build GRCh38
rs552577810 Chr.11: 1630527-1630527 on Build GRCh38
rs748873768 Chr.11: 1630217-1630240 on Build GRCh38
rs201911893 Chr.11: 1630410-1630410 on Build GRCh38
rs553180611 Chr.11: 1630240-1630240 on Build GRCh38
rs779885115 Chr.11: 1630298-1630298 on Build GRCh38
rs769429915 Chr.11: 1630215-1630243 on Build GRCh38
rs776572193 Chr.11: 1630440-1630440 on Build GRCh38
rs764403327 Chr.11: 1630364-1630377 on Build GRCh38
rs779963378 Chr.11: 1630466-1630467 on Build GRCh38
rs759249591 Chr.11: 1630177-1630177 on Build GRCh38
rs746993398 Chr.11: 1630506-1630506 on Build GRCh38
rs201598858 Chr.11: 1630472-1630472 on Build GRCh38
rs766783534 Chr.11: 1630198-1630198 on Build GRCh38
rs183750160 Chr.11: 1630530-1630530 on Build GRCh38
rs773970029 Chr.11: 1630389-1630415 on Build GRCh38
rs770407985 Chr.11: 1630408-1630422 on Build GRCh38
rs775297744 Chr.11: 1630217-1630220 on Build GRCh38
rs768506323 Chr.11: 1630385-1630386 on Build GRCh38
rs752926195 Chr.11: 1630207-1630208 on Build GRCh38
rs79933019 Chr.11: 1630151-1630151 on Build GRCh38
rs758887815 Chr.11: 1630361-1630364 on Build GRCh38
rs776016098 Chr.11: 1630192-1630192 on Build GRCh38
rs745755489 Chr.11: 1630380-1630381 on Build GRCh38
rs201822473 Chr.11: 1630229-1630229 on Build GRCh38

View Full Product Details